Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 51284628-51285122 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIN2
Entrez Gene ID
51411
Gene Name
bridging integrator 2
Gene Aliases
BRAP-1
UniGene
Hs.14770
Gene Chromosome Location
Chr.12: 51281038-51324680 on Build GRCh38
Transcripts
NM_001290007.1 NM_001290008.1 NM_001290009.1 NM_016293.3 XM_005268957.3 XM_005268958.3 XM_017019422.1

SNP Detail

SNP ID Chromosome Location
rs891449652 Chr.12: 51284990-51284990 on Build GRCh38
rs927985569 Chr.12: 51284956-51284956 on Build GRCh38
rs765843050 Chr.12: 51284727-51284727 on Build GRCh38
rs893638268 Chr.12: 51284656-51284656 on Build GRCh38
rs961439155 Chr.12: 51284841-51284841 on Build GRCh38
rs370435296 Chr.12: 51284882-51284882 on Build GRCh38
rs141365457 Chr.12: 51284731-51284731 on Build GRCh38
rs772966112 Chr.12: 51284818-51284818 on Build GRCh38
rs370534497 Chr.12: 51284809-51284809 on Build GRCh38
rs774798353 Chr.12: 51284697-51284697 on Build GRCh38
rs768978185 Chr.12: 51284792-51284792 on Build GRCh38
rs767279243 Chr.12: 51284945-51284945 on Build GRCh38
rs1043120938 Chr.12: 51285095-51285095 on Build GRCh38
rs371833676 Chr.12: 51284822-51284822 on Build GRCh38
rs759094699 Chr.12: 51284827-51284827 on Build GRCh38
rs781380098 Chr.12: 51284751-51284751 on Build GRCh38
rs769825439 Chr.12: 51284689-51284689 on Build GRCh38
rs982626969 Chr.12: 51284928-51284928 on Build GRCh38
rs1010712966 Chr.12: 51284674-51284674 on Build GRCh38
rs756187749 Chr.12: 51284769-51284769 on Build GRCh38
rs775949155 Chr.12: 51284826-51284826 on Build GRCh38
rs995895924 Chr.12: 51285101-51285101 on Build GRCh38
rs773054199 Chr.12: 51284712-51284712 on Build GRCh38
rs996930387 Chr.12: 51284736-51284736 on Build GRCh38
rs529539044 Chr.12: 51284974-51284974 on Build GRCh38
rs762272163 Chr.12: 51284700-51284700 on Build GRCh38
rs1053083779 Chr.12: 51284979-51284979 on Build GRCh38
rs1053169157 Chr.12: 51284667-51284667 on Build GRCh38
rs964758067 Chr.12: 51284772-51284772 on Build GRCh38
rs946634628 Chr.12: 51285068-51285068 on Build GRCh38
rs765158039 Chr.12: 51284831-51284831 on Build GRCh38
rs749398113 Chr.12: 51284791-51284791 on Build GRCh38
rs750694548 Chr.12: 51284763-51284763 on Build GRCh38
rs542915602 Chr.12: 51284994-51284994 on Build GRCh38
rs752425161 Chr.12: 51284745-51284745 on Build GRCh38
rs575269854 Chr.12: 51284842-51284842 on Build GRCh38
rs928463354 Chr.12: 51284968-51284968 on Build GRCh38
rs552960490 Chr.12: 51284729-51284729 on Build GRCh38
rs754579070 Chr.12: 51284931-51284931 on Build GRCh38
rs781400312 Chr.12: 51284680-51284680 on Build GRCh38
rs947041880 Chr.12: 51285091-51285091 on Build GRCh38
rs938029395 Chr.12: 51284963-51284963 on Build GRCh38
rs893690866 Chr.12: 51285054-51285054 on Build GRCh38
rs376240258 Chr.12: 51284691-51284691 on Build GRCh38
rs919988615 Chr.12: 51284878-51284878 on Build GRCh38
rs765176485 Chr.12: 51284739-51284739 on Build GRCh38
rs760206717 Chr.12: 51284713-51284713 on Build GRCh38
rs199543314 Chr.12: 51284719-51284719 on Build GRCh38
rs368044843 Chr.12: 51284820-51284820 on Build GRCh38
rs908714667 Chr.12: 51284848-51284848 on Build GRCh38
rs367655945 Chr.12: 51284693-51284693 on Build GRCh38
rs546956251 Chr.12: 51285100-51285100 on Build GRCh38
rs563580728 Chr.12: 51284933-51284933 on Build GRCh38
rs758258873 Chr.12: 51284747-51284747 on Build GRCh38
rs779958734 Chr.12: 51284780-51284780 on Build GRCh38
rs769637875 Chr.12: 51284713-51284714 on Build GRCh38
rs772610274 Chr.12: 51284811-51284811 on Build GRCh38
rs948783507 Chr.12: 51284926-51284926 on Build GRCh38
rs745946942 Chr.12: 51284681-51284681 on Build GRCh38
rs767273976 Chr.12: 51284705-51284705 on Build GRCh38
rs866046394 Chr.12: 51284718-51284718 on Build GRCh38
rs543979872 Chr.12: 51284906-51284906 on Build GRCh38
rs757416158 Chr.12: 51284678-51284678 on Build GRCh38
rs769237492 Chr.12: 51284696-51284696 on Build GRCh38
rs190671339 Chr.12: 51284799-51284799 on Build GRCh38
rs952967863 Chr.12: 51284839-51284839 on Build GRCh38
rs373966540 Chr.12: 51284813-51284813 on Build GRCh38
rs752522710 Chr.12: 51285055-51285055 on Build GRCh38
rs747963940 Chr.12: 51284673-51284673 on Build GRCh38

View Full Product Details