Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52286713-52287223 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT81
Entrez Gene ID
3887
Gene Name
keratin 81
Gene Aliases
HB1, Hb-1, KRTHB1, MLN137, ghHkb1, hHAKB2-1
UniGene
Hs.658118
Gene Chromosome Location
Chr.12: 52285913-52291515 on Build GRCh38
Transcripts
NM_002281.3 NM_001320198.1 XM_005268866.4 XM_017019296.1

Gene Symbol
KRT86
Entrez Gene ID
3892
Gene Name
keratin 86
Gene Aliases
HB6, Hb1, K86, KRTHB1, KRTHB6, MNX
UniGene
Hs.278658
Gene Chromosome Location
Chr.12: 52270507-52309163 on Build GRCh38
Transcripts
NM_002281.3 NM_001320198.1 XM_005268866.4 XM_017019296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
162065 c.1237G>A p.E413K Chr.12: 52287112-52287112 on Build GRCh38
5824363 c.1268C>A p.A423D Chr.12: 52286802-52286802 on Build GRCh38
400158 c.1161G>T p.R387S Chr.12: 52287188-52287188 on Build GRCh38
6996611 c.1258G>T p.G420C Chr.12: 52286812-52286812 on Build GRCh38
296595 c.1140G>A p.Q380Q Chr.12: 52287209-52287209 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751051005 Chr.12: 52287157-52287157 on Build GRCh38
rs113097323 Chr.12: 52286998-52286999 on Build GRCh38
rs749590869 Chr.12: 52286777-52286777 on Build GRCh38
rs746851539 Chr.12: 52286749-52286749 on Build GRCh38
rs756945756 Chr.12: 52286762-52286762 on Build GRCh38
rs764096477 Chr.12: 52286870-52286870 on Build GRCh38
rs934550315 Chr.12: 52286747-52286747 on Build GRCh38
rs57419521 Chr.12: 52287112-52287112 on Build GRCh38
rs370632532 Chr.12: 52286861-52286861 on Build GRCh38
rs777027305 Chr.12: 52287129-52287129 on Build GRCh38
rs534459175 Chr.12: 52287097-52287097 on Build GRCh38
rs998393339 Chr.12: 52286869-52286869 on Build GRCh38
rs991354798 Chr.12: 52286996-52286996 on Build GRCh38
rs1005543455 Chr.12: 52286969-52286969 on Build GRCh38
rs200188584 Chr.12: 52287069-52287069 on Build GRCh38
rs368237680 Chr.12: 52287101-52287101 on Build GRCh38
rs752916490 Chr.12: 52287201-52287201 on Build GRCh38
rs750276983 Chr.12: 52287171-52287171 on Build GRCh38
rs772669761 Chr.12: 52286812-52286812 on Build GRCh38
rs980728119 Chr.12: 52286936-52286936 on Build GRCh38
rs761110090 Chr.12: 52286825-52286825 on Build GRCh38
rs766783392 Chr.12: 52286847-52286847 on Build GRCh38
rs759224738 Chr.12: 52286858-52286858 on Build GRCh38
rs961783554 Chr.12: 52286793-52286793 on Build GRCh38
rs755276088 Chr.12: 52287085-52287085 on Build GRCh38
rs774624443 Chr.12: 52286785-52286785 on Build GRCh38
rs201447608 Chr.12: 52287126-52287126 on Build GRCh38
rs56821304 Chr.12: 52287145-52287145 on Build GRCh38
rs760521872 Chr.12: 52286811-52286811 on Build GRCh38
rs540172456 Chr.12: 52286994-52286994 on Build GRCh38
rs376393956 Chr.12: 52286773-52286773 on Build GRCh38
rs892582153 Chr.12: 52286999-52286999 on Build GRCh38
rs111382015 Chr.12: 52287122-52287122 on Build GRCh38
rs939356400 Chr.12: 52286989-52286991 on Build GRCh38
rs750223770 Chr.12: 52287073-52287073 on Build GRCh38
rs934290936 Chr.12: 52287091-52287091 on Build GRCh38
rs59448276 Chr.12: 52286982-52286982 on Build GRCh38
rs757215183 Chr.12: 52287066-52287066 on Build GRCh38
rs761893737 Chr.12: 52287161-52287161 on Build GRCh38
rs751509447 Chr.12: 52287055-52287055 on Build GRCh38
rs374751440 Chr.12: 52287088-52287088 on Build GRCh38
rs1016878787 Chr.12: 52286971-52286971 on Build GRCh38
rs771393943 Chr.12: 52287127-52287127 on Build GRCh38
rs141521550 Chr.12: 52287140-52287140 on Build GRCh38
rs1049840733 Chr.12: 52286879-52286879 on Build GRCh38
rs758407894 Chr.12: 52286742-52286742 on Build GRCh38
rs773529374 Chr.12: 52286813-52286813 on Build GRCh38
rs768970768 Chr.12: 52286784-52286784 on Build GRCh38
rs961632486 Chr.12: 52286973-52286973 on Build GRCh38
rs551405099 Chr.12: 52286981-52286981 on Build GRCh38
rs901024465 Chr.12: 52286835-52286835 on Build GRCh38
rs759844211 Chr.12: 52287139-52287139 on Build GRCh38
rs1014690630 Chr.12: 52286748-52286748 on Build GRCh38
rs74095619 Chr.12: 52287005-52287005 on Build GRCh38
rs771983295 Chr.12: 52287113-52287113 on Build GRCh38
rs1044307771 Chr.12: 52287008-52287008 on Build GRCh38
rs762328799 Chr.12: 52286865-52286865 on Build GRCh38
rs763579563 Chr.12: 52287155-52287155 on Build GRCh38
rs779295867 Chr.12: 52287198-52287198 on Build GRCh38
rs889867156 Chr.12: 52286887-52286887 on Build GRCh38
rs187305858 Chr.12: 52286951-52286951 on Build GRCh38
rs767515484 Chr.12: 52287168-52287168 on Build GRCh38
rs762654620 Chr.12: 52287149-52287149 on Build GRCh38
rs777522022 Chr.12: 52286745-52286745 on Build GRCh38
rs777990155 Chr.12: 52286855-52286855 on Build GRCh38
rs748130319 Chr.12: 52286794-52286794 on Build GRCh38
rs775624280 Chr.12: 52286785-52286788 on Build GRCh38
rs764846224 Chr.12: 52286860-52286860 on Build GRCh38
rs570237300 Chr.12: 52287010-52287010 on Build GRCh38
rs978452210 Chr.12: 52287032-52287032 on Build GRCh38
rs776972178 Chr.12: 52286857-52286857 on Build GRCh38
rs372998713 Chr.12: 52287180-52287180 on Build GRCh38

View Full Product Details