Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52362993-52363536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT85
Entrez Gene ID
3891
Gene Name
keratin 85
Gene Aliases
ECTD4, HB5, Hb-5, K85, KRTHB5, hHb5
UniGene
Hs.182507
Gene Chromosome Location
Chr.12: 52360006-52367525 on Build GRCh38
Transcripts
NM_001300810.1 NM_002283.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4822786 c.913C>T p.R305C Chr.12: 52363284-52363284 on Build GRCh38
6840840 c.157C>T p.R53C Chr.12: 52363404-52363404 on Build GRCh38
6698308 c.268G>A p.V90I Chr.12: 52363293-52363293 on Build GRCh38
3384452 c.951+8G>T p.? Chr.12: 52363238-52363238 on Build GRCh38
6840839 c.793C>T p.R265C Chr.12: 52363404-52363404 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs529513894 Chr.12: 52363283-52363283 on Build GRCh38
rs10876280 Chr.12: 52363129-52363129 on Build GRCh38
rs926546119 Chr.12: 52363099-52363099 on Build GRCh38
rs951202106 Chr.12: 52363500-52363500 on Build GRCh38
rs780063613 Chr.12: 52363255-52363255 on Build GRCh38
rs61731778 Chr.12: 52363273-52363273 on Build GRCh38
rs549707125 Chr.12: 52363284-52363284 on Build GRCh38
rs371939591 Chr.12: 52363397-52363397 on Build GRCh38
rs370871372 Chr.12: 52363017-52363017 on Build GRCh38
rs373727813 Chr.12: 52363350-52363350 on Build GRCh38
rs769092127 Chr.12: 52363021-52363021 on Build GRCh38
rs779303633 Chr.12: 52363325-52363325 on Build GRCh38
rs779667905 Chr.12: 52363459-52363459 on Build GRCh38
rs778330987 Chr.12: 52363346-52363346 on Build GRCh38
rs749721561 Chr.12: 52363020-52363020 on Build GRCh38
rs779006060 Chr.12: 52363410-52363410 on Build GRCh38
rs906348313 Chr.12: 52363154-52363154 on Build GRCh38
rs1030732567 Chr.12: 52363039-52363039 on Build GRCh38
rs1056315490 Chr.12: 52363483-52363483 on Build GRCh38
rs147392729 Chr.12: 52363277-52363277 on Build GRCh38
rs376584235 Chr.12: 52363461-52363461 on Build GRCh38
rs753936439 Chr.12: 52363307-52363307 on Build GRCh38
rs1029708661 Chr.12: 52363507-52363507 on Build GRCh38
rs572711402 Chr.12: 52363064-52363064 on Build GRCh38
rs375124943 Chr.12: 52363253-52363253 on Build GRCh38
rs752801617 Chr.12: 52363435-52363435 on Build GRCh38
rs201510373 Chr.12: 52363328-52363328 on Build GRCh38
rs377226861 Chr.12: 52363350-52363351 on Build GRCh38
rs956807383 Chr.12: 52363187-52363187 on Build GRCh38
rs759440939 Chr.12: 52363220-52363220 on Build GRCh38
rs745380009 Chr.12: 52363360-52363360 on Build GRCh38
rs745490028 Chr.12: 52363452-52363452 on Build GRCh38
rs368606625 Chr.12: 52363213-52363213 on Build GRCh38
rs780630642 Chr.12: 52363269-52363269 on Build GRCh38
rs758196560 Chr.12: 52363245-52363245 on Build GRCh38
rs749367463 Chr.12: 52363264-52363264 on Build GRCh38
rs779471419 Chr.12: 52363272-52363272 on Build GRCh38
rs376347432 Chr.12: 52363404-52363404 on Build GRCh38
rs760274769 Chr.12: 52363212-52363212 on Build GRCh38
rs778418922 Chr.12: 52363442-52363442 on Build GRCh38
rs776043030 Chr.12: 52363280-52363280 on Build GRCh38
rs1052266070 Chr.12: 52363446-52363446 on Build GRCh38
rs537447052 Chr.12: 52363499-52363499 on Build GRCh38
rs757635544 Chr.12: 52363448-52363448 on Build GRCh38
rs773004949 Chr.12: 52363275-52363275 on Build GRCh38
rs929696562 Chr.12: 52363192-52363192 on Build GRCh38
rs1003384251 Chr.12: 52363162-52363162 on Build GRCh38
rs780710780 Chr.12: 52363449-52363449 on Build GRCh38
rs1022407037 Chr.12: 52363495-52363495 on Build GRCh38
rs774630090 Chr.12: 52363024-52363024 on Build GRCh38
rs769205477 Chr.12: 52363457-52363457 on Build GRCh38
rs139583707 Chr.12: 52363296-52363296 on Build GRCh38
rs769278493 Chr.12: 52363371-52363371 on Build GRCh38
rs769277149 Chr.12: 52363018-52363019 on Build GRCh38
rs752499748 Chr.12: 52363239-52363239 on Build GRCh38
rs767737636 Chr.12: 52363246-52363246 on Build GRCh38
rs1037363880 Chr.12: 52363098-52363098 on Build GRCh38
rs756221275 Chr.12: 52363254-52363254 on Build GRCh38
rs139756703 Chr.12: 52363403-52363403 on Build GRCh38
rs375295365 Chr.12: 52363210-52363210 on Build GRCh38
rs765320745 Chr.12: 52363433-52363433 on Build GRCh38
rs747345900 Chr.12: 52363347-52363347 on Build GRCh38
rs1023527814 Chr.12: 52363142-52363142 on Build GRCh38
rs372918365 Chr.12: 52363297-52363297 on Build GRCh38
rs755483972 Chr.12: 52363373-52363373 on Build GRCh38
rs780883191 Chr.12: 52363351-52363351 on Build GRCh38
rs867020277 Chr.12: 52363177-52363177 on Build GRCh38
rs771918724 Chr.12: 52363205-52363205 on Build GRCh38
rs776651710 Chr.12: 52363217-52363217 on Build GRCh38
rs1056168827 Chr.12: 52363209-52363209 on Build GRCh38
rs199836290 Chr.12: 52363015-52363015 on Build GRCh38
rs1031462884 Chr.12: 52363173-52363173 on Build GRCh38
rs762237390 Chr.12: 52363453-52363454 on Build GRCh38
rs765187925 Chr.12: 52363228-52363228 on Build GRCh38
rs755074018 Chr.12: 52363320-52363320 on Build GRCh38
rs1044928299 Chr.12: 52363393-52363393 on Build GRCh38
rs373479477 Chr.12: 52363402-52363402 on Build GRCh38
rs146373501 Chr.12: 52363278-52363278 on Build GRCh38
rs780540192 Chr.12: 52363019-52363019 on Build GRCh38
rs114878563 Chr.12: 52363119-52363119 on Build GRCh38
rs919419022 Chr.12: 52363480-52363480 on Build GRCh38
rs777330819 Chr.12: 52363432-52363433 on Build GRCh38
rs759539209 Chr.12: 52363413-52363413 on Build GRCh38
rs758865856 Chr.12: 52363335-52363335 on Build GRCh38
rs557126652 Chr.12: 52363508-52363508 on Build GRCh38
rs754035337 Chr.12: 52363405-52363405 on Build GRCh38
rs752027000 Chr.12: 52363447-52363447 on Build GRCh38
rs918353485 Chr.12: 52363145-52363145 on Build GRCh38
rs146410564 Chr.12: 52363418-52363419 on Build GRCh38
rs375780859 Chr.12: 52363050-52363050 on Build GRCh38
rs768650216 Chr.12: 52363462-52363462 on Build GRCh38
rs767799696 Chr.12: 52363104-52363104 on Build GRCh38
rs768562539 Chr.12: 52363401-52363401 on Build GRCh38
rs763832331 Chr.12: 52363294-52363294 on Build GRCh38
rs759013876 Chr.12: 52363436-52363436 on Build GRCh38

View Full Product Details