Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52393867-52394367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT82
Entrez Gene ID
3888
Gene Name
keratin 82
Gene Aliases
HB2, Hb-2, KRTHB2
UniGene
Hs.134640
Gene Chromosome Location
Chr.12: 52393951-52406392 on Build GRCh38
Transcripts
NM_033033.3

SNP Detail

SNP ID Chromosome Location
rs551415621 Chr.12: 52394032-52394032 on Build GRCh38
rs1019375694 Chr.12: 52393929-52393929 on Build GRCh38
rs970054738 Chr.12: 52394113-52394113 on Build GRCh38
rs774528664 Chr.12: 52394166-52394185 on Build GRCh38
rs1051865836 Chr.12: 52393945-52393945 on Build GRCh38
rs1016038388 Chr.12: 52393978-52393978 on Build GRCh38
rs1791636 Chr.12: 52394304-52394304 on Build GRCh38
rs73304592 Chr.12: 52394085-52394085 on Build GRCh38
rs528190421 Chr.12: 52393968-52393968 on Build GRCh38
rs1026683716 Chr.12: 52393982-52393982 on Build GRCh38
rs1732262 Chr.12: 52394228-52394228 on Build GRCh38
rs1023372767 Chr.12: 52394108-52394108 on Build GRCh38
rs11170112 Chr.12: 52394255-52394255 on Build GRCh38
rs751340015 Chr.12: 52394169-52394169 on Build GRCh38
rs571268565 Chr.12: 52394039-52394039 on Build GRCh38
rs888822514 Chr.12: 52393951-52393951 on Build GRCh38
rs79191015 Chr.12: 52393907-52393907 on Build GRCh38
rs116844944 Chr.12: 52394104-52394104 on Build GRCh38
rs952603444 Chr.12: 52394001-52394001 on Build GRCh38
rs988382103 Chr.12: 52394179-52394179 on Build GRCh38
rs1026890899 Chr.12: 52394311-52394311 on Build GRCh38
rs898916401 Chr.12: 52394291-52394291 on Build GRCh38
rs555702496 Chr.12: 52394180-52394180 on Build GRCh38
rs1007304365 Chr.12: 52393954-52393954 on Build GRCh38
rs933226641 Chr.12: 52394181-52394181 on Build GRCh38
rs773045650 Chr.12: 52394017-52394017 on Build GRCh38
rs899301992 Chr.12: 52393986-52393986 on Build GRCh38
rs577711132 Chr.12: 52394267-52394267 on Build GRCh38
rs1037420424 Chr.12: 52394286-52394286 on Build GRCh38
rs193274974 Chr.12: 52394159-52394159 on Build GRCh38
rs1008382810 Chr.12: 52393918-52393918 on Build GRCh38
rs978793176 Chr.12: 52394126-52394126 on Build GRCh38
rs1019744272 Chr.12: 52394223-52394223 on Build GRCh38
rs142315080 Chr.12: 52394146-52394146 on Build GRCh38
rs145761240 Chr.12: 52394188-52394188 on Build GRCh38
rs904572782 Chr.12: 52394132-52394132 on Build GRCh38
rs927104155 Chr.12: 52394022-52394022 on Build GRCh38
rs374059428 Chr.12: 52394241-52394243 on Build GRCh38
rs961311662 Chr.12: 52393943-52393943 on Build GRCh38
rs1009088766 Chr.12: 52394209-52394209 on Build GRCh38
rs542486569 Chr.12: 52393896-52393896 on Build GRCh38
rs879863442 Chr.12: 52394054-52394054 on Build GRCh38
rs943028369 Chr.12: 52394257-52394257 on Build GRCh38

View Full Product Details