Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 52821368-52821878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT79
Entrez Gene ID
338785
Gene Name
keratin 79
Gene Aliases
K6L, KRT6L
UniGene
Hs.711471
Gene Chromosome Location
Chr.12: 52821447-52834293 on Build GRCh38
Transcripts
NM_175834.2

SNP Detail

SNP ID Chromosome Location
rs907680107 Chr.12: 52821715-52821715 on Build GRCh38
rs748340452 Chr.12: 52821536-52821536 on Build GRCh38
rs971702776 Chr.12: 52821684-52821684 on Build GRCh38
rs947703475 Chr.12: 52821727-52821727 on Build GRCh38
rs528741340 Chr.12: 52821785-52821785 on Build GRCh38
rs189130045 Chr.12: 52821685-52821685 on Build GRCh38
rs374045629 Chr.12: 52821560-52821560 on Build GRCh38
rs750077364 Chr.12: 52821851-52821851 on Build GRCh38
rs1057190427 Chr.12: 52821458-52821458 on Build GRCh38
rs551116360 Chr.12: 52821606-52821606 on Build GRCh38
rs894651096 Chr.12: 52821477-52821477 on Build GRCh38
rs762968948 Chr.12: 52821831-52821831 on Build GRCh38
rs74096641 Chr.12: 52821423-52821423 on Build GRCh38
rs146367809 Chr.12: 52821439-52821439 on Build GRCh38
rs751046717 Chr.12: 52821833-52821833 on Build GRCh38
rs553250416 Chr.12: 52821587-52821587 on Build GRCh38
rs759334192 Chr.12: 52821822-52821822 on Build GRCh38
rs948897023 Chr.12: 52821761-52821761 on Build GRCh38
rs964386957 Chr.12: 52821644-52821644 on Build GRCh38
rs771129993 Chr.12: 52821714-52821714 on Build GRCh38
rs970537889 Chr.12: 52821601-52821601 on Build GRCh38
rs751446930 Chr.12: 52821827-52821835 on Build GRCh38
rs922747998 Chr.12: 52821402-52821402 on Build GRCh38
rs148927112 Chr.12: 52821719-52821719 on Build GRCh38
rs975997201 Chr.12: 52821614-52821614 on Build GRCh38
rs529928750 Chr.12: 52821810-52821810 on Build GRCh38
rs1037634533 Chr.12: 52821463-52821463 on Build GRCh38
rs375519220 Chr.12: 52821705-52821705 on Build GRCh38
rs953259123 Chr.12: 52821637-52821637 on Build GRCh38
rs1043645655 Chr.12: 52821767-52821767 on Build GRCh38
rs559558190 Chr.12: 52821731-52821731 on Build GRCh38
rs898797341 Chr.12: 52821478-52821478 on Build GRCh38
rs764190693 Chr.12: 52821832-52821832 on Build GRCh38
rs1040152052 Chr.12: 52821396-52821396 on Build GRCh38
rs755271063 Chr.12: 52821852-52821852 on Build GRCh38
rs367731035 Chr.12: 52821839-52821839 on Build GRCh38
rs879183329 Chr.12: 52821672-52821672 on Build GRCh38
rs530880727 Chr.12: 52821855-52821855 on Build GRCh38
rs552567898 Chr.12: 52821422-52821422 on Build GRCh38
rs775507912 Chr.12: 52821830-52821830 on Build GRCh38
rs535715654 Chr.12: 52821607-52821607 on Build GRCh38
rs756770047 Chr.12: 52821836-52821836 on Build GRCh38
rs940512481 Chr.12: 52821718-52821718 on Build GRCh38
rs564796188 Chr.12: 52821720-52821720 on Build GRCh38
rs989664672 Chr.12: 52821634-52821634 on Build GRCh38
rs773571250 Chr.12: 52821669-52821669 on Build GRCh38
rs372266125 Chr.12: 52821619-52821619 on Build GRCh38
rs1021729412 Chr.12: 52821542-52821542 on Build GRCh38
rs576825553 Chr.12: 52821709-52821709 on Build GRCh38
rs904627664 Chr.12: 52821543-52821543 on Build GRCh38
rs765127106 Chr.12: 52821825-52821825 on Build GRCh38
rs933138607 Chr.12: 52821416-52821416 on Build GRCh38
rs563550936 Chr.12: 52821819-52821819 on Build GRCh38
rs764706317 Chr.12: 52821740-52821740 on Build GRCh38
rs542078722 Chr.12: 52821818-52821818 on Build GRCh38
rs369498157 Chr.12: 52821656-52821656 on Build GRCh38
rs145734424 Chr.12: 52821680-52821680 on Build GRCh38
rs1023803110 Chr.12: 52821569-52821569 on Build GRCh38

View Full Product Details