Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 53973042-53973550 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOTAIR
Entrez Gene ID
100124700
Gene Name
HOX transcript antisense RNA
Gene Aliases
HOXAS, HOXC-AS4, HOXC11-AS1, NCRNA00072
UniGene
Hs.197076
Gene Chromosome Location
Chr.12: 53962308-53974956 on Build GRCh38
Transcripts
NR_047517.1 NM_014212.3

Gene Symbol
HOXC11
Entrez Gene ID
3227
Gene Name
homeobox C11
Gene Aliases
HOX3H
UniGene
Hs.127562
Gene Chromosome Location
Chr.12: 53973126-53976419 on Build GRCh38
Transcripts
NR_047517.1 NM_014212.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
389057 c.45C>A p.R15R Chr.12: 53973286-53973286 on Build GRCh38
7276651 c.84C>T p.C28C Chr.12: 53973325-53973325 on Build GRCh38
5964126 c.250C>T p.H84Y Chr.12: 53973491-53973491 on Build GRCh38
5841410 c.154_155delCC p.Q53fs*141 Chr.12: 53973395-53973396 on Build GRCh38
5841409 c.76G>T p.G26W Chr.12: 53973317-53973317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777615105 Chr.12: 53973247-53973247 on Build GRCh38
rs541827325 Chr.12: 53973109-53973109 on Build GRCh38
rs757003417 Chr.12: 53973335-53973335 on Build GRCh38
rs201200152 Chr.12: 53973529-53973529 on Build GRCh38
rs754972917 Chr.12: 53973227-53973227 on Build GRCh38
rs867894513 Chr.12: 53973241-53973241 on Build GRCh38
rs972524159 Chr.12: 53973363-53973363 on Build GRCh38
rs199765463 Chr.12: 53973315-53973315 on Build GRCh38
rs777910326 Chr.12: 53973328-53973328 on Build GRCh38
rs766936743 Chr.12: 53973212-53973212 on Build GRCh38
rs868032096 Chr.12: 53973410-53973410 on Build GRCh38
rs758550561 Chr.12: 53973324-53973324 on Build GRCh38
rs762569969 Chr.12: 53973290-53973290 on Build GRCh38
rs771851038 Chr.12: 53973494-53973494 on Build GRCh38
rs374141798 Chr.12: 53973224-53973224 on Build GRCh38
rs932159008 Chr.12: 53973143-53973143 on Build GRCh38
rs919338272 Chr.12: 53973072-53973072 on Build GRCh38
rs752730449 Chr.12: 53973427-53973427 on Build GRCh38
rs779994792 Chr.12: 53973466-53973466 on Build GRCh38
rs748775178 Chr.12: 53973381-53973381 on Build GRCh38
rs372496237 Chr.12: 53973211-53973211 on Build GRCh38
rs756007846 Chr.12: 53973464-53973464 on Build GRCh38
rs985878207 Chr.12: 53973156-53973156 on Build GRCh38
rs765711639 Chr.12: 53973422-53973422 on Build GRCh38
rs768321467 Chr.12: 53973473-53973473 on Build GRCh38
rs926578087 Chr.12: 53973457-53973457 on Build GRCh38
rs374695143 Chr.12: 53973426-53973426 on Build GRCh38
rs563309038 Chr.12: 53973183-53973183 on Build GRCh38
rs759883766 Chr.12: 53973509-53973509 on Build GRCh38
rs201521088 Chr.12: 53973253-53973253 on Build GRCh38
rs34028255 Chr.12: 53973259-53973259 on Build GRCh38
rs553000303 Chr.12: 53973094-53973094 on Build GRCh38
rs746825359 Chr.12: 53973332-53973332 on Build GRCh38
rs757448481 Chr.12: 53973450-53973450 on Build GRCh38
rs755493064 Chr.12: 53973311-53973311 on Build GRCh38
rs866318457 Chr.12: 53973313-53973313 on Build GRCh38
rs146084910 Chr.12: 53973298-53973298 on Build GRCh38
rs976335536 Chr.12: 53973119-53973119 on Build GRCh38
rs200921214 Chr.12: 53973235-53973235 on Build GRCh38
rs4759315 Chr.12: 53973277-53973277 on Build GRCh38
rs745410942 Chr.12: 53973285-53973285 on Build GRCh38
rs367923255 Chr.12: 53973482-53973482 on Build GRCh38
rs769755642 Chr.12: 53973370-53973370 on Build GRCh38
rs764327229 Chr.12: 53973447-53973447 on Build GRCh38
rs567250610 Chr.12: 53973136-53973136 on Build GRCh38
rs763832970 Chr.12: 53973295-53973295 on Build GRCh38
rs754436960 Chr.12: 53973309-53973309 on Build GRCh38
rs367629889 Chr.12: 53973405-53973405 on Build GRCh38
rs372792014 Chr.12: 53973202-53973202 on Build GRCh38
rs768447878 Chr.12: 53973382-53973382 on Build GRCh38
rs1012749118 Chr.12: 53973276-53973276 on Build GRCh38
rs374603975 Chr.12: 53973339-53973339 on Build GRCh38
rs761527831 Chr.12: 53973408-53973408 on Build GRCh38
rs1000921625 Chr.12: 53973330-53973330 on Build GRCh38
rs908965688 Chr.12: 53973161-53973161 on Build GRCh38
rs945638731 Chr.12: 53973519-53973519 on Build GRCh38
rs771665384 Chr.12: 53973292-53973292 on Build GRCh38
rs747870649 Chr.12: 53973488-53973488 on Build GRCh38
rs771475169 Chr.12: 53973411-53973411 on Build GRCh38
rs377372195 Chr.12: 53973229-53973229 on Build GRCh38
rs779496207 Chr.12: 53973378-53973378 on Build GRCh38
rs750226190 Chr.12: 53973458-53973458 on Build GRCh38
rs372124938 Chr.12: 53973499-53973499 on Build GRCh38
rs537732736 Chr.12: 53973178-53973178 on Build GRCh38
rs1035995876 Chr.12: 53973199-53973199 on Build GRCh38
rs750802083 Chr.12: 53973210-53973210 on Build GRCh38
rs187224438 Chr.12: 53973130-53973130 on Build GRCh38
rs35477816 Chr.12: 53973450-53973451 on Build GRCh38
rs767025016 Chr.12: 53973307-53973307 on Build GRCh38
rs762061699 Chr.12: 53973243-53973243 on Build GRCh38
rs941746193 Chr.12: 53973184-53973184 on Build GRCh38
rs777751809 Chr.12: 53973294-53973294 on Build GRCh38
rs984861461 Chr.12: 53973453-53973453 on Build GRCh38
rs956711637 Chr.12: 53973449-53973449 on Build GRCh38
rs781022669 Chr.12: 53973452-53973452 on Build GRCh38
rs183488417 Chr.12: 53973095-53973095 on Build GRCh38
rs926659562 Chr.12: 53973104-53973104 on Build GRCh38
rs903945259 Chr.12: 53973308-53973308 on Build GRCh38
rs938240917 Chr.12: 53973131-53973132 on Build GRCh38
rs17849492 Chr.12: 53973384-53973384 on Build GRCh38
rs745841501 Chr.12: 53973357-53973357 on Build GRCh38
rs894334146 Chr.12: 53973273-53973273 on Build GRCh38
rs201705157 Chr.12: 53973432-53973432 on Build GRCh38
rs746737594 Chr.12: 53973250-53973250 on Build GRCh38
rs748782060 Chr.12: 53973470-53973470 on Build GRCh38
rs529380030 Chr.12: 53973523-53973523 on Build GRCh38
rs141903332 Chr.12: 53973289-53973289 on Build GRCh38
rs773727228 Chr.12: 53973297-53973297 on Build GRCh38
rs773228082 Chr.12: 53973526-53973526 on Build GRCh38

View Full Product Details