Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54375742-54376286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC102724050
Entrez Gene ID
102724050
Gene Name
uncharacterized LOC102724050
Gene Aliases
-
UniGene
Hs.670941
Gene Chromosome Location
Chr.12: 54353691-54467030 on Build GRCh38
Transcripts
NR_120486.1 NR_120487.1 NM_001130967.2 NM_001130968.2 NM_001290001.1 NM_001290002.1 NM_001290004.1 NM_015481.2 XM_005268783.4 XM_006719342.2 XM_011538168.2 XM_011538169.2 XM_011538170.1 XM_011538171.2 XM_017019174.1

Gene Symbol
ZNF385A
Entrez Gene ID
25946
Gene Name
zinc finger protein 385A
Gene Aliases
HZF, RZF, ZFP385, ZNF385
UniGene
Hs.505653
Gene Chromosome Location
Chr.12: 54369129-54391609 on Build GRCh38
Transcripts
NR_120486.1 NR_120487.1 NM_001130967.2 NM_001130968.2 NM_001290001.1 NM_001290002.1 NM_001290004.1 NM_015481.2 XM_005268783.4 XM_006719342.2 XM_011538168.2 XM_011538169.2 XM_011538170.1 XM_011538171.2 XM_017019174.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6829775 c.92A>C p.D31A Chr.12: 54375950-54375950 on Build GRCh38
6484046 c.133C>G p.P45A Chr.12: 54375909-54375909 on Build GRCh38
6829772 c.194C>A p.S65Y Chr.12: 54375848-54375848 on Build GRCh38
6829781 c.153G>A p.R51R Chr.12: 54375889-54375889 on Build GRCh38
6829780 c.153G>A p.R51R Chr.12: 54375889-54375889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780146658 Chr.12: 54375991-54375991 on Build GRCh38
rs548645857 Chr.12: 54376084-54376084 on Build GRCh38
rs540007544 Chr.12: 54375964-54375964 on Build GRCh38
rs780091648 Chr.12: 54375890-54375890 on Build GRCh38
rs560738184 Chr.12: 54375901-54375901 on Build GRCh38
rs781736220 Chr.12: 54375885-54375885 on Build GRCh38
rs774027657 Chr.12: 54376115-54376115 on Build GRCh38
rs2700153 Chr.12: 54376080-54376080 on Build GRCh38
rs760679071 Chr.12: 54375916-54375916 on Build GRCh38
rs1050557978 Chr.12: 54375969-54375969 on Build GRCh38
rs761676279 Chr.12: 54375824-54375824 on Build GRCh38
rs975409648 Chr.12: 54376083-54376083 on Build GRCh38
rs149814119 Chr.12: 54376235-54376235 on Build GRCh38
rs771698908 Chr.12: 54375910-54375910 on Build GRCh38
rs760266544 Chr.12: 54375796-54375796 on Build GRCh38
rs756605649 Chr.12: 54375987-54375987 on Build GRCh38
rs1049146643 Chr.12: 54376037-54376037 on Build GRCh38
rs550128131 Chr.12: 54375925-54375925 on Build GRCh38
rs751934638 Chr.12: 54375828-54375828 on Build GRCh38
rs765315823 Chr.12: 54376003-54376005 on Build GRCh38
rs967637552 Chr.12: 54376012-54376012 on Build GRCh38
rs376574982 Chr.12: 54375798-54375798 on Build GRCh38
rs537042403 Chr.12: 54376180-54376180 on Build GRCh38
rs767712479 Chr.12: 54375979-54375979 on Build GRCh38
rs569976098 Chr.12: 54376145-54376145 on Build GRCh38
rs376929390 Chr.12: 54375980-54375980 on Build GRCh38
rs1009310631 Chr.12: 54376247-54376247 on Build GRCh38
rs747996419 Chr.12: 54375909-54375909 on Build GRCh38
rs774062871 Chr.12: 54375908-54375908 on Build GRCh38
rs386763164 Chr.12: 54376147-54376148 on Build GRCh38
rs920020383 Chr.12: 54376107-54376107 on Build GRCh38
rs759313388 Chr.12: 54375949-54375949 on Build GRCh38
rs949307033 Chr.12: 54376138-54376138 on Build GRCh38
rs754962910 Chr.12: 54376000-54376000 on Build GRCh38
rs931382626 Chr.12: 54376113-54376113 on Build GRCh38
rs764968698 Chr.12: 54375962-54375962 on Build GRCh38
rs778041996 Chr.12: 54375822-54375822 on Build GRCh38
rs577132026 Chr.12: 54376254-54376254 on Build GRCh38
rs370356057 Chr.12: 54375992-54375992 on Build GRCh38
rs551613567 Chr.12: 54376051-54376051 on Build GRCh38
rs532407591 Chr.12: 54375973-54375973 on Build GRCh38
rs565839223 Chr.12: 54376031-54376031 on Build GRCh38
rs776252685 Chr.12: 54375933-54375933 on Build GRCh38
rs754674136 Chr.12: 54375819-54375819 on Build GRCh38
rs769873316 Chr.12: 54375888-54375888 on Build GRCh38
rs201589447 Chr.12: 54375913-54375913 on Build GRCh38
rs1047374383 Chr.12: 54376114-54376114 on Build GRCh38
rs932130644 Chr.12: 54375891-54375891 on Build GRCh38
rs753604521 Chr.12: 54375809-54375809 on Build GRCh38
rs757518593 Chr.12: 54375857-54375857 on Build GRCh38
rs1001888913 Chr.12: 54376198-54376198 on Build GRCh38
rs905082475 Chr.12: 54376191-54376191 on Build GRCh38
rs750698231 Chr.12: 54376007-54376008 on Build GRCh38
rs777908583 Chr.12: 54376140-54376140 on Build GRCh38
rs111505697 Chr.12: 54375886-54375886 on Build GRCh38
rs976622251 Chr.12: 54376002-54376003 on Build GRCh38
rs542680871 Chr.12: 54375895-54375895 on Build GRCh38
rs534253559 Chr.12: 54376239-54376239 on Build GRCh38
rs1053417357 Chr.12: 54376212-54376212 on Build GRCh38
rs565168005 Chr.12: 54375967-54375967 on Build GRCh38
rs200627454 Chr.12: 54376197-54376197 on Build GRCh38

View Full Product Details