Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55294598-55295141 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR6C6
Entrez Gene ID
283365
Gene Name
olfactory receptor family 6 subfamily C member 6
Gene Aliases
-
UniGene
Hs.554555
Gene Chromosome Location
Chr.12: 55294288-55295232 on Build GRCh38
Transcripts
NM_001005493.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3462987 c.493G>A p.D165N Chr.12: 55294740-55294740 on Build GRCh38
1293492 c.470_471CA>TT p.P157L Chr.12: 55294762-55294763 on Build GRCh38
941260 c.240G>T p.L80F Chr.12: 55294993-55294993 on Build GRCh38
4903531 c.148G>A p.D50N Chr.12: 55295085-55295085 on Build GRCh38
6137553 c.459C>T p.I153I Chr.12: 55294774-55294774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762101280 Chr.12: 55295085-55295085 on Build GRCh38
rs774853693 Chr.12: 55295083-55295083 on Build GRCh38
rs149778052 Chr.12: 55294646-55294646 on Build GRCh38
rs772385046 Chr.12: 55294626-55294626 on Build GRCh38
rs151019652 Chr.12: 55294625-55294625 on Build GRCh38
rs768927891 Chr.12: 55294665-55294666 on Build GRCh38
rs78183586 Chr.12: 55295111-55295111 on Build GRCh38
rs770802926 Chr.12: 55294636-55294636 on Build GRCh38
rs763560106 Chr.12: 55295051-55295054 on Build GRCh38
rs746648924 Chr.12: 55295056-55295056 on Build GRCh38
rs76796682 Chr.12: 55294634-55294634 on Build GRCh38
rs763849128 Chr.12: 55294672-55294672 on Build GRCh38
rs770356780 Chr.12: 55294807-55294814 on Build GRCh38
rs761623941 Chr.12: 55294763-55294763 on Build GRCh38
rs760918759 Chr.12: 55295106-55295106 on Build GRCh38
rs765801314 Chr.12: 55294817-55294817 on Build GRCh38
rs761535964 Chr.12: 55294887-55294887 on Build GRCh38
rs779790933 Chr.12: 55294951-55294951 on Build GRCh38
rs956997118 Chr.12: 55294926-55294926 on Build GRCh38
rs779396403 Chr.12: 55294707-55294707 on Build GRCh38
rs11171402 Chr.12: 55294664-55294664 on Build GRCh38
rs35212493 Chr.12: 55295030-55295031 on Build GRCh38
rs375624704 Chr.12: 55294988-55294988 on Build GRCh38
rs546567397 Chr.12: 55295013-55295013 on Build GRCh38
rs770467066 Chr.12: 55295002-55295002 on Build GRCh38
rs781318586 Chr.12: 55294735-55294735 on Build GRCh38
rs372117452 Chr.12: 55295049-55295049 on Build GRCh38
rs541789315 Chr.12: 55294825-55294825 on Build GRCh38
rs77411445 Chr.12: 55295069-55295069 on Build GRCh38
rs143781018 Chr.12: 55294721-55294721 on Build GRCh38
rs138409643 Chr.12: 55294910-55294910 on Build GRCh38
rs938033756 Chr.12: 55294831-55294831 on Build GRCh38
rs765014918 Chr.12: 55294660-55294660 on Build GRCh38
rs750001109 Chr.12: 55294793-55294793 on Build GRCh38
rs761613179 Chr.12: 55295096-55295097 on Build GRCh38
rs987019097 Chr.12: 55294704-55294704 on Build GRCh38
rs112475102 Chr.12: 55294919-55294919 on Build GRCh38
rs767120777 Chr.12: 55294791-55294791 on Build GRCh38
rs765222572 Chr.12: 55295098-55295103 on Build GRCh38
rs937584820 Chr.12: 55294792-55294792 on Build GRCh38
rs529569476 Chr.12: 55295041-55295041 on Build GRCh38
rs906134909 Chr.12: 55294743-55294743 on Build GRCh38
rs1055212635 Chr.12: 55294851-55294851 on Build GRCh38
rs751985604 Chr.12: 55294922-55294922 on Build GRCh38
rs760280603 Chr.12: 55294901-55294901 on Build GRCh38
rs372388367 Chr.12: 55294957-55294957 on Build GRCh38
rs760088389 Chr.12: 55294794-55294794 on Build GRCh38
rs369845779 Chr.12: 55295008-55295008 on Build GRCh38
rs1055354978 Chr.12: 55294838-55294838 on Build GRCh38
rs767865021 Chr.12: 55295088-55295088 on Build GRCh38
rs775116735 Chr.12: 55294746-55294746 on Build GRCh38
rs751973071 Chr.12: 55294918-55294918 on Build GRCh38
rs200826443 Chr.12: 55295093-55295093 on Build GRCh38
rs750454046 Chr.12: 55294928-55294928 on Build GRCh38
rs764508423 Chr.12: 55294914-55294914 on Build GRCh38
rs772806047 Chr.12: 55294892-55294892 on Build GRCh38
rs766540478 Chr.12: 55295109-55295109 on Build GRCh38
rs766924846 Chr.12: 55294679-55294679 on Build GRCh38
rs896718733 Chr.12: 55294867-55294867 on Build GRCh38
rs778075050 Chr.12: 55294730-55294730 on Build GRCh38
rs752951320 Chr.12: 55294818-55294818 on Build GRCh38
rs773410825 Chr.12: 55294882-55294882 on Build GRCh38
rs757537272 Chr.12: 55294850-55294850 on Build GRCh38
rs866508134 Chr.12: 55294927-55294927 on Build GRCh38
rs542816698 Chr.12: 55294880-55294880 on Build GRCh38
rs777969874 Chr.12: 55294847-55294847 on Build GRCh38
rs141613478 Chr.12: 55295043-55295043 on Build GRCh38
rs760146733 Chr.12: 55295029-55295029 on Build GRCh38
rs371680600 Chr.12: 55294742-55294742 on Build GRCh38
rs975471004 Chr.12: 55295028-55295028 on Build GRCh38
rs75303496 Chr.12: 55294670-55294670 on Build GRCh38
rs754391945 Chr.12: 55294689-55294689 on Build GRCh38
rs888020821 Chr.12: 55295098-55295100 on Build GRCh38
rs763311940 Chr.12: 55294929-55294930 on Build GRCh38
rs764551855 Chr.12: 55295012-55295012 on Build GRCh38
rs771005694 Chr.12: 55294733-55294733 on Build GRCh38
rs267603560 Chr.12: 55294637-55294637 on Build GRCh38
rs986766608 Chr.12: 55295068-55295068 on Build GRCh38
rs748655339 Chr.12: 55294722-55294722 on Build GRCh38
rs148383430 Chr.12: 55295034-55295034 on Build GRCh38
rs748980249 Chr.12: 55294875-55294875 on Build GRCh38
rs745843004 Chr.12: 55294906-55294906 on Build GRCh38
rs1013862610 Chr.12: 55294899-55294899 on Build GRCh38
rs756851218 Chr.12: 55294678-55294678 on Build GRCh38
rs775189138 Chr.12: 55295024-55295025 on Build GRCh38
rs762187412 Chr.12: 55295027-55295027 on Build GRCh38
rs763496600 Chr.12: 55295009-55295009 on Build GRCh38
rs145153717 Chr.12: 55294944-55294944 on Build GRCh38
rs200792871 Chr.12: 55294958-55294958 on Build GRCh38
rs781231440 Chr.12: 55294864-55294864 on Build GRCh38
rs267603561 Chr.12: 55294741-55294741 on Build GRCh38
rs199671649 Chr.12: 55295115-55295115 on Build GRCh38
rs150512978 Chr.12: 55295045-55295045 on Build GRCh38
rs868041374 Chr.12: 55294692-55294692 on Build GRCh38
rs539665121 Chr.12: 55295079-55295079 on Build GRCh38
rs145260899 Chr.12: 55295116-55295116 on Build GRCh38
rs142751888 Chr.12: 55295097-55295097 on Build GRCh38
rs201844969 Chr.12: 55294869-55294869 on Build GRCh38
rs755598368 Chr.12: 55294693-55294693 on Build GRCh38
rs754707350 Chr.12: 55294747-55294747 on Build GRCh38
rs988853973 Chr.12: 55294681-55294681 on Build GRCh38
rs746450515 Chr.12: 55294993-55294993 on Build GRCh38
rs530742573 Chr.12: 55294874-55294874 on Build GRCh38
rs200328804 Chr.12: 55294979-55294979 on Build GRCh38
rs774070804 Chr.12: 55294878-55294878 on Build GRCh38
rs753409640 Chr.12: 55295078-55295079 on Build GRCh38
rs757658611 Chr.12: 55295048-55295048 on Build GRCh38
rs985358114 Chr.12: 55294985-55294985 on Build GRCh38
rs552948172 Chr.12: 55294716-55294716 on Build GRCh38
rs920025742 Chr.12: 55295091-55295091 on Build GRCh38
rs937598234 Chr.12: 55295095-55295095 on Build GRCh38
rs773008930 Chr.12: 55294992-55294992 on Build GRCh38
rs562689518 Chr.12: 55294889-55294889 on Build GRCh38
rs774068305 Chr.12: 55294759-55294759 on Build GRCh38
rs145746232 Chr.12: 55294756-55294756 on Build GRCh38
rs771655486 Chr.12: 55294968-55294968 on Build GRCh38
rs763439814 Chr.12: 55294911-55294911 on Build GRCh38
rs781704171 Chr.12: 55294690-55294692 on Build GRCh38
rs192785175 Chr.12: 55294727-55294727 on Build GRCh38
rs200002470 Chr.12: 55294684-55294684 on Build GRCh38
rs374548635 Chr.12: 55294814-55294814 on Build GRCh38
rs750993797 Chr.12: 55294676-55294676 on Build GRCh38
rs138420259 Chr.12: 55295082-55295082 on Build GRCh38
rs78315631 Chr.12: 55295033-55295033 on Build GRCh38
rs117851256 Chr.12: 55295110-55295110 on Build GRCh38
rs201362654 Chr.12: 55295074-55295074 on Build GRCh38
rs866701494 Chr.12: 55295077-55295078 on Build GRCh38
rs746999886 Chr.12: 55294731-55294731 on Build GRCh38
rs773132166 Chr.12: 55294631-55294631 on Build GRCh38
rs751805569 Chr.12: 55294842-55294842 on Build GRCh38

View Full Product Details