Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55637605-55638115 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR10P1
Entrez Gene ID
121130
Gene Name
olfactory receptor family 10 subfamily P member 1
Gene Aliases
OR10P1P, OR10P2P, OR10P3P, OR12-7, OST701
UniGene
Hs.554551
Gene Chromosome Location
Chr.12: 55636892-55637833 on Build GRCh38
Transcripts
NM_206899.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6137535 c.796G>A p.G266S Chr.12: 55637687-55637687 on Build GRCh38
322213 c.901G>T p.A301S Chr.12: 55637792-55637792 on Build GRCh38
301197 c.820G>A p.V274I Chr.12: 55637711-55637711 on Build GRCh38
3463148 c.867C>T p.I289I Chr.12: 55637758-55637758 on Build GRCh38
4709619 c.839delC p.T280fs*19 Chr.12: 55637730-55637730 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779886280 Chr.12: 55637791-55637791 on Build GRCh38
rs10876839 Chr.12: 55638079-55638079 on Build GRCh38
rs778961287 Chr.12: 55637674-55637674 on Build GRCh38
rs776924950 Chr.12: 55637811-55637811 on Build GRCh38
rs930635138 Chr.12: 55638081-55638081 on Build GRCh38
rs957695070 Chr.12: 55638007-55638007 on Build GRCh38
rs76049580 Chr.12: 55637782-55637782 on Build GRCh38
rs761298035 Chr.12: 55637739-55637739 on Build GRCh38
rs774620519 Chr.12: 55637786-55637786 on Build GRCh38
rs368136407 Chr.12: 55637776-55637776 on Build GRCh38
rs981520346 Chr.12: 55637941-55637941 on Build GRCh38
rs767572823 Chr.12: 55637838-55637838 on Build GRCh38
rs1055811658 Chr.12: 55637864-55637864 on Build GRCh38
rs187125465 Chr.12: 55638015-55638015 on Build GRCh38
rs755666158 Chr.12: 55637810-55637810 on Build GRCh38
rs752705189 Chr.12: 55637771-55637771 on Build GRCh38
rs940856009 Chr.12: 55638016-55638016 on Build GRCh38
rs752296913 Chr.12: 55637783-55637783 on Build GRCh38
rs755768351 Chr.12: 55637789-55637789 on Build GRCh38
rs769635291 Chr.12: 55637708-55637708 on Build GRCh38
rs748787014 Chr.12: 55637793-55637793 on Build GRCh38
rs562128796 Chr.12: 55637983-55637983 on Build GRCh38
rs267603572 Chr.12: 55637792-55637792 on Build GRCh38
rs988621209 Chr.12: 55637918-55637926 on Build GRCh38
rs1012542484 Chr.12: 55637724-55637724 on Build GRCh38
rs927269835 Chr.12: 55637719-55637719 on Build GRCh38
rs118082633 Chr.12: 55638037-55638037 on Build GRCh38
rs375663637 Chr.12: 55637798-55637798 on Build GRCh38
rs781178778 Chr.12: 55637695-55637695 on Build GRCh38
rs762402694 Chr.12: 55637635-55637635 on Build GRCh38
rs954065500 Chr.12: 55637649-55637649 on Build GRCh38
rs999863862 Chr.12: 55637973-55637973 on Build GRCh38
rs780369555 Chr.12: 55637717-55637717 on Build GRCh38
rs759742221 Chr.12: 55637756-55637756 on Build GRCh38
rs140882217 Chr.12: 55637746-55637746 on Build GRCh38
rs759897189 Chr.12: 55637819-55637819 on Build GRCh38
rs762375977 Chr.12: 55637710-55637710 on Build GRCh38
rs763284810 Chr.12: 55637826-55637826 on Build GRCh38
rs747709358 Chr.12: 55637873-55637873 on Build GRCh38
rs758099374 Chr.12: 55637661-55637661 on Build GRCh38
rs573446976 Chr.12: 55637921-55637921 on Build GRCh38
rs866903129 Chr.12: 55637863-55637863 on Build GRCh38
rs72482258 Chr.12: 55638023-55638023 on Build GRCh38
rs1050786043 Chr.12: 55638078-55638078 on Build GRCh38
rs770654411 Chr.12: 55637686-55637686 on Build GRCh38
rs772594253 Chr.12: 55637711-55637711 on Build GRCh38
rs575283712 Chr.12: 55637841-55637841 on Build GRCh38
rs374603359 Chr.12: 55637772-55637772 on Build GRCh38
rs184213016 Chr.12: 55637948-55637948 on Build GRCh38
rs547623506 Chr.12: 55638022-55638022 on Build GRCh38
rs745482705 Chr.12: 55637704-55637704 on Build GRCh38
rs536972157 Chr.12: 55637679-55637679 on Build GRCh38
rs750735614 Chr.12: 55637641-55637641 on Build GRCh38
rs750720150 Chr.12: 55637727-55637728 on Build GRCh38
rs937344253 Chr.12: 55637984-55637984 on Build GRCh38
rs2555035 Chr.12: 55637920-55637920 on Build GRCh38
rs755234101 Chr.12: 55637673-55637673 on Build GRCh38
rs760979917 Chr.12: 55637657-55637657 on Build GRCh38
rs376369727 Chr.12: 55637859-55637859 on Build GRCh38
rs773952811 Chr.12: 55637799-55637799 on Build GRCh38
rs775595582 Chr.12: 55637823-55637823 on Build GRCh38
rs7309433 Chr.12: 55638053-55638053 on Build GRCh38
rs778274860 Chr.12: 55637872-55637872 on Build GRCh38
rs899242379 Chr.12: 55637689-55637689 on Build GRCh38
rs570772947 Chr.12: 55638088-55638088 on Build GRCh38
rs777151955 Chr.12: 55637885-55637885 on Build GRCh38
rs906438990 Chr.12: 55637953-55637953 on Build GRCh38
rs115204039 Chr.12: 55638070-55638070 on Build GRCh38
rs146503734 Chr.12: 55637709-55637709 on Build GRCh38
rs775176418 Chr.12: 55637633-55637633 on Build GRCh38
rs962295609 Chr.12: 55638056-55638056 on Build GRCh38
rs754727461 Chr.12: 55637861-55637861 on Build GRCh38
rs771363225 Chr.12: 55637876-55637876 on Build GRCh38
rs971093760 Chr.12: 55637752-55637752 on Build GRCh38
rs376517791 Chr.12: 55637842-55637842 on Build GRCh38
rs953132234 Chr.12: 55638034-55638034 on Build GRCh38
rs11171631 Chr.12: 55637827-55637827 on Build GRCh38
rs780047644 Chr.12: 55637852-55637852 on Build GRCh38
rs751429640 Chr.12: 55637780-55637780 on Build GRCh38
rs199780580 Chr.12: 55637638-55637638 on Build GRCh38
rs534859426 Chr.12: 55637754-55637754 on Build GRCh38
rs757125761 Chr.12: 55637830-55637830 on Build GRCh38
rs550597985 Chr.12: 55637668-55637668 on Build GRCh38
rs565312254 Chr.12: 55637900-55637900 on Build GRCh38
rs138844754 Chr.12: 55637892-55637892 on Build GRCh38
rs143887647 Chr.12: 55637680-55637680 on Build GRCh38
rs113745484 Chr.12: 55637676-55637676 on Build GRCh38
rs985388863 Chr.12: 55637664-55637664 on Build GRCh38
rs896868853 Chr.12: 55637795-55637795 on Build GRCh38
rs765401594 Chr.12: 55637820-55637820 on Build GRCh38
rs758712551 Chr.12: 55637861-55637875 on Build GRCh38
rs112739911 Chr.12: 55637770-55637770 on Build GRCh38
rs752834026 Chr.12: 55637675-55637675 on Build GRCh38

View Full Product Details