Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55987020-55987515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB5B
Entrez Gene ID
5869
Gene Name
RAB5B, member RAS oncogene family
Gene Aliases
-
UniGene
Hs.157659
Gene Chromosome Location
Chr.12: 55973913-55996683 on Build GRCh38
Transcripts
NM_001252036.1 NM_001252037.1 NM_002868.3 XM_005269051.2 XM_005269052.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5494644 c.158T>C p.I53T Chr.12: 55987118-55987118 on Build GRCh38
941457 c.76C>T p.L26L Chr.12: 55987036-55987036 on Build GRCh38
941458 c.130C>T p.Q44* Chr.12: 55987090-55987090 on Build GRCh38
7092755 c.82G>A p.E28K Chr.12: 55987042-55987042 on Build GRCh38
5375600 c.86C>T p.S29F Chr.12: 55987046-55987046 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779738899 Chr.12: 55987339-55987339 on Build GRCh38
rs748775328 Chr.12: 55987167-55987170 on Build GRCh38
rs991111441 Chr.12: 55987459-55987459 on Build GRCh38
rs759235479 Chr.12: 55987164-55987164 on Build GRCh38
rs769431200 Chr.12: 55987167-55987169 on Build GRCh38
rs985739487 Chr.12: 55987335-55987335 on Build GRCh38
rs1045901995 Chr.12: 55987164-55987164 on Build GRCh38
rs566051029 Chr.12: 55987180-55987180 on Build GRCh38
rs747430136 Chr.12: 55987167-55987168 on Build GRCh38
rs770835155 Chr.12: 55987136-55987136 on Build GRCh38
rs34223637 Chr.12: 55987166-55987167 on Build GRCh38
rs1008771464 Chr.12: 55987370-55987370 on Build GRCh38
rs961868078 Chr.12: 55987444-55987444 on Build GRCh38
rs751181190 Chr.12: 55987245-55987245 on Build GRCh38
rs962265208 Chr.12: 55987206-55987206 on Build GRCh38
rs957353178 Chr.12: 55987071-55987071 on Build GRCh38
rs778834831 Chr.12: 55987076-55987076 on Build GRCh38
rs949811624 Chr.12: 55987479-55987479 on Build GRCh38
rs988744305 Chr.12: 55987463-55987463 on Build GRCh38
rs903316385 Chr.12: 55987264-55987264 on Build GRCh38
rs930303523 Chr.12: 55987225-55987225 on Build GRCh38
rs560251258 Chr.12: 55987309-55987309 on Build GRCh38
rs1019205044 Chr.12: 55987415-55987415 on Build GRCh38
rs972409782 Chr.12: 55987447-55987447 on Build GRCh38
rs148835571 Chr.12: 55987483-55987483 on Build GRCh38
rs577955469 Chr.12: 55987286-55987286 on Build GRCh38
rs748206537 Chr.12: 55987101-55987101 on Build GRCh38
rs901871239 Chr.12: 55987277-55987277 on Build GRCh38
rs950257900 Chr.12: 55987259-55987259 on Build GRCh38
rs971974650 Chr.12: 55987213-55987213 on Build GRCh38
rs755651879 Chr.12: 55987138-55987138 on Build GRCh38
rs746934152 Chr.12: 55987131-55987131 on Build GRCh38
rs955440756 Chr.12: 55987318-55987318 on Build GRCh38
rs765031067 Chr.12: 55987169-55987169 on Build GRCh38
rs761499790 Chr.12: 55987303-55987303 on Build GRCh38
rs999515461 Chr.12: 55987301-55987301 on Build GRCh38
rs144336111 Chr.12: 55987222-55987222 on Build GRCh38
rs183304949 Chr.12: 55987458-55987458 on Build GRCh38
rs556485643 Chr.12: 55987275-55987275 on Build GRCh38
rs772072840 Chr.12: 55987116-55987116 on Build GRCh38
rs975919056 Chr.12: 55987094-55987094 on Build GRCh38
rs911853555 Chr.12: 55987199-55987199 on Build GRCh38
rs971498907 Chr.12: 55987484-55987484 on Build GRCh38
rs915649305 Chr.12: 55987246-55987246 on Build GRCh38
rs576363481 Chr.12: 55987250-55987250 on Build GRCh38
rs755230046 Chr.12: 55987072-55987072 on Build GRCh38
rs572070989 Chr.12: 55987400-55987400 on Build GRCh38
rs746342722 Chr.12: 55987134-55987135 on Build GRCh38
rs920495653 Chr.12: 55987456-55987456 on Build GRCh38
rs749501744 Chr.12: 55987066-55987066 on Build GRCh38
rs912871681 Chr.12: 55987476-55987476 on Build GRCh38
rs1014839266 Chr.12: 55987271-55987271 on Build GRCh38
rs538176415 Chr.12: 55987263-55987263 on Build GRCh38
rs765608332 Chr.12: 55987487-55987487 on Build GRCh38
rs953219939 Chr.12: 55987168-55987168 on Build GRCh38
rs773134788 Chr.12: 55987117-55987117 on Build GRCh38
rs766749669 Chr.12: 55987312-55987312 on Build GRCh38
rs763623639 Chr.12: 55987096-55987096 on Build GRCh38
rs915694870 Chr.12: 55987464-55987464 on Build GRCh38
rs78661386 Chr.12: 55987184-55987184 on Build GRCh38
rs758831808 Chr.12: 55987167-55987167 on Build GRCh38
rs754217889 Chr.12: 55987313-55987313 on Build GRCh38
rs553902200 Chr.12: 55987142-55987142 on Build GRCh38
rs997582958 Chr.12: 55987293-55987293 on Build GRCh38
rs987286694 Chr.12: 55987173-55987173 on Build GRCh38
rs545226497 Chr.12: 55987294-55987294 on Build GRCh38
rs375382798 Chr.12: 55987221-55987221 on Build GRCh38
rs906017985 Chr.12: 55987183-55987183 on Build GRCh38
rs747566231 Chr.12: 55987129-55987129 on Build GRCh38
rs61937246 Chr.12: 55987181-55987181 on Build GRCh38

View Full Product Details