Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 56162165-56162671 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMARCC2
Entrez Gene ID
6601
Gene Name
SWI/SNF related, matrix associated, actin dependent regulator of chromatin subfamily c member 2
Gene Aliases
BAF170, CRACC2, Rsc8
UniGene
Hs.236030
Gene Chromosome Location
Chr.12: 56161852-56189567 on Build GRCh38
Transcripts
NM_001130420.1 NM_003075.3 NM_139067.2 XM_005269101.1 XM_005269102.1 XM_005269103.1 XM_005269104.1 XM_011538693.2 XM_017019884.1 XM_017019885.1 XM_017019886.1 XM_017019887.1

SNP Detail

SNP ID Chromosome Location
rs552827280 Chr.12: 56162614-56162614 on Build GRCh38
rs904631684 Chr.12: 56162190-56162190 on Build GRCh38
rs369945276 Chr.12: 56162488-56162488 on Build GRCh38
rs1000121145 Chr.12: 56162193-56162193 on Build GRCh38
rs918556252 Chr.12: 56162283-56162283 on Build GRCh38
rs187598740 Chr.12: 56162272-56162272 on Build GRCh38
rs973172422 Chr.12: 56162588-56162588 on Build GRCh38
rs543450682 Chr.12: 56162257-56162257 on Build GRCh38
rs1018265845 Chr.12: 56162194-56162194 on Build GRCh38
rs1051121316 Chr.12: 56162311-56162311 on Build GRCh38
rs1041868706 Chr.12: 56162394-56162394 on Build GRCh38
rs941586683 Chr.12: 56162233-56162233 on Build GRCh38
rs569980958 Chr.12: 56162508-56162508 on Build GRCh38
rs566275386 Chr.12: 56162385-56162385 on Build GRCh38
rs954789778 Chr.12: 56162529-56162529 on Build GRCh38
rs556530439 Chr.12: 56162412-56162414 on Build GRCh38
rs1058342 Chr.12: 56162603-56162603 on Build GRCh38
rs904611731 Chr.12: 56162398-56162398 on Build GRCh38
rs565192781 Chr.12: 56162269-56162269 on Build GRCh38
rs917500132 Chr.12: 56162331-56162331 on Build GRCh38
rs927017310 Chr.12: 56162256-56162256 on Build GRCh38
rs981849292 Chr.12: 56162192-56162192 on Build GRCh38
rs372065045 Chr.12: 56162309-56162309 on Build GRCh38
rs963120287 Chr.12: 56162579-56162579 on Build GRCh38
rs781151522 Chr.12: 56162595-56162595 on Build GRCh38
rs973056572 Chr.12: 56162248-56162248 on Build GRCh38
rs918623602 Chr.12: 56162566-56162566 on Build GRCh38
rs777430784 Chr.12: 56162533-56162533 on Build GRCh38
rs531595622 Chr.12: 56162212-56162212 on Build GRCh38
rs1004685028 Chr.12: 56162555-56162555 on Build GRCh38
rs893741242 Chr.12: 56162406-56162407 on Build GRCh38
rs907584326 Chr.12: 56162234-56162234 on Build GRCh38
rs375352481 Chr.12: 56162371-56162371 on Build GRCh38
rs890308960 Chr.12: 56162312-56162312 on Build GRCh38
rs577354478 Chr.12: 56162395-56162398 on Build GRCh38
rs986874327 Chr.12: 56162599-56162599 on Build GRCh38
rs746757371 Chr.12: 56162545-56162545 on Build GRCh38
rs71459346 Chr.12: 56162231-56162231 on Build GRCh38
rs954541346 Chr.12: 56162216-56162216 on Build GRCh38
rs772793690 Chr.12: 56162401-56162402 on Build GRCh38
rs963131199 Chr.12: 56162245-56162245 on Build GRCh38
rs996133707 Chr.12: 56162202-56162202 on Build GRCh38
rs946337863 Chr.12: 56162639-56162639 on Build GRCh38
rs910149548 Chr.12: 56162223-56162223 on Build GRCh38
rs796530906 Chr.12: 56162413-56162414 on Build GRCh38
rs937570385 Chr.12: 56162306-56162306 on Build GRCh38
rs755680249 Chr.12: 56162217-56162217 on Build GRCh38
rs946193603 Chr.12: 56162335-56162335 on Build GRCh38
rs758207608 Chr.12: 56162570-56162570 on Build GRCh38
rs745785785 Chr.12: 56162602-56162602 on Build GRCh38
rs558530398 Chr.12: 56162556-56162556 on Build GRCh38
rs1039662553 Chr.12: 56162466-56162466 on Build GRCh38
rs74836828 Chr.12: 56162383-56162383 on Build GRCh38
rs928157591 Chr.12: 56162197-56162197 on Build GRCh38
rs34102859 Chr.12: 56162605-56162605 on Build GRCh38
rs1003319047 Chr.12: 56162496-56162496 on Build GRCh38
rs1020666586 Chr.12: 56162448-56162448 on Build GRCh38
rs34426006 Chr.12: 56162452-56162452 on Build GRCh38
rs1000278484 Chr.12: 56162458-56162458 on Build GRCh38
rs943202741 Chr.12: 56162384-56162384 on Build GRCh38
rs530350143 Chr.12: 56162410-56162410 on Build GRCh38
rs933972839 Chr.12: 56162580-56162580 on Build GRCh38
rs74092005 Chr.12: 56162479-56162479 on Build GRCh38
rs758981147 Chr.12: 56162343-56162343 on Build GRCh38
rs964984946 Chr.12: 56162200-56162200 on Build GRCh38
rs570428512 Chr.12: 56162592-56162592 on Build GRCh38
rs972763956 Chr.12: 56162238-56162238 on Build GRCh38
rs917466084 Chr.12: 56162615-56162615 on Build GRCh38

View Full Product Details