Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57232341-57232840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHMT2
Entrez Gene ID
6472
Gene Name
serine hydroxymethyltransferase 2
Gene Aliases
GLYA, HEL-S-51e, SHMT
UniGene
Hs.741179
Gene Chromosome Location
Chr.12: 57229573-57234935 on Build GRCh38
Transcripts
NM_001166356.1 NM_001166357.1 NM_001166358.1 NM_001166359.1 NM_005412.5 NR_029415.1 NR_029416.1 NR_029417.1 NR_048562.1 XM_011538675.2 XM_011538676.2 XM_011538677.2 XM_011538678.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5473418 c.363G>A p.R121R Chr.12: 57232509-57232509 on Build GRCh38
941998 c.847C>T p.R283* Chr.12: 57232833-57232833 on Build GRCh38
5216805 c.310delA p.T105fs*11 Chr.12: 57232456-57232456 on Build GRCh38
6777889 c.523G>A p.A175T Chr.12: 57232797-57232797 on Build GRCh38
4502811 c.622C>T p.Q208* Chr.12: 57232480-57232480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780089826 Chr.12: 57232730-57232730 on Build GRCh38
rs768920289 Chr.12: 57232662-57232662 on Build GRCh38
rs77260799 Chr.12: 57232661-57232661 on Build GRCh38
rs891950839 Chr.12: 57232362-57232362 on Build GRCh38
rs781224640 Chr.12: 57232763-57232763 on Build GRCh38
rs921745312 Chr.12: 57232727-57232727 on Build GRCh38
rs780147063 Chr.12: 57232623-57232623 on Build GRCh38
rs750285738 Chr.12: 57232814-57232814 on Build GRCh38
rs756109246 Chr.12: 57232741-57232741 on Build GRCh38
rs770219588 Chr.12: 57232681-57232681 on Build GRCh38
rs373381797 Chr.12: 57232698-57232698 on Build GRCh38
rs368859292 Chr.12: 57232682-57232682 on Build GRCh38
rs781677213 Chr.12: 57232599-57232601 on Build GRCh38
rs759605261 Chr.12: 57232547-57232547 on Build GRCh38
rs148573932 Chr.12: 57232791-57232791 on Build GRCh38
rs78650532 Chr.12: 57232370-57232370 on Build GRCh38
rs771507850 Chr.12: 57232796-57232796 on Build GRCh38
rs749243816 Chr.12: 57232522-57232522 on Build GRCh38
rs758392212 Chr.12: 57232723-57232723 on Build GRCh38
rs773574090 Chr.12: 57232692-57232692 on Build GRCh38
rs747029594 Chr.12: 57232748-57232748 on Build GRCh38
rs757310852 Chr.12: 57232521-57232521 on Build GRCh38
rs774360462 Chr.12: 57232546-57232546 on Build GRCh38
rs768724092 Chr.12: 57232751-57232751 on Build GRCh38
rs1053766115 Chr.12: 57232630-57232630 on Build GRCh38
rs891986656 Chr.12: 57232650-57232650 on Build GRCh38
rs544942528 Chr.12: 57232369-57232369 on Build GRCh38
rs551746000 Chr.12: 57232651-57232651 on Build GRCh38
rs745839664 Chr.12: 57232613-57232613 on Build GRCh38
rs201088482 Chr.12: 57232565-57232565 on Build GRCh38
rs754320831 Chr.12: 57232478-57232478 on Build GRCh38
rs377502791 Chr.12: 57232699-57232699 on Build GRCh38
rs375798229 Chr.12: 57232589-57232589 on Build GRCh38
rs749291993 Chr.12: 57232400-57232400 on Build GRCh38
rs769745150 Chr.12: 57232375-57232375 on Build GRCh38
rs184950916 Chr.12: 57232514-57232514 on Build GRCh38
rs199498720 Chr.12: 57232499-57232499 on Build GRCh38
rs778817041 Chr.12: 57232712-57232712 on Build GRCh38
rs972643721 Chr.12: 57232453-57232453 on Build GRCh38
rs951807571 Chr.12: 57232672-57232672 on Build GRCh38
rs747827761 Chr.12: 57232507-57232507 on Build GRCh38
rs753409171 Chr.12: 57232805-57232805 on Build GRCh38
rs760276535 Chr.12: 57232700-57232700 on Build GRCh38
rs763718059 Chr.12: 57232802-57232802 on Build GRCh38
rs370198751 Chr.12: 57232447-57232447 on Build GRCh38
rs748468819 Chr.12: 57232612-57232612 on Build GRCh38
rs928635466 Chr.12: 57232605-57232605 on Build GRCh38
rs1025511284 Chr.12: 57232461-57232461 on Build GRCh38
rs141575043 Chr.12: 57232669-57232669 on Build GRCh38
rs751086075 Chr.12: 57232487-57232487 on Build GRCh38
rs1011353467 Chr.12: 57232367-57232367 on Build GRCh38
rs900833068 Chr.12: 57232775-57232775 on Build GRCh38
rs764464165 Chr.12: 57232476-57232476 on Build GRCh38
rs762014126 Chr.12: 57232666-57232666 on Build GRCh38
rs751759388 Chr.12: 57232498-57232498 on Build GRCh38
rs757740427 Chr.12: 57232479-57232479 on Build GRCh38
rs765603191 Chr.12: 57232583-57232583 on Build GRCh38
rs983040249 Chr.12: 57232687-57232687 on Build GRCh38
rs772117225 Chr.12: 57232616-57232616 on Build GRCh38
rs572293970 Chr.12: 57232373-57232373 on Build GRCh38
rs1053253659 Chr.12: 57232361-57232361 on Build GRCh38
rs944643577 Chr.12: 57232431-57232431 on Build GRCh38
rs750296262 Chr.12: 57232713-57232713 on Build GRCh38
rs760986751 Chr.12: 57232465-57232465 on Build GRCh38
rs760935958 Chr.12: 57232562-57232562 on Build GRCh38
rs756952031 Chr.12: 57232706-57232706 on Build GRCh38
rs757082236 Chr.12: 57232590-57232590 on Build GRCh38
rs763559667 Chr.12: 57232694-57232694 on Build GRCh38
rs2229716 Chr.12: 57232799-57232799 on Build GRCh38
rs1019331798 Chr.12: 57232735-57232735 on Build GRCh38
rs770832478 Chr.12: 57232536-57232536 on Build GRCh38
rs754527342 Chr.12: 57232494-57232494 on Build GRCh38
rs994115895 Chr.12: 57232778-57232778 on Build GRCh38
rs767579674 Chr.12: 57232449-57232449 on Build GRCh38
rs775692309 Chr.12: 57232464-57232464 on Build GRCh38
rs778673704 Chr.12: 57232592-57232592 on Build GRCh38
rs2229715 Chr.12: 57232784-57232784 on Build GRCh38
rs151127748 Chr.12: 57232806-57232806 on Build GRCh38
rs553430961 Chr.12: 57232582-57232582 on Build GRCh38
rs760592358 Chr.12: 57232797-57232797 on Build GRCh38
rs773697432 Chr.12: 57232787-57232787 on Build GRCh38
rs1000229352 Chr.12: 57232534-57232534 on Build GRCh38
rs1025966420 Chr.12: 57232779-57232779 on Build GRCh38
rs562939107 Chr.12: 57232588-57232588 on Build GRCh38
rs932716913 Chr.12: 57232496-57232496 on Build GRCh38
rs767188559 Chr.12: 57232585-57232585 on Build GRCh38
rs372077401 Chr.12: 57232587-57232587 on Build GRCh38
rs748454200 Chr.12: 57232783-57232783 on Build GRCh38
rs764937227 Chr.12: 57232809-57232809 on Build GRCh38
rs765905599 Chr.12: 57232486-57232486 on Build GRCh38
rs561041442 Chr.12: 57232560-57232560 on Build GRCh38
rs753556116 Chr.12: 57232701-57232701 on Build GRCh38
rs1006574917 Chr.12: 57232385-57232385 on Build GRCh38
rs763316143 Chr.12: 57232372-57232372 on Build GRCh38
rs775639509 Chr.12: 57232561-57232561 on Build GRCh38
rs143192247 Chr.12: 57232564-57232564 on Build GRCh38
rs771012653 Chr.12: 57232410-57232410 on Build GRCh38
rs189884111 Chr.12: 57232629-57232629 on Build GRCh38
rs550014245 Chr.12: 57232584-57232584 on Build GRCh38
rs377091363 Chr.12: 57232513-57232513 on Build GRCh38
rs773238288 Chr.12: 57232378-57232378 on Build GRCh38
rs558952821 Chr.12: 57232581-57232582 on Build GRCh38
rs891807785 Chr.12: 57232803-57232803 on Build GRCh38
rs375126310 Chr.12: 57232416-57232416 on Build GRCh38
rs755954996 Chr.12: 57232508-57232508 on Build GRCh38

View Full Product Details