Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57474783-57475281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP9
Entrez Gene ID
64333
Gene Name
Rho GTPase activating protein 9
Gene Aliases
10C, RGL1
UniGene
Hs.437126
Gene Chromosome Location
Chr.12: 57472255-57488814 on Build GRCh38
Transcripts
NM_001080156.2 NM_001080157.1 NM_001319850.1 NM_001319851.1 NM_001319852.1 NM_032496.3 XM_005269083.2 XM_005269084.2 XM_005269085.2 XM_011538656.2 XM_011538657.2 XM_011538658.2 XM_011538659.2 XM_017019800.1 XM_017019801.1 XM_017019802.1 XM_017019803.1 XM_017019804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2151509 c.1706G>A p.R569K Chr.12: 57474877-57474877 on Build GRCh38
6603742 c.1620C>T p.F540F Chr.12: 57474963-57474963 on Build GRCh38
6603749 c.1687A>G p.I563V Chr.12: 57474896-57474896 on Build GRCh38
942050 c.1694C>A p.A565D Chr.12: 57474889-57474889 on Build GRCh38
4529239 c.1655G>A p.G552E Chr.12: 57474928-57474928 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs12820647 Chr.12: 57475179-57475179 on Build GRCh38
rs962667858 Chr.12: 57474963-57474963 on Build GRCh38
rs12820264 Chr.12: 57475046-57475046 on Build GRCh38
rs926188968 Chr.12: 57475131-57475131 on Build GRCh38
rs751692675 Chr.12: 57474840-57474840 on Build GRCh38
rs761311082 Chr.12: 57474950-57474950 on Build GRCh38
rs935965323 Chr.12: 57474814-57474814 on Build GRCh38
rs868149695 Chr.12: 57474825-57474825 on Build GRCh38
rs913668662 Chr.12: 57475039-57475039 on Build GRCh38
rs908949866 Chr.12: 57475226-57475226 on Build GRCh38
rs757610187 Chr.12: 57474996-57474996 on Build GRCh38
rs770094014 Chr.12: 57475227-57475227 on Build GRCh38
rs752970332 Chr.12: 57474991-57474991 on Build GRCh38
rs1003968459 Chr.12: 57475237-57475237 on Build GRCh38
rs563411616 Chr.12: 57475049-57475049 on Build GRCh38
rs762039853 Chr.12: 57475004-57475005 on Build GRCh38
rs112671297 Chr.12: 57474845-57474845 on Build GRCh38
rs756516836 Chr.12: 57474875-57474875 on Build GRCh38
rs552244987 Chr.12: 57475079-57475079 on Build GRCh38
rs779593493 Chr.12: 57475004-57475004 on Build GRCh38
rs547261969 Chr.12: 57475230-57475230 on Build GRCh38
rs764047965 Chr.12: 57474865-57474866 on Build GRCh38
rs768292722 Chr.12: 57475025-57475025 on Build GRCh38
rs191828785 Chr.12: 57475199-57475199 on Build GRCh38
rs774977816 Chr.12: 57474916-57474916 on Build GRCh38
rs746558556 Chr.12: 57474928-57474928 on Build GRCh38
rs1017225576 Chr.12: 57474955-57474955 on Build GRCh38
rs768239598 Chr.12: 57474934-57474934 on Build GRCh38
rs766347449 Chr.12: 57474982-57474982 on Build GRCh38
rs772944068 Chr.12: 57474962-57474962 on Build GRCh38
rs762791620 Chr.12: 57474981-57474981 on Build GRCh38
rs530827072 Chr.12: 57475064-57475064 on Build GRCh38
rs112133601 Chr.12: 57474890-57474890 on Build GRCh38
rs751461476 Chr.12: 57474987-57474987 on Build GRCh38
rs12821831 Chr.12: 57475101-57475101 on Build GRCh38
rs921302560 Chr.12: 57475150-57475150 on Build GRCh38
rs1039739637 Chr.12: 57474816-57474816 on Build GRCh38
rs115071370 Chr.12: 57474818-57474818 on Build GRCh38
rs1051104571 Chr.12: 57475208-57475208 on Build GRCh38
rs977623835 Chr.12: 57475100-57475100 on Build GRCh38
rs374366947 Chr.12: 57474866-57474866 on Build GRCh38
rs984971325 Chr.12: 57475205-57475205 on Build GRCh38
rs370660961 Chr.12: 57474937-57474937 on Build GRCh38
rs960428934 Chr.12: 57474994-57474994 on Build GRCh38
rs936303034 Chr.12: 57475134-57475134 on Build GRCh38
rs748323792 Chr.12: 57474868-57474868 on Build GRCh38
rs571126001 Chr.12: 57475096-57475096 on Build GRCh38
rs35105444 Chr.12: 57474914-57474915 on Build GRCh38
rs201097332 Chr.12: 57474862-57474862 on Build GRCh38
rs1040090202 Chr.12: 57475144-57475144 on Build GRCh38
rs753691489 Chr.12: 57474866-57474867 on Build GRCh38
rs368828844 Chr.12: 57474993-57474993 on Build GRCh38
rs548301559 Chr.12: 57475010-57475010 on Build GRCh38
rs372082509 Chr.12: 57475250-57475250 on Build GRCh38
rs913527050 Chr.12: 57474994-57474994 on Build GRCh38
rs878871158 Chr.12: 57474826-57474826 on Build GRCh38
rs765398007 Chr.12: 57475025-57475025 on Build GRCh38
rs957390073 Chr.12: 57474978-57474978 on Build GRCh38
rs904773989 Chr.12: 57474810-57474810 on Build GRCh38
rs765081715 Chr.12: 57474953-57474953 on Build GRCh38
rs777935198 Chr.12: 57474879-57474879 on Build GRCh38
rs12820256 Chr.12: 57475036-57475036 on Build GRCh38
rs767529393 Chr.12: 57474988-57474988 on Build GRCh38
rs756359689 Chr.12: 57474992-57474992 on Build GRCh38
rs771400557 Chr.12: 57474903-57474903 on Build GRCh38

View Full Product Details