Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57612839-57613336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF25
Entrez Gene ID
115557
Gene Name
Rho guanine nucleotide exchange factor 25
Gene Aliases
GEFT, p63RhoGEF
UniGene
Hs.61581
Gene Chromosome Location
Chr.12: 57610180-57617245 on Build GRCh38
Transcripts
NM_001111270.2 NM_182947.3 NR_046223.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7319232 c.492C>G p.T164T Chr.12: 57613326-57613326 on Build GRCh38
1147080 c.272T>G p.L91R Chr.12: 57612987-57612987 on Build GRCh38
4550029 c.372G>T p.L124L Chr.12: 57613323-57613323 on Build GRCh38
6604184 c.231G>A p.A77A Chr.12: 57612946-57612946 on Build GRCh38
7125277 c.186C>T p.L62L Chr.12: 57613018-57613018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150735183 Chr.12: 57613086-57613086 on Build GRCh38
rs369276230 Chr.12: 57612983-57612983 on Build GRCh38
rs144451436 Chr.12: 57612936-57612936 on Build GRCh38
rs145779061 Chr.12: 57613120-57613120 on Build GRCh38
rs375392703 Chr.12: 57613169-57613169 on Build GRCh38
rs759188636 Chr.12: 57612932-57612932 on Build GRCh38
rs936283782 Chr.12: 57612939-57612939 on Build GRCh38
rs750263568 Chr.12: 57613147-57613147 on Build GRCh38
rs761488499 Chr.12: 57613136-57613136 on Build GRCh38
rs771640862 Chr.12: 57613094-57613094 on Build GRCh38
rs775790345 Chr.12: 57612886-57612886 on Build GRCh38
rs766273748 Chr.12: 57613300-57613300 on Build GRCh38
rs748430628 Chr.12: 57613065-57613065 on Build GRCh38
rs773228709 Chr.12: 57613297-57613297 on Build GRCh38
rs757174434 Chr.12: 57613042-57613042 on Build GRCh38
rs199644875 Chr.12: 57612889-57612889 on Build GRCh38
rs78229578 Chr.12: 57612925-57612925 on Build GRCh38
rs763923142 Chr.12: 57613037-57613037 on Build GRCh38
rs992359568 Chr.12: 57612877-57612877 on Build GRCh38
rs761676190 Chr.12: 57613285-57613285 on Build GRCh38
rs913658142 Chr.12: 57613064-57613064 on Build GRCh38
rs112509455 Chr.12: 57612927-57612927 on Build GRCh38
rs769420060 Chr.12: 57613296-57613296 on Build GRCh38
rs377531308 Chr.12: 57613031-57613031 on Build GRCh38
rs147842061 Chr.12: 57612949-57612949 on Build GRCh38
rs762459033 Chr.12: 57612909-57612909 on Build GRCh38
rs767849431 Chr.12: 57613306-57613306 on Build GRCh38
rs751402460 Chr.12: 57613293-57613293 on Build GRCh38
rs151122035 Chr.12: 57612996-57612996 on Build GRCh38
rs766400509 Chr.12: 57613157-57613157 on Build GRCh38
rs776131100 Chr.12: 57613269-57613269 on Build GRCh38
rs962657738 Chr.12: 57613213-57613213 on Build GRCh38
rs533686624 Chr.12: 57613043-57613043 on Build GRCh38
rs768946799 Chr.12: 57612980-57612980 on Build GRCh38
rs116461827 Chr.12: 57613180-57613180 on Build GRCh38
rs201573584 Chr.12: 57613303-57613303 on Build GRCh38
rs746105629 Chr.12: 57612964-57612964 on Build GRCh38
rs765288175 Chr.12: 57613146-57613146 on Build GRCh38
rs372636594 Chr.12: 57612982-57612982 on Build GRCh38
rs780245693 Chr.12: 57612972-57612972 on Build GRCh38
rs746483028 Chr.12: 57613244-57613244 on Build GRCh38
rs371577552 Chr.12: 57612955-57612955 on Build GRCh38
rs775275007 Chr.12: 57613028-57613028 on Build GRCh38
rs779667522 Chr.12: 57613243-57613243 on Build GRCh38
rs771728815 Chr.12: 57613234-57613234 on Build GRCh38
rs140959163 Chr.12: 57613055-57613055 on Build GRCh38
rs371120356 Chr.12: 57613153-57613153 on Build GRCh38
rs199703624 Chr.12: 57613081-57613081 on Build GRCh38
rs531612180 Chr.12: 57613131-57613131 on Build GRCh38
rs529397542 Chr.12: 57612977-57612977 on Build GRCh38
rs1011902979 Chr.12: 57612871-57612871 on Build GRCh38
rs926144701 Chr.12: 57613101-57613101 on Build GRCh38
rs913645403 Chr.12: 57612888-57612888 on Build GRCh38
rs759601029 Chr.12: 57613305-57613305 on Build GRCh38
rs749700695 Chr.12: 57613233-57613233 on Build GRCh38
rs145325473 Chr.12: 57612991-57612991 on Build GRCh38
rs770383336 Chr.12: 57613071-57613071 on Build GRCh38
rs1038444571 Chr.12: 57613282-57613282 on Build GRCh38
rs200472423 Chr.12: 57612945-57612945 on Build GRCh38
rs759170188 Chr.12: 57612997-57613008 on Build GRCh38
rs781496619 Chr.12: 57613058-57613058 on Build GRCh38
rs750562465 Chr.12: 57613047-57613047 on Build GRCh38
rs1051080064 Chr.12: 57613163-57613163 on Build GRCh38
rs376912565 Chr.12: 57612966-57612966 on Build GRCh38
rs377278375 Chr.12: 57613111-57613111 on Build GRCh38
rs199652772 Chr.12: 57613054-57613054 on Build GRCh38
rs1017036242 Chr.12: 57612873-57612873 on Build GRCh38
rs768470341 Chr.12: 57613256-57613256 on Build GRCh38
rs139793201 Chr.12: 57612953-57612953 on Build GRCh38
rs751811460 Chr.12: 57613167-57613167 on Build GRCh38
rs769212408 Chr.12: 57612891-57612891 on Build GRCh38
rs551752562 Chr.12: 57613050-57613050 on Build GRCh38
rs778346875 Chr.12: 57613227-57613227 on Build GRCh38
rs753845308 Chr.12: 57613040-57613040 on Build GRCh38
rs369675789 Chr.12: 57613095-57613095 on Build GRCh38
rs962524675 Chr.12: 57613015-57613015 on Build GRCh38
rs762840477 Chr.12: 57613299-57613299 on Build GRCh38
rs202113715 Chr.12: 57612922-57612922 on Build GRCh38
rs61754584 Chr.12: 57613308-57613308 on Build GRCh38
rs369756788 Chr.12: 57612946-57612946 on Build GRCh38
rs777121084 Chr.12: 57612897-57612897 on Build GRCh38
rs140645509 Chr.12: 57613307-57613307 on Build GRCh38
rs756429172 Chr.12: 57613191-57613191 on Build GRCh38
rs767335428 Chr.12: 57612997-57612997 on Build GRCh38
rs979136030 Chr.12: 57613085-57613085 on Build GRCh38
rs372239766 Chr.12: 57613158-57613158 on Build GRCh38
rs921491978 Chr.12: 57613130-57613130 on Build GRCh38

View Full Product Details