Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 58880502-58881014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRIG3
Entrez Gene ID
121227
Gene Name
leucine rich repeats and immunoglobulin like domains 3
Gene Aliases
LIG3
UniGene
Hs.253736
Gene Chromosome Location
Chr.12: 58869351-58920540 on Build GRCh38
Transcripts
NM_001136051.2 NM_153377.4 XM_011537893.2 XM_017018790.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4811771 c.1546G>A p.E516K Chr.12: 58880656-58880656 on Build GRCh38
4947595 c.1474C>G p.L492V Chr.12: 58880728-58880728 on Build GRCh38
1581179 c.1679T>C p.M560T Chr.12: 58880703-58880703 on Build GRCh38
3987120 c.1491C>T p.G497G Chr.12: 58880711-58880711 on Build GRCh38
4043991 c.1710G>T p.E570D Chr.12: 58880672-58880672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768073832 Chr.12: 58880605-58880605 on Build GRCh38
rs370483520 Chr.12: 58880677-58880677 on Build GRCh38
rs936444813 Chr.12: 58880796-58880796 on Build GRCh38
rs117245609 Chr.12: 58880683-58880683 on Build GRCh38
rs900160911 Chr.12: 58880542-58880542 on Build GRCh38
rs777712307 Chr.12: 58880908-58880908 on Build GRCh38
rs886291215 Chr.12: 58880597-58880597 on Build GRCh38
rs768163806 Chr.12: 58880725-58880725 on Build GRCh38
rs371122124 Chr.12: 58880799-58880799 on Build GRCh38
rs757991739 Chr.12: 58880754-58880754 on Build GRCh38
rs541602051 Chr.12: 58880757-58880757 on Build GRCh38
rs773267873 Chr.12: 58880556-58880556 on Build GRCh38
rs771847000 Chr.12: 58880563-58880563 on Build GRCh38
rs763416675 Chr.12: 58880559-58880559 on Build GRCh38
rs751377320 Chr.12: 58880771-58880771 on Build GRCh38
rs749327139 Chr.12: 58880806-58880806 on Build GRCh38
rs373268203 Chr.12: 58880543-58880543 on Build GRCh38
rs753414425 Chr.12: 58880615-58880615 on Build GRCh38
rs60376933 Chr.12: 58880710-58880710 on Build GRCh38
rs754067261 Chr.12: 58880700-58880701 on Build GRCh38
rs868591398 Chr.12: 58880661-58880661 on Build GRCh38
rs775349426 Chr.12: 58880746-58880746 on Build GRCh38
rs374118120 Chr.12: 58880967-58880967 on Build GRCh38
rs776193260 Chr.12: 58880859-58880859 on Build GRCh38
rs113261838 Chr.12: 58880760-58880760 on Build GRCh38
rs750152739 Chr.12: 58880744-58880744 on Build GRCh38
rs200549484 Chr.12: 58880628-58880628 on Build GRCh38
rs202006711 Chr.12: 58880827-58880827 on Build GRCh38
rs769731705 Chr.12: 58880682-58880682 on Build GRCh38
rs151252867 Chr.12: 58880974-58880974 on Build GRCh38
rs201261850 Chr.12: 58880869-58880869 on Build GRCh38
rs573595042 Chr.12: 58880961-58880961 on Build GRCh38
rs111986525 Chr.12: 58880673-58880673 on Build GRCh38
rs761255915 Chr.12: 58880861-58880861 on Build GRCh38
rs200611962 Chr.12: 58880676-58880676 on Build GRCh38
rs967961963 Chr.12: 58880723-58880723 on Build GRCh38
rs780953080 Chr.12: 58880895-58880895 on Build GRCh38
rs781149509 Chr.12: 58880794-58880794 on Build GRCh38
rs746739539 Chr.12: 58880674-58880674 on Build GRCh38
rs749353285 Chr.12: 58880926-58880926 on Build GRCh38
rs753222629 Chr.12: 58880730-58880730 on Build GRCh38
rs762474881 Chr.12: 58880877-58880877 on Build GRCh38
rs111815783 Chr.12: 58880707-58880707 on Build GRCh38
rs752698837 Chr.12: 58880897-58880897 on Build GRCh38
rs779709378 Chr.12: 58880672-58880672 on Build GRCh38
rs566187872 Chr.12: 58880711-58880711 on Build GRCh38
rs867057683 Chr.12: 58880924-58880924 on Build GRCh38
rs752171079 Chr.12: 58880595-58880595 on Build GRCh38
rs377193373 Chr.12: 58880675-58880675 on Build GRCh38
rs774290942 Chr.12: 58880855-58880855 on Build GRCh38
rs368520634 Chr.12: 58880876-58880876 on Build GRCh38
rs764637222 Chr.12: 58880742-58880742 on Build GRCh38
rs771075384 Chr.12: 58880825-58880825 on Build GRCh38
rs779698653 Chr.12: 58880769-58880769 on Build GRCh38
rs765977176 Chr.12: 58880887-58880887 on Build GRCh38
rs763361453 Chr.12: 58880936-58880936 on Build GRCh38
rs761822687 Chr.12: 58880532-58880532 on Build GRCh38
rs745483651 Chr.12: 58880635-58880635 on Build GRCh38
rs142306271 Chr.12: 58880724-58880724 on Build GRCh38
rs181837033 Chr.12: 58880935-58880935 on Build GRCh38
rs758268109 Chr.12: 58880665-58880665 on Build GRCh38
rs766632145 Chr.12: 58880588-58880588 on Build GRCh38
rs755540790 Chr.12: 58880604-58880604 on Build GRCh38
rs376173782 Chr.12: 58880736-58880736 on Build GRCh38
rs1051649387 Chr.12: 58880562-58880562 on Build GRCh38
rs756013904 Chr.12: 58880898-58880898 on Build GRCh38
rs751183492 Chr.12: 58880892-58880892 on Build GRCh38
rs754726146 Chr.12: 58880779-58880779 on Build GRCh38
rs934087528 Chr.12: 58880582-58880582 on Build GRCh38
rs142965808 Chr.12: 58880856-58880856 on Build GRCh38
rs775217155 Chr.12: 58880901-58880902 on Build GRCh38
rs972514451 Chr.12: 58880969-58880969 on Build GRCh38
rs374580006 Chr.12: 58880854-58880854 on Build GRCh38
rs756785803 Chr.12: 58880621-58880621 on Build GRCh38
rs982272481 Chr.12: 58880770-58880770 on Build GRCh38
rs754818016 Chr.12: 58880894-58880894 on Build GRCh38

View Full Product Details