Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 62500355-62500882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MON2
Entrez Gene ID
23041
Gene Name
MON2 homolog, regulator of endosome-to-Golgi trafficking
Gene Aliases
-
UniGene
Hs.389378
Gene Chromosome Location
Chr.12: 62466817-62598956 on Build GRCh38
Transcripts
NM_001278469.1 NM_001278470.1 NM_001278471.1 NM_001278472.1 NM_015026.2 XM_017019041.1 XM_017019042.1 XM_017019043.1 XM_017019044.1 XM_017019045.1 XM_017019046.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7220433 c.639T>A p.A213A Chr.12: 62500856-62500856 on Build GRCh38
6137613 c.643G>C p.D215H Chr.12: 62500860-62500860 on Build GRCh38
5942477 c.663G>A p.Q221Q Chr.12: 62500880-62500880 on Build GRCh38
7220432 c.639T>A p.A213A Chr.12: 62500856-62500856 on Build GRCh38
4443835 c.589G>A p.V197I Chr.12: 62500806-62500806 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs74095804 Chr.12: 62500634-62500634 on Build GRCh38
rs190608016 Chr.12: 62500671-62500671 on Build GRCh38
rs559213204 Chr.12: 62500628-62500628 on Build GRCh38
rs200692514 Chr.12: 62500778-62500778 on Build GRCh38
rs978165303 Chr.12: 62500604-62500604 on Build GRCh38
rs202015814 Chr.12: 62500803-62500803 on Build GRCh38
rs753721541 Chr.12: 62500806-62500806 on Build GRCh38
rs1054738446 Chr.12: 62500661-62500661 on Build GRCh38
rs1002649466 Chr.12: 62500383-62500383 on Build GRCh38
rs977963216 Chr.12: 62500420-62500420 on Build GRCh38
rs967955154 Chr.12: 62500418-62500418 on Build GRCh38
rs115864448 Chr.12: 62500760-62500760 on Build GRCh38
rs867174805 Chr.12: 62500854-62500854 on Build GRCh38
rs17120341 Chr.12: 62500856-62500856 on Build GRCh38
rs923608655 Chr.12: 62500427-62500427 on Build GRCh38
rs992335184 Chr.12: 62500487-62500487 on Build GRCh38
rs777706383 Chr.12: 62500800-62500800 on Build GRCh38
rs933320411 Chr.12: 62500649-62500649 on Build GRCh38
rs960374073 Chr.12: 62500457-62500457 on Build GRCh38
rs909506466 Chr.12: 62500589-62500589 on Build GRCh38
rs757284923 Chr.12: 62500818-62500818 on Build GRCh38
rs569655843 Chr.12: 62500426-62500426 on Build GRCh38
rs201958152 Chr.12: 62500855-62500855 on Build GRCh38
rs538221616 Chr.12: 62500505-62500505 on Build GRCh38
rs780456859 Chr.12: 62500844-62500844 on Build GRCh38
rs749232048 Chr.12: 62500536-62500536 on Build GRCh38
rs916312152 Chr.12: 62500488-62500488 on Build GRCh38
rs984967075 Chr.12: 62500564-62500564 on Build GRCh38
rs574678759 Chr.12: 62500743-62500743 on Build GRCh38
rs923468629 Chr.12: 62500521-62500521 on Build GRCh38
rs762182674 Chr.12: 62500739-62500739 on Build GRCh38
rs568074105 Chr.12: 62500837-62500837 on Build GRCh38
rs201677867 Chr.12: 62500614-62500615 on Build GRCh38
rs1044234582 Chr.12: 62500728-62500728 on Build GRCh38
rs372969306 Chr.12: 62500749-62500749 on Build GRCh38
rs889198812 Chr.12: 62500693-62500693 on Build GRCh38
rs940956435 Chr.12: 62500591-62500591 on Build GRCh38
rs760877619 Chr.12: 62500736-62500736 on Build GRCh38
rs769248535 Chr.12: 62500738-62500738 on Build GRCh38
rs781131771 Chr.12: 62500413-62500413 on Build GRCh38
rs201434593 Chr.12: 62500753-62500753 on Build GRCh38
rs1022140719 Chr.12: 62500767-62500767 on Build GRCh38
rs903654041 Chr.12: 62500853-62500853 on Build GRCh38
rs901872703 Chr.12: 62500644-62500644 on Build GRCh38
rs778558969 Chr.12: 62500823-62500823 on Build GRCh38
rs755640335 Chr.12: 62500787-62500787 on Build GRCh38
rs948678630 Chr.12: 62500706-62500706 on Build GRCh38
rs532546106 Chr.12: 62500764-62500764 on Build GRCh38
rs757492964 Chr.12: 62500784-62500786 on Build GRCh38
rs1006711481 Chr.12: 62500384-62500384 on Build GRCh38
rs143913309 Chr.12: 62500682-62500682 on Build GRCh38

View Full Product Details