Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 64404244-64404778 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XPOT
Entrez Gene ID
11260
Gene Name
exportin for tRNA
Gene Aliases
XPO3
UniGene
Hs.592579
Gene Chromosome Location
Chr.12: 64404350-64451125 on Build GRCh38
Transcripts
NM_007235.4

SNP Detail

SNP ID Chromosome Location
rs1001420500 Chr.12: 64404738-64404738 on Build GRCh38
rs1021150537 Chr.12: 64404727-64404727 on Build GRCh38
rs571447496 Chr.12: 64404509-64404509 on Build GRCh38
rs910743242 Chr.12: 64404350-64404350 on Build GRCh38
rs1026229292 Chr.12: 64404735-64404735 on Build GRCh38
rs1002047700 Chr.12: 64404587-64404587 on Build GRCh38
rs992176805 Chr.12: 64404467-64404484 on Build GRCh38
rs945372175 Chr.12: 64404406-64404406 on Build GRCh38
rs986426999 Chr.12: 64404467-64404467 on Build GRCh38
rs898986384 Chr.12: 64404719-64404719 on Build GRCh38
rs896986571 Chr.12: 64404508-64404508 on Build GRCh38
rs566976705 Chr.12: 64404728-64404728 on Build GRCh38
rs966199993 Chr.12: 64404473-64404481 on Build GRCh38
rs944863481 Chr.12: 64404383-64404383 on Build GRCh38
rs994739233 Chr.12: 64404600-64404600 on Build GRCh38
rs896351038 Chr.12: 64404454-64404454 on Build GRCh38
rs1010092322 Chr.12: 64404576-64404576 on Build GRCh38
rs551306702 Chr.12: 64404493-64404493 on Build GRCh38
rs906573483 Chr.12: 64404579-64404579 on Build GRCh38
rs370121068 Chr.12: 64404402-64404402 on Build GRCh38
rs912757833 Chr.12: 64404267-64404267 on Build GRCh38
rs1008570635 Chr.12: 64404747-64404747 on Build GRCh38
rs966233571 Chr.12: 64404563-64404570 on Build GRCh38
rs555693814 Chr.12: 64404645-64404645 on Build GRCh38
rs756243461 Chr.12: 64404551-64404552 on Build GRCh38
rs944217284 Chr.12: 64404273-64404273 on Build GRCh38
rs575865293 Chr.12: 64404666-64404666 on Build GRCh38
rs1026429448 Chr.12: 64404601-64404601 on Build GRCh38
rs538504325 Chr.12: 64404712-64404712 on Build GRCh38
rs548914270 Chr.12: 64404334-64404334 on Build GRCh38
rs994790759 Chr.12: 64404721-64404721 on Build GRCh38
rs547156706 Chr.12: 64404562-64404562 on Build GRCh38
rs889493802 Chr.12: 64404700-64404700 on Build GRCh38
rs1018997154 Chr.12: 64404754-64404754 on Build GRCh38
rs146483101 Chr.12: 64404566-64404566 on Build GRCh38
rs917818922 Chr.12: 64404500-64404500 on Build GRCh38
rs976182837 Chr.12: 64404475-64404476 on Build GRCh38
rs987666979 Chr.12: 64404614-64404614 on Build GRCh38
rs963310509 Chr.12: 64404595-64404595 on Build GRCh38
rs913924708 Chr.12: 64404683-64404683 on Build GRCh38
rs964448958 Chr.12: 64404758-64404758 on Build GRCh38
rs950651951 Chr.12: 64404736-64404736 on Build GRCh38
rs967263748 Chr.12: 64404576-64404577 on Build GRCh38
rs975565975 Chr.12: 64404638-64404638 on Build GRCh38
rs949205673 Chr.12: 64404503-64404503 on Build GRCh38
rs1013398492 Chr.12: 64404455-64404455 on Build GRCh38
rs966854467 Chr.12: 64404482-64404490 on Build GRCh38
rs371595748 Chr.12: 64404265-64404265 on Build GRCh38
rs1036931044 Chr.12: 64404644-64404644 on Build GRCh38
rs990783325 Chr.12: 64404596-64404596 on Build GRCh38
rs568580764 Chr.12: 64404547-64404548 on Build GRCh38
rs531598672 Chr.12: 64404417-64404417 on Build GRCh38
rs925332765 Chr.12: 64404484-64404484 on Build GRCh38
rs1017782814 Chr.12: 64404470-64404481 on Build GRCh38
rs1033485428 Chr.12: 64404592-64404592 on Build GRCh38
rs556310177 Chr.12: 64404624-64404624 on Build GRCh38
rs960665182 Chr.12: 64404410-64404430 on Build GRCh38
rs6581560 Chr.12: 64404338-64404338 on Build GRCh38
rs1006943970 Chr.12: 64404548-64404555 on Build GRCh38
rs966918335 Chr.12: 64404732-64404733 on Build GRCh38
rs903845717 Chr.12: 64404410-64404410 on Build GRCh38
rs950405410 Chr.12: 64404613-64404613 on Build GRCh38
rs889471568 Chr.12: 64404523-64404523 on Build GRCh38
rs1046466166 Chr.12: 64404578-64404578 on Build GRCh38
rs1039808045 Chr.12: 64404313-64404313 on Build GRCh38
rs1036898403 Chr.12: 64404506-64404506 on Build GRCh38
rs976150178 Chr.12: 64404389-64404389 on Build GRCh38
rs1048605359 Chr.12: 64404518-64404518 on Build GRCh38
rs937998901 Chr.12: 64404491-64404491 on Build GRCh38
rs918455235 Chr.12: 64404657-64404657 on Build GRCh38
rs936945965 Chr.12: 64404659-64404659 on Build GRCh38
rs533958353 Chr.12: 64404559-64404559 on Build GRCh38
rs1002268328 Chr.12: 64404714-64404714 on Build GRCh38
rs979923815 Chr.12: 64404483-64404483 on Build GRCh38
rs770410319 Chr.12: 64404481-64404482 on Build GRCh38
rs997748185 Chr.12: 64404725-64404725 on Build GRCh38
rs1055435522 Chr.12: 64404437-64404437 on Build GRCh38
rs778369120 Chr.12: 64404633-64404633 on Build GRCh38
rs896872647 Chr.12: 64404466-64404467 on Build GRCh38
rs906632129 Chr.12: 64404713-64404713 on Build GRCh38
rs892447006 Chr.12: 64404569-64404569 on Build GRCh38
rs1021119408 Chr.12: 64404567-64404570 on Build GRCh38
rs796579421 Chr.12: 64404546-64404547 on Build GRCh38
rs945392811 Chr.12: 64404697-64404697 on Build GRCh38
rs140906992 Chr.12: 64404620-64404620 on Build GRCh38
rs937983940 Chr.12: 64404415-64404415 on Build GRCh38
rs528899274 Chr.12: 64404325-64404325 on Build GRCh38
rs961047062 Chr.12: 64404270-64404270 on Build GRCh38
rs1015352422 Chr.12: 64404459-64404459 on Build GRCh38
rs921408058 Chr.12: 64404655-64404655 on Build GRCh38
rs770848940 Chr.12: 64404715-64404715 on Build GRCh38
rs755734840 Chr.12: 64404548-64404551 on Build GRCh38
rs559802664 Chr.12: 64404322-64404322 on Build GRCh38
rs1032433231 Chr.12: 64404738-64404739 on Build GRCh38
rs546828743 Chr.12: 64404705-64404705 on Build GRCh38
rs1026688977 Chr.12: 64404467-64404481 on Build GRCh38

View Full Product Details