Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 69677042-69677458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BEST3
Entrez Gene ID
144453
Gene Name
bestrophin 3
Gene Aliases
VMD2L3
UniGene
Hs.280782
Gene Chromosome Location
Chr.12: 69643508-69699416 on Build GRCh38
Transcripts
NM_001282613.1 NM_001282614.1 NM_032735.2 NM_152439.3 XM_006719256.3 XM_011537961.1 XM_011537962.2 XM_011537963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4044534 c.647G>A p.R216Q Chr.12: 69677247-69677247 on Build GRCh38
4044531 c.711C>T p.T237T Chr.12: 69677183-69677183 on Build GRCh38
4627136 c.736A>G p.T246A Chr.12: 69677047-69677047 on Build GRCh38
6265679 c.666C>A p.S222R Chr.12: 69677228-69677228 on Build GRCh38
3464635 c.693G>A p.G231G Chr.12: 69677201-69677201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755555260 Chr.12: 69677234-69677234 on Build GRCh38
rs767324127 Chr.12: 69677077-69677077 on Build GRCh38
rs377359402 Chr.12: 69677252-69677252 on Build GRCh38
rs763222464 Chr.12: 69677288-69677288 on Build GRCh38
rs116917484 Chr.12: 69677297-69677297 on Build GRCh38
rs710718 Chr.12: 69677162-69677162 on Build GRCh38
rs771720622 Chr.12: 69677166-69677166 on Build GRCh38
rs753205163 Chr.12: 69677242-69677242 on Build GRCh38
rs11609972 Chr.12: 69677113-69677113 on Build GRCh38
rs747033835 Chr.12: 69677184-69677184 on Build GRCh38
rs755438978 Chr.12: 69677118-69677118 on Build GRCh38
rs778735903 Chr.12: 69677161-69677161 on Build GRCh38
rs202161560 Chr.12: 69677266-69677266 on Build GRCh38
rs748065485 Chr.12: 69677269-69677269 on Build GRCh38
rs745517606 Chr.12: 69677248-69677248 on Build GRCh38
rs1033666888 Chr.12: 69677403-69677403 on Build GRCh38
rs927233694 Chr.12: 69677164-69677164 on Build GRCh38
rs552071746 Chr.12: 69677320-69677320 on Build GRCh38
rs746472418 Chr.12: 69677259-69677259 on Build GRCh38
rs758498539 Chr.12: 69677101-69677101 on Build GRCh38
rs1009703094 Chr.12: 69677338-69677338 on Build GRCh38
rs200887965 Chr.12: 69677075-69677075 on Build GRCh38
rs768172565 Chr.12: 69677261-69677261 on Build GRCh38
rs763098809 Chr.12: 69677197-69677197 on Build GRCh38
rs767876879 Chr.12: 69677241-69677241 on Build GRCh38
rs751581328 Chr.12: 69677232-69677232 on Build GRCh38
rs140310153 Chr.12: 69677196-69677196 on Build GRCh38
rs759561779 Chr.12: 69677300-69677300 on Build GRCh38
rs766515260 Chr.12: 69677218-69677218 on Build GRCh38
rs78300740 Chr.12: 69677081-69677081 on Build GRCh38
rs1002528550 Chr.12: 69677395-69677395 on Build GRCh38
rs79464531 Chr.12: 69677114-69677114 on Build GRCh38
rs370129964 Chr.12: 69677279-69677279 on Build GRCh38
rs892646497 Chr.12: 69677324-69677324 on Build GRCh38
rs556546964 Chr.12: 69677243-69677243 on Build GRCh38
rs752565721 Chr.12: 69677175-69677179 on Build GRCh38
rs17106943 Chr.12: 69677298-69677298 on Build GRCh38
rs372783693 Chr.12: 69677247-69677247 on Build GRCh38
rs1005503702 Chr.12: 69677336-69677336 on Build GRCh38
rs903935398 Chr.12: 69677365-69677365 on Build GRCh38
rs748436719 Chr.12: 69677135-69677135 on Build GRCh38
rs145342394 Chr.12: 69677314-69677314 on Build GRCh38
rs776531139 Chr.12: 69677193-69677193 on Build GRCh38
rs779761425 Chr.12: 69677253-69677253 on Build GRCh38
rs1022389255 Chr.12: 69677358-69677358 on Build GRCh38
rs768412741 Chr.12: 69677219-69677219 on Build GRCh38
rs755812933 Chr.12: 69677216-69677232 on Build GRCh38
rs772990158 Chr.12: 69677287-69677287 on Build GRCh38
rs1002238892 Chr.12: 69677384-69677384 on Build GRCh38
rs906835323 Chr.12: 69677394-69677394 on Build GRCh38
rs769681556 Chr.12: 69677362-69677362 on Build GRCh38
rs147988441 Chr.12: 69677354-69677354 on Build GRCh38
rs369833223 Chr.12: 69677119-69677119 on Build GRCh38
rs768457172 Chr.12: 69677192-69677192 on Build GRCh38
rs199950482 Chr.12: 69677195-69677195 on Build GRCh38
rs1014611827 Chr.12: 69677346-69677346 on Build GRCh38
rs375603707 Chr.12: 69677182-69677182 on Build GRCh38

View Full Product Details