Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 70366472-70367014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNMB4
Entrez Gene ID
27345
Gene Name
potassium calcium-activated channel subfamily M regulatory beta subunit 4
Gene Aliases
-
UniGene
Hs.525529
Gene Chromosome Location
Chr.12: 70366220-70434292 on Build GRCh38
Transcripts
NM_014505.5 XM_011538188.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6377566 c.55C>T p.R19W Chr.12: 70366789-70366789 on Build GRCh38
1299836 c.234C>T p.G78G Chr.12: 70366968-70366968 on Build GRCh38
6502523 c.87C>A p.V29V Chr.12: 70366821-70366821 on Build GRCh38
1606578 c.189C>T p.G63G Chr.12: 70366923-70366923 on Build GRCh38
1606577 c.112T>C p.C38R Chr.12: 70366846-70366846 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748946457 Chr.12: 70366723-70366723 on Build GRCh38
rs11178200 Chr.12: 70366983-70366983 on Build GRCh38
rs778420332 Chr.12: 70366840-70366840 on Build GRCh38
rs905208972 Chr.12: 70366600-70366600 on Build GRCh38
rs1021147459 Chr.12: 70366811-70366811 on Build GRCh38
rs765553568 Chr.12: 70366887-70366887 on Build GRCh38
rs374031800 Chr.12: 70366810-70366810 on Build GRCh38
rs372077662 Chr.12: 70366882-70366882 on Build GRCh38
rs777327993 Chr.12: 70366722-70366722 on Build GRCh38
rs771540859 Chr.12: 70366857-70366857 on Build GRCh38
rs772056073 Chr.12: 70366730-70366730 on Build GRCh38
rs763162546 Chr.12: 70366893-70366893 on Build GRCh38
rs982940274 Chr.12: 70366803-70366803 on Build GRCh38
rs35578708 Chr.12: 70366639-70366640 on Build GRCh38
rs1053886258 Chr.12: 70366639-70366639 on Build GRCh38
rs144200148 Chr.12: 70366884-70366884 on Build GRCh38
rs762125328 Chr.12: 70366766-70366766 on Build GRCh38
rs532514417 Chr.12: 70366706-70366706 on Build GRCh38
rs930192522 Chr.12: 70366507-70366507 on Build GRCh38
rs760504525 Chr.12: 70366744-70366744 on Build GRCh38
rs745499444 Chr.12: 70366964-70366964 on Build GRCh38
rs768436591 Chr.12: 70366878-70366878 on Build GRCh38
rs546679597 Chr.12: 70366536-70366536 on Build GRCh38
rs775457202 Chr.12: 70366859-70366859 on Build GRCh38
rs763928883 Chr.12: 70366746-70366746 on Build GRCh38
rs1001320332 Chr.12: 70366625-70366626 on Build GRCh38
rs973179380 Chr.12: 70366505-70366505 on Build GRCh38
rs753166736 Chr.12: 70366933-70366933 on Build GRCh38
rs755344952 Chr.12: 70366800-70366800 on Build GRCh38
rs887809772 Chr.12: 70366689-70366689 on Build GRCh38
rs372457321 Chr.12: 70366717-70366717 on Build GRCh38
rs778577165 Chr.12: 70366957-70366957 on Build GRCh38
rs780649197 Chr.12: 70366716-70366716 on Build GRCh38
rs966850209 Chr.12: 70366828-70366828 on Build GRCh38
rs181227533 Chr.12: 70366860-70366860 on Build GRCh38
rs1045013735 Chr.12: 70366543-70366543 on Build GRCh38
rs775402368 Chr.12: 70366733-70366733 on Build GRCh38
rs963430853 Chr.12: 70366497-70366497 on Build GRCh38
rs1030236200 Chr.12: 70366705-70366705 on Build GRCh38
rs753525779 Chr.12: 70366747-70366747 on Build GRCh38
rs765309715 Chr.12: 70366777-70366777 on Build GRCh38
rs551085353 Chr.12: 70366721-70366721 on Build GRCh38
rs959103639 Chr.12: 70366986-70366986 on Build GRCh38
rs757331385 Chr.12: 70366685-70366685 on Build GRCh38
rs755516521 Chr.12: 70366911-70366911 on Build GRCh38
rs758535497 Chr.12: 70366791-70366791 on Build GRCh38
rs1055177787 Chr.12: 70366699-70366699 on Build GRCh38
rs751639013 Chr.12: 70366902-70366902 on Build GRCh38
rs756951139 Chr.12: 70366821-70366821 on Build GRCh38
rs1047300068 Chr.12: 70366528-70366528 on Build GRCh38
rs756431148 Chr.12: 70366938-70366938 on Build GRCh38
rs752078274 Chr.12: 70366794-70366794 on Build GRCh38
rs949373201 Chr.12: 70366540-70366541 on Build GRCh38
rs750428962 Chr.12: 70366708-70366708 on Build GRCh38
rs868360661 Chr.12: 70366909-70366909 on Build GRCh38
rs1007495741 Chr.12: 70366725-70366726 on Build GRCh38
rs745449083 Chr.12: 70366845-70366845 on Build GRCh38
rs1009444132 Chr.12: 70366673-70366673 on Build GRCh38
rs534480835 Chr.12: 70366917-70366917 on Build GRCh38
rs766541538 Chr.12: 70366898-70366898 on Build GRCh38
rs757999060 Chr.12: 70366987-70366987 on Build GRCh38
rs748397922 Chr.12: 70366818-70366818 on Build GRCh38
rs920054465 Chr.12: 70366506-70366506 on Build GRCh38
rs200100737 Chr.12: 70366891-70366891 on Build GRCh38
rs370331603 Chr.12: 70366758-70366758 on Build GRCh38
rs112372380 Chr.12: 70366727-70366727 on Build GRCh38
rs565254738 Chr.12: 70366646-70366646 on Build GRCh38
rs779070594 Chr.12: 70366693-70366693 on Build GRCh38
rs536892206 Chr.12: 70366784-70366784 on Build GRCh38

View Full Product Details