Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71610269-71610771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZFC3H1
Entrez Gene ID
196441
Gene Name
zinc finger C3H1-type containing
Gene Aliases
CCDC131, CSRC2, PSRC2
UniGene
Hs.245798
Gene Chromosome Location
Chr.12: 71609599-71663969 on Build GRCh38
Transcripts
NM_144982.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2101248 c.5966A>T p.H1989L Chr.12: 71610432-71610432 on Build GRCh38
5418076 c.5849C>T p.A1950V Chr.12: 71610549-71610549 on Build GRCh38
316669 c.5925G>T p.E1975D Chr.12: 71610473-71610473 on Build GRCh38
2101250 c.5921A>G p.N1974S Chr.12: 71610477-71610477 on Build GRCh38
5985070 c.5854G>C p.E1952Q Chr.12: 71610544-71610544 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200754250 Chr.12: 71610666-71610666 on Build GRCh38
rs754795352 Chr.12: 71610397-71610397 on Build GRCh38
rs779019770 Chr.12: 71610669-71610669 on Build GRCh38
rs762389932 Chr.12: 71610548-71610548 on Build GRCh38
rs983805899 Chr.12: 71610427-71610427 on Build GRCh38
rs746897223 Chr.12: 71610395-71610395 on Build GRCh38
rs559086986 Chr.12: 71610497-71610497 on Build GRCh38
rs752486633 Chr.12: 71610588-71610588 on Build GRCh38
rs749336047 Chr.12: 71610426-71610426 on Build GRCh38
rs1035679793 Chr.12: 71610322-71610322 on Build GRCh38
rs769012731 Chr.12: 71610710-71610710 on Build GRCh38
rs781026825 Chr.12: 71610586-71610586 on Build GRCh38
rs777873223 Chr.12: 71610603-71610603 on Build GRCh38
rs180798100 Chr.12: 71610453-71610453 on Build GRCh38
rs17227269 Chr.12: 71610311-71610311 on Build GRCh38
rs900013015 Chr.12: 71610320-71610320 on Build GRCh38
rs377232932 Chr.12: 71610713-71610713 on Build GRCh38
rs1025222086 Chr.12: 71610383-71610383 on Build GRCh38
rs993716434 Chr.12: 71610371-71610371 on Build GRCh38
rs761622277 Chr.12: 71610507-71610507 on Build GRCh38
rs747862217 Chr.12: 71610429-71610430 on Build GRCh38
rs776071927 Chr.12: 71610706-71610706 on Build GRCh38
rs771095831 Chr.12: 71610698-71610698 on Build GRCh38
rs988075118 Chr.12: 71610687-71610687 on Build GRCh38
rs951067622 Chr.12: 71610412-71610412 on Build GRCh38
rs556359938 Chr.12: 71610665-71610665 on Build GRCh38
rs368660102 Chr.12: 71610527-71610527 on Build GRCh38
rs774744524 Chr.12: 71610447-71610447 on Build GRCh38
rs905348789 Chr.12: 71610301-71610301 on Build GRCh38
rs748421946 Chr.12: 71610408-71610408 on Build GRCh38
rs772702024 Chr.12: 71610697-71610697 on Build GRCh38
rs887433472 Chr.12: 71610345-71610345 on Build GRCh38
rs751060461 Chr.12: 71610746-71610746 on Build GRCh38
rs199727681 Chr.12: 71610735-71610735 on Build GRCh38
rs755874773 Chr.12: 71610602-71610602 on Build GRCh38
rs765868931 Chr.12: 71610559-71610559 on Build GRCh38
rs755730438 Chr.12: 71610591-71610592 on Build GRCh38
rs958539453 Chr.12: 71610632-71610632 on Build GRCh38
rs754891556 Chr.12: 71610579-71610579 on Build GRCh38
rs770987929 Chr.12: 71610440-71610440 on Build GRCh38
rs933857478 Chr.12: 71610562-71610562 on Build GRCh38
rs769995118 Chr.12: 71610421-71610421 on Build GRCh38
rs757782971 Chr.12: 71610337-71610342 on Build GRCh38
rs896766924 Chr.12: 71610323-71610323 on Build GRCh38
rs78500528 Chr.12: 71610425-71610425 on Build GRCh38
rs538020425 Chr.12: 71610560-71610560 on Build GRCh38
rs1006339739 Chr.12: 71610348-71610348 on Build GRCh38
rs964201091 Chr.12: 71610523-71610523 on Build GRCh38
rs1035500218 Chr.12: 71610600-71610600 on Build GRCh38
rs780930691 Chr.12: 71610399-71610399 on Build GRCh38
rs762632066 Chr.12: 71610718-71610718 on Build GRCh38
rs760080438 Chr.12: 71610452-71610452 on Build GRCh38
rs1014841917 Chr.12: 71610407-71610407 on Build GRCh38
rs750031886 Chr.12: 71610547-71610547 on Build GRCh38
rs184313013 Chr.12: 71610545-71610545 on Build GRCh38
rs1016712439 Chr.12: 71610512-71610512 on Build GRCh38
rs1050630503 Chr.12: 71610340-71610340 on Build GRCh38
rs553396550 Chr.12: 71610695-71610695 on Build GRCh38
rs761133469 Chr.12: 71610707-71610707 on Build GRCh38
rs749479655 Chr.12: 71610616-71610616 on Build GRCh38

View Full Product Details