Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 85023051-85023551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN19
Entrez Gene ID
144448
Gene Name
tetraspanin 19
Gene Aliases
-
UniGene
Hs.156962
Gene Chromosome Location
Chr.12: 85014311-85036277 on Build GRCh38
Transcripts
NM_001100917.1 XM_005268669.4 XM_017018862.1 XM_017018863.1 XM_017018864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6520533 c.300G>T p.Q100H Chr.12: 85023365-85023365 on Build GRCh38
267652 c.314C>T p.A105V Chr.12: 85023351-85023351 on Build GRCh38
6372515 c.269C>T p.A90V Chr.12: 85023396-85023396 on Build GRCh38
5811785 c.312A>C p.S104S Chr.12: 85023353-85023353 on Build GRCh38
86620 c.291T>C p.F97F Chr.12: 85023374-85023374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs976335369 Chr.12: 85023418-85023418 on Build GRCh38
rs769539587 Chr.12: 85023361-85023361 on Build GRCh38
rs200541817 Chr.12: 85023398-85023398 on Build GRCh38
rs746147732 Chr.12: 85023334-85023334 on Build GRCh38
rs909281464 Chr.12: 85023126-85023126 on Build GRCh38
rs957117986 Chr.12: 85023255-85023255 on Build GRCh38
rs781688358 Chr.12: 85023138-85023138 on Build GRCh38
rs770415163 Chr.12: 85023385-85023385 on Build GRCh38
rs969429807 Chr.12: 85023370-85023370 on Build GRCh38
rs768271502 Chr.12: 85023328-85023328 on Build GRCh38
rs369573770 Chr.12: 85023277-85023277 on Build GRCh38
rs544523576 Chr.12: 85023237-85023237 on Build GRCh38
rs1012802459 Chr.12: 85023165-85023165 on Build GRCh38
rs983441470 Chr.12: 85023480-85023480 on Build GRCh38
rs191054062 Chr.12: 85023499-85023499 on Build GRCh38
rs372781647 Chr.12: 85023400-85023400 on Build GRCh38
rs1031167370 Chr.12: 85023315-85023315 on Build GRCh38
rs747908512 Chr.12: 85023356-85023356 on Build GRCh38
rs539468278 Chr.12: 85023259-85023259 on Build GRCh38
rs779641175 Chr.12: 85023182-85023182 on Build GRCh38
rs560424052 Chr.12: 85023157-85023157 on Build GRCh38
rs375386781 Chr.12: 85023402-85023402 on Build GRCh38
rs568971846 Chr.12: 85023175-85023175 on Build GRCh38
rs976751684 Chr.12: 85023083-85023083 on Build GRCh38
rs973914031 Chr.12: 85023481-85023481 on Build GRCh38
rs747926600 Chr.12: 85023420-85023420 on Build GRCh38
rs753085601 Chr.12: 85023293-85023293 on Build GRCh38
rs182983181 Chr.12: 85023362-85023362 on Build GRCh38
rs767162536 Chr.12: 85023397-85023397 on Build GRCh38
rs754404410 Chr.12: 85023409-85023409 on Build GRCh38
rs186462173 Chr.12: 85023443-85023443 on Build GRCh38
rs1050812335 Chr.12: 85023513-85023513 on Build GRCh38
rs757428763 Chr.12: 85023325-85023325 on Build GRCh38
rs1025457414 Chr.12: 85023368-85023368 on Build GRCh38
rs773897888 Chr.12: 85023392-85023392 on Build GRCh38
rs757803847 Chr.12: 85023142-85023142 on Build GRCh38
rs879933598 Chr.12: 85023511-85023511 on Build GRCh38
rs373039970 Chr.12: 85023306-85023306 on Build GRCh38
rs796799964 Chr.12: 85023476-85023476 on Build GRCh38
rs998465439 Chr.12: 85023189-85023189 on Build GRCh38
rs756724641 Chr.12: 85023299-85023299 on Build GRCh38
rs965261786 Chr.12: 85023380-85023380 on Build GRCh38
rs776335499 Chr.12: 85023340-85023340 on Build GRCh38
rs1018556674 Chr.12: 85023266-85023266 on Build GRCh38
rs929785759 Chr.12: 85023504-85023504 on Build GRCh38
rs544768551 Chr.12: 85023455-85023455 on Build GRCh38
rs904084544 Chr.12: 85023298-85023298 on Build GRCh38
rs894302895 Chr.12: 85023143-85023143 on Build GRCh38
rs12578769 Chr.12: 85023524-85023524 on Build GRCh38
rs759156650 Chr.12: 85023396-85023396 on Build GRCh38
rs765291707 Chr.12: 85023432-85023432 on Build GRCh38
rs778988909 Chr.12: 85023332-85023332 on Build GRCh38
rs1047782313 Chr.12: 85023103-85023103 on Build GRCh38
rs890813376 Chr.12: 85023206-85023206 on Build GRCh38
rs780898960 Chr.12: 85023351-85023351 on Build GRCh38
rs368223404 Chr.12: 85023364-85023364 on Build GRCh38
rs773145660 Chr.12: 85023363-85023363 on Build GRCh38
rs1038152295 Chr.12: 85023122-85023122 on Build GRCh38
rs749803790 Chr.12: 85023087-85023087 on Build GRCh38
rs758634998 Chr.12: 85023335-85023335 on Build GRCh38
rs929308852 Chr.12: 85023095-85023095 on Build GRCh38
rs942403394 Chr.12: 85023526-85023526 on Build GRCh38
rs942126435 Chr.12: 85023140-85023140 on Build GRCh38
rs751138751 Chr.12: 85023372-85023372 on Build GRCh38

View Full Product Details