Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 92420301-92420812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLLU1OS
Entrez Gene ID
574016
Gene Name
chronic lymphocytic leukemia up-regulated 1 opposite strand
Gene Aliases
-
UniGene
Hs.551199
Gene Chromosome Location
Chr.12: 92420086-92428449 on Build GRCh38
Transcripts
NM_001025232.1 XM_017019673.1

SNP Detail

SNP ID Chromosome Location
rs1039943491 Chr.12: 92420413-92420413 on Build GRCh38
rs942854265 Chr.12: 92420664-92420664 on Build GRCh38
rs566604538 Chr.12: 92420398-92420398 on Build GRCh38
rs575218287 Chr.12: 92420491-92420491 on Build GRCh38
rs1051214424 Chr.12: 92420402-92420402 on Build GRCh38
rs145874423 Chr.12: 92420668-92420668 on Build GRCh38
rs908028078 Chr.12: 92420574-92420574 on Build GRCh38
rs958551251 Chr.12: 92420524-92420524 on Build GRCh38
rs963625434 Chr.12: 92420579-92420579 on Build GRCh38
rs941340156 Chr.12: 92420394-92420394 on Build GRCh38
rs991742075 Chr.12: 92420547-92420547 on Build GRCh38
rs893350376 Chr.12: 92420711-92420711 on Build GRCh38
rs780664939 Chr.12: 92420410-92420410 on Build GRCh38
rs1020819218 Chr.12: 92420772-92420772 on Build GRCh38
rs967223829 Chr.12: 92420773-92420773 on Build GRCh38
rs958825093 Chr.12: 92420342-92420342 on Build GRCh38
rs1040296023 Chr.12: 92420687-92420687 on Build GRCh38
rs1047291870 Chr.12: 92420548-92420548 on Build GRCh38
rs780425944 Chr.12: 92420335-92420335 on Build GRCh38
rs1002573362 Chr.12: 92420503-92420503 on Build GRCh38
rs1035445277 Chr.12: 92420523-92420523 on Build GRCh38
rs530661630 Chr.12: 92420474-92420475 on Build GRCh38
rs925900843 Chr.12: 92420444-92420444 on Build GRCh38
rs912666948 Chr.12: 92420656-92420656 on Build GRCh38
rs555030688 Chr.12: 92420439-92420439 on Build GRCh38
rs991090276 Chr.12: 92420490-92420490 on Build GRCh38
rs967653293 Chr.12: 92420434-92420434 on Build GRCh38
rs374847152 Chr.12: 92420326-92420326 on Build GRCh38
rs527239272 Chr.12: 92420628-92420628 on Build GRCh38
rs1037942491 Chr.12: 92420595-92420595 on Build GRCh38
rs986863883 Chr.12: 92420636-92420636 on Build GRCh38
rs769197987 Chr.12: 92420388-92420388 on Build GRCh38
rs1038396561 Chr.12: 92420397-92420397 on Build GRCh38
rs1016156957 Chr.12: 92420573-92420573 on Build GRCh38
rs1011217916 Chr.12: 92420455-92420455 on Build GRCh38
rs138954272 Chr.12: 92420411-92420411 on Build GRCh38
rs74602068 Chr.12: 92420692-92420692 on Build GRCh38
rs534607825 Chr.12: 92420747-92420747 on Build GRCh38
rs577859593 Chr.12: 92420609-92420609 on Build GRCh38
rs890454193 Chr.12: 92420696-92420696 on Build GRCh38
rs930052146 Chr.12: 92420399-92420399 on Build GRCh38
rs1008979588 Chr.12: 92420717-92420717 on Build GRCh38
rs940886909 Chr.12: 92420578-92420578 on Build GRCh38
rs539242825 Chr.12: 92420336-92420336 on Build GRCh38
rs918582155 Chr.12: 92420605-92420605 on Build GRCh38
rs74765089 Chr.12: 92420365-92420365 on Build GRCh38
rs142162684 Chr.12: 92420607-92420607 on Build GRCh38
rs138482153 Chr.12: 92420787-92420787 on Build GRCh38
rs947649443 Chr.12: 92420785-92420785 on Build GRCh38
rs954750035 Chr.12: 92420407-92420407 on Build GRCh38
rs1019912383 Chr.12: 92420482-92420482 on Build GRCh38
rs537869695 Chr.12: 92420596-92420596 on Build GRCh38
rs893392397 Chr.12: 92420437-92420437 on Build GRCh38

View Full Product Details