Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95251844-95252386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VEZT
Entrez Gene ID
55591
Gene Name
vezatin, adherens junctions transmembrane protein
Gene Aliases
VEZATIN
UniGene
Hs.24135
Gene Chromosome Location
Chr.12: 95217746-95302799 on Build GRCh38
Transcripts
NM_017599.3 NR_038241.1 NR_038242.1 XM_006719477.3 XM_006719478.2 XM_006719479.2 XM_006719480.3 XM_006719481.2 XM_006719482.3 XM_006719483.3 XM_006719484.3 XM_006719485.2 XM_006719486.3 XM_006719490.3 XM_011538543.1 XM_011538544.1 XM_011538545.2 XM_011538546.1 XM_011538547.1 XM_011538548.2 XM_011538549.1 XM_011538550.2 XM_011538551.2 XM_011538552.1 XM_011538553.2 XM_017019586.1 XM_017019587.1 XM_017019588.1 XM_017019589.1 XM_017019590.1 XM_017019591.1 XM_017019592.1 XM_017019593.1 XM_017019594.1 XM_017019595.1 XM_017019596.1 XM_017019597.1 XM_017019598.1 XM_017019599.1 XM_017019600.1 XM_017019601.1 XM_017019602.1 XM_017019603.1 XM_017019604.1 XM_017019605.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695354 c.64G>T p.D22Y Chr.12: 95251967-95251967 on Build GRCh38
2046068 c.75C>T p.H25H Chr.12: 95251978-95251978 on Build GRCh38
6346402 c.102G>T p.L34F Chr.12: 95252005-95252005 on Build GRCh38
5473438 c.168+9G>A p.? Chr.12: 95252080-95252080 on Build GRCh38
3813225 c.85G>A p.E29K Chr.12: 95251988-95251988 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945052973 Chr.12: 95252339-95252339 on Build GRCh38
rs971296176 Chr.12: 95252246-95252246 on Build GRCh38
rs755715193 Chr.12: 95252114-95252114 on Build GRCh38
rs35706662 Chr.12: 95252325-95252326 on Build GRCh38
rs898560084 Chr.12: 95251872-95251872 on Build GRCh38
rs912678693 Chr.12: 95252331-95252331 on Build GRCh38
rs750408156 Chr.12: 95252081-95252081 on Build GRCh38
rs1033808514 Chr.12: 95252277-95252277 on Build GRCh38
rs184302617 Chr.12: 95252080-95252080 on Build GRCh38
rs974181907 Chr.12: 95252250-95252250 on Build GRCh38
rs759484148 Chr.12: 95251941-95251941 on Build GRCh38
rs781679052 Chr.12: 95252172-95252172 on Build GRCh38
rs766250527 Chr.12: 95252085-95252085 on Build GRCh38
rs745446290 Chr.12: 95252057-95252057 on Build GRCh38
rs879024738 Chr.12: 95252262-95252262 on Build GRCh38
rs961621337 Chr.12: 95252249-95252249 on Build GRCh38
rs779535400 Chr.12: 95251974-95251974 on Build GRCh38
rs1006599338 Chr.12: 95252136-95252136 on Build GRCh38
rs368375304 Chr.12: 95252147-95252147 on Build GRCh38
rs957824795 Chr.12: 95252283-95252283 on Build GRCh38
rs368650050 Chr.12: 95252300-95252300 on Build GRCh38
rs200805952 Chr.12: 95252079-95252079 on Build GRCh38
rs754731408 Chr.12: 95252086-95252086 on Build GRCh38
rs192728802 Chr.12: 95251931-95251931 on Build GRCh38
rs768263816 Chr.12: 95251995-95251995 on Build GRCh38
rs528291796 Chr.12: 95251884-95251884 on Build GRCh38
rs892360729 Chr.12: 95252347-95252347 on Build GRCh38
rs778578749 Chr.12: 95252055-95252055 on Build GRCh38
rs766443670 Chr.12: 95251924-95251924 on Build GRCh38
rs375841949 Chr.12: 95252035-95252035 on Build GRCh38
rs932875660 Chr.12: 95252295-95252295 on Build GRCh38
rs758083648 Chr.12: 95252082-95252082 on Build GRCh38
rs770363379 Chr.12: 95252001-95252001 on Build GRCh38
rs749848729 Chr.12: 95252116-95252116 on Build GRCh38
rs199826172 Chr.12: 95252119-95252119 on Build GRCh38
rs1025210345 Chr.12: 95252195-95252195 on Build GRCh38
rs569269744 Chr.12: 95251997-95251997 on Build GRCh38
rs778091233 Chr.12: 95252115-95252115 on Build GRCh38
rs550777059 Chr.12: 95251944-95251944 on Build GRCh38
rs148301080 Chr.12: 95252066-95252066 on Build GRCh38
rs376496279 Chr.12: 95251993-95251993 on Build GRCh38
rs991815451 Chr.12: 95252148-95252148 on Build GRCh38
rs750130561 Chr.12: 95251967-95251967 on Build GRCh38
rs757253007 Chr.12: 95251957-95251957 on Build GRCh38
rs768963606 Chr.12: 95252118-95252119 on Build GRCh38
rs751725754 Chr.12: 95251965-95251965 on Build GRCh38
rs767580297 Chr.12: 95252062-95252062 on Build GRCh38
rs2306883 Chr.12: 95251922-95251922 on Build GRCh38
rs189255225 Chr.12: 95252338-95252338 on Build GRCh38
rs763185588 Chr.12: 95252027-95252027 on Build GRCh38
rs546341336 Chr.12: 95251889-95251889 on Build GRCh38
rs960644564 Chr.12: 95252122-95252122 on Build GRCh38
rs771916494 Chr.12: 95252132-95252132 on Build GRCh38
rs554034129 Chr.12: 95251958-95251958 on Build GRCh38
rs773771104 Chr.12: 95252007-95252007 on Build GRCh38
rs201628210 Chr.12: 95252051-95252051 on Build GRCh38
rs981003103 Chr.12: 95252260-95252260 on Build GRCh38
rs781002656 Chr.12: 95252100-95252100 on Build GRCh38
rs908778171 Chr.12: 95252162-95252162 on Build GRCh38
rs972178736 Chr.12: 95251875-95251875 on Build GRCh38
rs374806354 Chr.12: 95252112-95252112 on Build GRCh38
rs747592861 Chr.12: 95252101-95252101 on Build GRCh38
rs774414199 Chr.12: 95252048-95252048 on Build GRCh38
rs369909426 Chr.12: 95251946-95251946 on Build GRCh38
rs753841930 Chr.12: 95251898-95251898 on Build GRCh38
rs962246982 Chr.12: 95252171-95252171 on Build GRCh38
rs746700350 Chr.12: 95251991-95251991 on Build GRCh38
rs777232804 Chr.12: 95251883-95251883 on Build GRCh38
rs545115163 Chr.12: 95252157-95252157 on Build GRCh38
rs778667079 Chr.12: 95251961-95251961 on Build GRCh38
rs944001935 Chr.12: 95252337-95252337 on Build GRCh38
rs373135467 Chr.12: 95251972-95251972 on Build GRCh38
rs1050266480 Chr.12: 95252317-95252317 on Build GRCh38
rs575210110 Chr.12: 95252344-95252344 on Build GRCh38
rs889207085 Chr.12: 95252118-95252118 on Build GRCh38
rs770275255 Chr.12: 95251886-95251886 on Build GRCh38
rs370984462 Chr.12: 95251999-95251999 on Build GRCh38

View Full Product Details