Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 95256343-95256838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VEZT
Entrez Gene ID
55591
Gene Name
vezatin, adherens junctions transmembrane protein
Gene Aliases
VEZATIN
UniGene
Hs.24135
Gene Chromosome Location
Chr.12: 95217746-95302799 on Build GRCh38
Transcripts
NM_017599.3 NR_038241.1 NR_038242.1 XM_006719477.3 XM_006719478.2 XM_006719479.2 XM_006719480.3 XM_006719481.2 XM_006719482.3 XM_006719483.3 XM_006719484.3 XM_006719485.2 XM_006719486.3 XM_006719490.3 XM_011538543.1 XM_011538544.1 XM_011538545.2 XM_011538546.1 XM_011538547.1 XM_011538548.2 XM_011538549.1 XM_011538550.2 XM_011538551.2 XM_011538552.1 XM_011538553.2 XM_017019586.1 XM_017019587.1 XM_017019588.1 XM_017019589.1 XM_017019590.1 XM_017019591.1 XM_017019592.1 XM_017019593.1 XM_017019594.1 XM_017019595.1 XM_017019596.1 XM_017019597.1 XM_017019598.1 XM_017019599.1 XM_017019600.1 XM_017019601.1 XM_017019602.1 XM_017019603.1 XM_017019604.1 XM_017019605.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1744721 c.23A>C p.K8T Chr.12: 95256621-95256621 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs573710455 Chr.12: 95256398-95256398 on Build GRCh38
rs907640682 Chr.12: 95256725-95256725 on Build GRCh38
rs1027427147 Chr.12: 95256553-95256553 on Build GRCh38
rs776698290 Chr.12: 95256594-95256594 on Build GRCh38
rs1018830362 Chr.12: 95256493-95256493 on Build GRCh38
rs1008982062 Chr.12: 95256454-95256454 on Build GRCh38
rs187428041 Chr.12: 95256432-95256432 on Build GRCh38
rs192855802 Chr.12: 95256473-95256473 on Build GRCh38
rs530989817 Chr.12: 95256630-95256630 on Build GRCh38
rs768728822 Chr.12: 95256386-95256386 on Build GRCh38
rs528900381 Chr.12: 95256763-95256763 on Build GRCh38
rs1031536303 Chr.12: 95256385-95256385 on Build GRCh38
rs958048390 Chr.12: 95256389-95256389 on Build GRCh38
rs561739624 Chr.12: 95256716-95256716 on Build GRCh38
rs78637755 Chr.12: 95256795-95256795 on Build GRCh38
rs955729840 Chr.12: 95256427-95256427 on Build GRCh38
rs917482420 Chr.12: 95256636-95256636 on Build GRCh38
rs111603798 Chr.12: 95256790-95256790 on Build GRCh38
rs547378392 Chr.12: 95256773-95256773 on Build GRCh38
rs974525465 Chr.12: 95256518-95256518 on Build GRCh38
rs748406379 Chr.12: 95256648-95256648 on Build GRCh38
rs999933093 Chr.12: 95256402-95256402 on Build GRCh38
rs747160656 Chr.12: 95256378-95256378 on Build GRCh38
rs533306754 Chr.12: 95256814-95256814 on Build GRCh38
rs984164215 Chr.12: 95256557-95256557 on Build GRCh38
rs999606438 Chr.12: 95256375-95256375 on Build GRCh38
rs578165818 Chr.12: 95256524-95256524 on Build GRCh38
rs951912084 Chr.12: 95256556-95256556 on Build GRCh38
rs1047460514 Chr.12: 95256694-95256694 on Build GRCh38
rs543558301 Chr.12: 95256672-95256672 on Build GRCh38
rs781436798 Chr.12: 95256495-95256495 on Build GRCh38
rs182876165 Chr.12: 95256617-95256617 on Build GRCh38
rs939999398 Chr.12: 95256593-95256593 on Build GRCh38
rs545366763 Chr.12: 95256572-95256572 on Build GRCh38
rs760708694 Chr.12: 95256656-95256656 on Build GRCh38
rs752173785 Chr.12: 95256406-95256406 on Build GRCh38
rs772225733 Chr.12: 95256646-95256646 on Build GRCh38
rs979870606 Chr.12: 95256574-95256574 on Build GRCh38

View Full Product Details