Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6571094-6571536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TAS1R1
Entrez Gene ID
80835
Gene Name
taste 1 receptor member 1
Gene Aliases
GM148, GPR70, T1R1, TR1
UniGene
Hs.124574
Gene Chromosome Location
Chr.1: 6555278-6579757 on Build GRCh38
Transcripts
NM_138697.3 NM_177540.2 XM_011542203.2 XM_011542206.2 XM_017002402.1 XM_017002403.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1179460 c.455C>T p.A152V Chr.1: 6571172-6571172 on Build GRCh38
315788 c.387C>A p.L129L Chr.1: 6571104-6571104 on Build GRCh38
5381421 c.396C>T p.D132D Chr.1: 6571113-6571113 on Build GRCh38
1158344 c.416C>T p.T139M Chr.1: 6571133-6571133 on Build GRCh38
3491892 c.412C>T p.P138S Chr.1: 6571129-6571129 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750390456 Chr.1: 6571198-6571198 on Build GRCh38
rs576080914 Chr.1: 6571375-6571375 on Build GRCh38
rs143667096 Chr.1: 6571199-6571199 on Build GRCh38
rs1040270096 Chr.1: 6571463-6571463 on Build GRCh38
rs1032790716 Chr.1: 6571297-6571297 on Build GRCh38
rs928131342 Chr.1: 6571191-6571191 on Build GRCh38
rs34514006 Chr.1: 6571293-6571293 on Build GRCh38
rs777929919 Chr.1: 6571158-6571158 on Build GRCh38
rs372013334 Chr.1: 6571155-6571155 on Build GRCh38
rs141817074 Chr.1: 6571133-6571133 on Build GRCh38
rs969955522 Chr.1: 6571222-6571222 on Build GRCh38
rs900393831 Chr.1: 6571488-6571488 on Build GRCh38
rs763740735 Chr.1: 6571129-6571129 on Build GRCh38
rs746615401 Chr.1: 6571145-6571148 on Build GRCh38
rs200157217 Chr.1: 6571141-6571141 on Build GRCh38
rs1006048263 Chr.1: 6571175-6571175 on Build GRCh38
rs982380380 Chr.1: 6571372-6571372 on Build GRCh38
rs376414342 Chr.1: 6571132-6571132 on Build GRCh38
rs964532315 Chr.1: 6571295-6571295 on Build GRCh38
rs941569853 Chr.1: 6571414-6571414 on Build GRCh38
rs544913726 Chr.1: 6571503-6571503 on Build GRCh38
rs531813245 Chr.1: 6571349-6571349 on Build GRCh38
rs897258178 Chr.1: 6571449-6571449 on Build GRCh38
rs1038833938 Chr.1: 6571415-6571415 on Build GRCh38
rs928164343 Chr.1: 6571396-6571396 on Build GRCh38
rs796989656 Chr.1: 6571299-6571299 on Build GRCh38
rs571661587 Chr.1: 6571428-6571428 on Build GRCh38
rs150596151 Chr.1: 6571185-6571185 on Build GRCh38
rs1008829439 Chr.1: 6571458-6571458 on Build GRCh38
rs747431304 Chr.1: 6571162-6571162 on Build GRCh38
rs761595154 Chr.1: 6571195-6571195 on Build GRCh38
rs907524950 Chr.1: 6571330-6571330 on Build GRCh38
rs369342532 Chr.1: 6571134-6571134 on Build GRCh38
rs776022950 Chr.1: 6571260-6571260 on Build GRCh38
rs751433512 Chr.1: 6571130-6571130 on Build GRCh38
rs1010943486 Chr.1: 6571216-6571216 on Build GRCh38
rs139619592 Chr.1: 6571186-6571186 on Build GRCh38
rs759967739 Chr.1: 6571176-6571176 on Build GRCh38
rs200714568 Chr.1: 6571168-6571168 on Build GRCh38
rs755957533 Chr.1: 6571135-6571135 on Build GRCh38
rs776804403 Chr.1: 6571169-6571169 on Build GRCh38
rs896522499 Chr.1: 6571472-6571472 on Build GRCh38
rs544861976 Chr.1: 6571209-6571209 on Build GRCh38
rs555141750 Chr.1: 6571124-6571124 on Build GRCh38
rs145544066 Chr.1: 6571192-6571192 on Build GRCh38
rs11587438 Chr.1: 6571371-6571371 on Build GRCh38
rs749650803 Chr.1: 6571310-6571310 on Build GRCh38
rs767473177 Chr.1: 6571196-6571196 on Build GRCh38
rs1055696139 Chr.1: 6571404-6571404 on Build GRCh38
rs371088376 Chr.1: 6571263-6571263 on Build GRCh38
rs35788812 Chr.1: 6571232-6571233 on Build GRCh38
rs915369950 Chr.1: 6571405-6571405 on Build GRCh38
rs1014121920 Chr.1: 6571482-6571482 on Build GRCh38
rs567380715 Chr.1: 6571328-6571328 on Build GRCh38
rs549174066 Chr.1: 6571302-6571302 on Build GRCh38
rs770975255 Chr.1: 6571242-6571242 on Build GRCh38
rs922815906 Chr.1: 6571343-6571343 on Build GRCh38
rs11122096 Chr.1: 6571259-6571259 on Build GRCh38
rs747126192 Chr.1: 6571240-6571240 on Build GRCh38
rs140548974 Chr.1: 6571204-6571204 on Build GRCh38
rs1027414856 Chr.1: 6571300-6571300 on Build GRCh38
rs946770653 Chr.1: 6571411-6571411 on Build GRCh38
rs763652546 Chr.1: 6571188-6571188 on Build GRCh38
rs781720509 Chr.1: 6571243-6571243 on Build GRCh38
rs1018787224 Chr.1: 6571291-6571291 on Build GRCh38
rs777646343 Chr.1: 6571211-6571211 on Build GRCh38
rs59679399 Chr.1: 6571298-6571298 on Build GRCh38
rs183933096 Chr.1: 6571507-6571507 on Build GRCh38
rs902533772 Chr.1: 6571435-6571435 on Build GRCh38
rs748409523 Chr.1: 6571148-6571148 on Build GRCh38
rs371838719 Chr.1: 6571207-6571207 on Build GRCh38
rs770349140 Chr.1: 6571184-6571184 on Build GRCh38
rs1008754385 Chr.1: 6571271-6571271 on Build GRCh38
rs746421706 Chr.1: 6571256-6571256 on Build GRCh38
rs1056397282 Chr.1: 6571464-6571464 on Build GRCh38
rs911363357 Chr.1: 6571403-6571403 on Build GRCh38
rs938170063 Chr.1: 6571398-6571398 on Build GRCh38

View Full Product Details