Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9039633-9040144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A5
Entrez Gene ID
6518
Gene Name
solute carrier family 2 member 5
Gene Aliases
GLUT-5, GLUT5
UniGene
Hs.530003
Gene Chromosome Location
Chr.1: 9036941-9093407 on Build GRCh38
Transcripts
NM_003039.2 XM_005263491.3 XM_017002133.1 XM_017002134.1 XM_017002135.1 XM_017002136.1 XM_017002137.1 XM_017002138.1 XM_017002139.1 XM_017002140.1 XM_017002141.1 XM_017002142.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4727072 c.678C>T p.D226D Chr.1: 9040083-9040083 on Build GRCh38
5828027 c.537C>T p.R179R Chr.1: 9039971-9039971 on Build GRCh38
5984500 c.693G>C p.K231N Chr.1: 9040068-9040068 on Build GRCh38
7390412 c.787A>G p.I263V Chr.1: 9039898-9039898 on Build GRCh38
6243989 c.767C>A p.A256E Chr.1: 9039918-9039918 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576402186 Chr.1: 9039671-9039671 on Build GRCh38
rs745684759 Chr.1: 9039801-9039801 on Build GRCh38
rs749076748 Chr.1: 9039878-9039878 on Build GRCh38
rs777064133 Chr.1: 9039839-9039839 on Build GRCh38
rs1006768820 Chr.1: 9040005-9040005 on Build GRCh38
rs752467828 Chr.1: 9040071-9040071 on Build GRCh38
rs763809145 Chr.1: 9039861-9039861 on Build GRCh38
rs751149909 Chr.1: 9039863-9039863 on Build GRCh38
rs760704672 Chr.1: 9039831-9039831 on Build GRCh38
rs753742704 Chr.1: 9040052-9040052 on Build GRCh38
rs776019904 Chr.1: 9040008-9040008 on Build GRCh38
rs767578985 Chr.1: 9039865-9039865 on Build GRCh38
rs200453525 Chr.1: 9040004-9040004 on Build GRCh38
rs59266751 Chr.1: 9039911-9039911 on Build GRCh38
rs760181959 Chr.1: 9040045-9040045 on Build GRCh38
rs764956075 Chr.1: 9039760-9039760 on Build GRCh38
rs758065939 Chr.1: 9039770-9039770 on Build GRCh38
rs781436741 Chr.1: 9040078-9040078 on Build GRCh38
rs548746170 Chr.1: 9040040-9040040 on Build GRCh38
rs574449436 Chr.1: 9039779-9039779 on Build GRCh38
rs766297608 Chr.1: 9039971-9039971 on Build GRCh38
rs74927644 Chr.1: 9039867-9039867 on Build GRCh38
rs762563926 Chr.1: 9039856-9039856 on Build GRCh38
rs906629444 Chr.1: 9040113-9040113 on Build GRCh38
rs757458776 Chr.1: 9039999-9039999 on Build GRCh38
rs770235701 Chr.1: 9040007-9040007 on Build GRCh38
rs774839167 Chr.1: 9040035-9040035 on Build GRCh38
rs896797817 Chr.1: 9039955-9039955 on Build GRCh38
rs753596002 Chr.1: 9039972-9039972 on Build GRCh38
rs772105932 Chr.1: 9039698-9039698 on Build GRCh38
rs773317655 Chr.1: 9039827-9039827 on Build GRCh38
rs756060508 Chr.1: 9039870-9039870 on Build GRCh38
rs202043600 Chr.1: 9040061-9040061 on Build GRCh38
rs932393603 Chr.1: 9039678-9039678 on Build GRCh38
rs372643132 Chr.1: 9039843-9039843 on Build GRCh38
rs761601855 Chr.1: 9040037-9040037 on Build GRCh38
rs768959238 Chr.1: 9039675-9039675 on Build GRCh38
rs995408895 Chr.1: 9039759-9039759 on Build GRCh38
rs765076168 Chr.1: 9040064-9040064 on Build GRCh38
rs752225785 Chr.1: 9039769-9039769 on Build GRCh38
rs757512248 Chr.1: 9040074-9040074 on Build GRCh38
rs769079052 Chr.1: 9040031-9040031 on Build GRCh38
rs547815903 Chr.1: 9039946-9039946 on Build GRCh38
rs750494112 Chr.1: 9040095-9040095 on Build GRCh38
rs754230868 Chr.1: 9039714-9039714 on Build GRCh38
rs200136395 Chr.1: 9039790-9039790 on Build GRCh38
rs771448887 Chr.1: 9039832-9039832 on Build GRCh38
rs139477702 Chr.1: 9039853-9039853 on Build GRCh38
rs767190491 Chr.1: 9039950-9039950 on Build GRCh38
rs148873763 Chr.1: 9039898-9039898 on Build GRCh38
rs779890796 Chr.1: 9039826-9039826 on Build GRCh38
rs754707926 Chr.1: 9039973-9039973 on Build GRCh38
rs766239277 Chr.1: 9040050-9040050 on Build GRCh38
rs12075583 Chr.1: 9039976-9039976 on Build GRCh38
rs759769831 Chr.1: 9039756-9039756 on Build GRCh38
rs747297349 Chr.1: 9039991-9039991 on Build GRCh38
rs756783690 Chr.1: 9039864-9039864 on Build GRCh38
rs769897 Chr.1: 9039668-9039668 on Build GRCh38
rs200930417 Chr.1: 9039907-9039907 on Build GRCh38
rs201546596 Chr.1: 9039892-9039892 on Build GRCh38
rs368017715 Chr.1: 9039687-9039687 on Build GRCh38
rs140582852 Chr.1: 9039960-9039960 on Build GRCh38
rs781298229 Chr.1: 9040000-9040000 on Build GRCh38
rs897095346 Chr.1: 9039713-9039713 on Build GRCh38
rs978115347 Chr.1: 9039844-9039844 on Build GRCh38
rs13306769 Chr.1: 9039696-9039696 on Build GRCh38
rs537378085 Chr.1: 9039666-9039666 on Build GRCh38
rs756163145 Chr.1: 9040119-9040119 on Build GRCh38
rs566162546 Chr.1: 9039968-9039968 on Build GRCh38
rs766840134 Chr.1: 9039705-9039705 on Build GRCh38
rs749630125 Chr.1: 9040026-9040026 on Build GRCh38
rs756038839 Chr.1: 9039866-9039866 on Build GRCh38
rs759948969 Chr.1: 9039841-9039841 on Build GRCh38
rs751093090 Chr.1: 9039788-9039788 on Build GRCh38
rs150154380 Chr.1: 9039893-9039893 on Build GRCh38
rs770313806 Chr.1: 9039906-9039906 on Build GRCh38
rs772625824 Chr.1: 9040041-9040041 on Build GRCh38
rs774094285 Chr.1: 9039918-9039918 on Build GRCh38
rs959196431 Chr.1: 9039761-9039761 on Build GRCh38
rs761217627 Chr.1: 9039704-9039704 on Build GRCh38
rs370295956 Chr.1: 9039914-9039914 on Build GRCh38
rs759464202 Chr.1: 9040059-9040059 on Build GRCh38
rs12564090 Chr.1: 9039847-9039847 on Build GRCh38
rs367787656 Chr.1: 9039791-9039791 on Build GRCh38
rs779760381 Chr.1: 9039877-9039877 on Build GRCh38
rs769421798 Chr.1: 9039813-9039813 on Build GRCh38

View Full Product Details