Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 100402109-100402642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A8
Entrez Gene ID
246213
Gene Name
solute carrier family 17 member 8
Gene Aliases
DFNA25, VGLUT3
UniGene
Hs.116871
Gene Chromosome Location
Chr.12: 100357079-100422059 on Build GRCh38
Transcripts
NM_001145288.1 NM_139319.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6135017 c.801G>T p.L267F Chr.12: 100402377-100402377 on Build GRCh38
6135016 c.945G>T p.P315P Chr.12: 100402637-100402637 on Build GRCh38
241623 c.773G>T p.G258V Chr.12: 100402349-100402349 on Build GRCh38
72663 c.813G>T p.E271D Chr.12: 100402389-100402389 on Build GRCh38
361709 c.881G>T p.G294V Chr.12: 100402457-100402457 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748402878 Chr.12: 100402530-100402530 on Build GRCh38
rs143665730 Chr.12: 100402356-100402356 on Build GRCh38
rs149747614 Chr.12: 100402173-100402173 on Build GRCh38
rs760361819 Chr.12: 100402592-100402592 on Build GRCh38
rs371524742 Chr.12: 100402521-100402521 on Build GRCh38
rs891224281 Chr.12: 100402162-100402162 on Build GRCh38
rs987948345 Chr.12: 100402144-100402144 on Build GRCh38
rs751132044 Chr.12: 100402342-100402342 on Build GRCh38
rs762308166 Chr.12: 100402340-100402340 on Build GRCh38
rs1050204178 Chr.12: 100402554-100402554 on Build GRCh38
rs771575770 Chr.12: 100402580-100402580 on Build GRCh38
rs774862251 Chr.12: 100402576-100402576 on Build GRCh38
rs751707588 Chr.12: 100402503-100402503 on Build GRCh38
rs757238953 Chr.12: 100402493-100402493 on Build GRCh38
rs141300183 Chr.12: 100402606-100402606 on Build GRCh38
rs773521129 Chr.12: 100402447-100402447 on Build GRCh38
rs200016579 Chr.12: 100402479-100402479 on Build GRCh38
rs1036470599 Chr.12: 100402422-100402422 on Build GRCh38
rs763702478 Chr.12: 100402605-100402605 on Build GRCh38
rs749854399 Chr.12: 100402419-100402419 on Build GRCh38
rs765872785 Chr.12: 100402341-100402341 on Build GRCh38
rs747133144 Chr.12: 100402424-100402424 on Build GRCh38
rs774930645 Chr.12: 100402588-100402588 on Build GRCh38
rs764189569 Chr.12: 100402448-100402448 on Build GRCh38
rs202114741 Chr.12: 100402558-100402558 on Build GRCh38
rs145107694 Chr.12: 100402608-100402608 on Build GRCh38
rs780356911 Chr.12: 100402292-100402292 on Build GRCh38
rs781210409 Chr.12: 100402529-100402529 on Build GRCh38
rs531623409 Chr.12: 100402152-100402152 on Build GRCh38
rs967768954 Chr.12: 100402246-100402246 on Build GRCh38
rs727503424 Chr.12: 100402430-100402430 on Build GRCh38
rs759169696 Chr.12: 100402350-100402350 on Build GRCh38
rs775224185 Chr.12: 100402471-100402471 on Build GRCh38
rs770205149 Chr.12: 100402437-100402437 on Build GRCh38
rs565848388 Chr.12: 100402334-100402334 on Build GRCh38
rs756353446 Chr.12: 100402545-100402545 on Build GRCh38
rs925541919 Chr.12: 100402450-100402450 on Build GRCh38
rs965844682 Chr.12: 100402139-100402139 on Build GRCh38
rs773787667 Chr.12: 100402325-100402325 on Build GRCh38
rs780266930 Chr.12: 100402415-100402415 on Build GRCh38
rs753819412 Chr.12: 100402390-100402390 on Build GRCh38
rs764989629 Chr.12: 100402248-100402248 on Build GRCh38
rs765214819 Chr.12: 100402612-100402612 on Build GRCh38
rs145592515 Chr.12: 100402319-100402319 on Build GRCh38
rs770157840 Chr.12: 100402320-100402320 on Build GRCh38
rs752395647 Chr.12: 100402365-100402365 on Build GRCh38
rs753632695 Chr.12: 100402492-100402492 on Build GRCh38
rs951655689 Chr.12: 100402275-100402275 on Build GRCh38
rs747229365 Chr.12: 100402308-100402308 on Build GRCh38
rs1007466960 Chr.12: 100402579-100402579 on Build GRCh38
rs979477866 Chr.12: 100402262-100402262 on Build GRCh38
rs763300852 Chr.12: 100402371-100402371 on Build GRCh38
rs961895662 Chr.12: 100402235-100402235 on Build GRCh38
rs771479939 Chr.12: 100402454-100402454 on Build GRCh38
rs759081653 Chr.12: 100402464-100402464 on Build GRCh38
rs778054930 Chr.12: 100402560-100402560 on Build GRCh38
rs926368196 Chr.12: 100402311-100402311 on Build GRCh38
rs757207684 Chr.12: 100402413-100402413 on Build GRCh38
rs568303762 Chr.12: 100402185-100402185 on Build GRCh38
rs771390050 Chr.12: 100402573-100402573 on Build GRCh38
rs1049798150 Chr.12: 100402153-100402153 on Build GRCh38
rs1019179122 Chr.12: 100402195-100402195 on Build GRCh38
rs773822821 Chr.12: 100402458-100402458 on Build GRCh38
rs748409949 Chr.12: 100402435-100402435 on Build GRCh38
rs113357443 Chr.12: 100402315-100402315 on Build GRCh38
rs576335529 Chr.12: 100402347-100402347 on Build GRCh38
rs368496606 Chr.12: 100402375-100402375 on Build GRCh38
rs751647521 Chr.12: 100402515-100402515 on Build GRCh38
rs537937181 Chr.12: 100402494-100402494 on Build GRCh38
rs148882860 Chr.12: 100402434-100402434 on Build GRCh38
rs745326282 Chr.12: 100402577-100402577 on Build GRCh38
rs763880720 Chr.12: 100402380-100402380 on Build GRCh38
rs760320160 Chr.12: 100402472-100402472 on Build GRCh38
rs7970087 Chr.12: 100402291-100402291 on Build GRCh38

View Full Product Details