Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 107520401-107520801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD11
Entrez Gene ID
121551
Gene Name
BTB domain containing 11
Gene Aliases
ABTB2B
UniGene
Hs.271272
Gene Chromosome Location
Chr.12: 107317168-107659642 on Build GRCh38
Transcripts
NM_001018072.1 XM_005268645.3 XM_011537909.2 XM_011537910.2 XM_017018807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6356457 c.1276G>A p.A426T Chr.12: 107520627-107520627 on Build GRCh38
6613321 c.1183C>T p.R395W Chr.12: 107520534-107520534 on Build GRCh38
4425597 c.1185G>A p.R395R Chr.12: 107520536-107520536 on Build GRCh38
5374053 c.1277C>T p.A426V Chr.12: 107520628-107520628 on Build GRCh38
7251006 c.1223C>T p.S408L Chr.12: 107520574-107520574 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930264439 Chr.12: 107520748-107520748 on Build GRCh38
rs183163162 Chr.12: 107520751-107520751 on Build GRCh38
rs766749863 Chr.12: 107520693-107520693 on Build GRCh38
rs773009357 Chr.12: 107520624-107520624 on Build GRCh38
rs746748967 Chr.12: 107520519-107520519 on Build GRCh38
rs138357783 Chr.12: 107520506-107520506 on Build GRCh38
rs760022263 Chr.12: 107520456-107520456 on Build GRCh38
rs780085559 Chr.12: 107520668-107520668 on Build GRCh38
rs749518256 Chr.12: 107520529-107520529 on Build GRCh38
rs778473618 Chr.12: 107520494-107520494 on Build GRCh38
rs1053131826 Chr.12: 107520428-107520428 on Build GRCh38
rs901332458 Chr.12: 107520744-107520744 on Build GRCh38
rs765978569 Chr.12: 107520630-107520630 on Build GRCh38
rs760959357 Chr.12: 107520629-107520629 on Build GRCh38
rs572418839 Chr.12: 107520540-107520540 on Build GRCh38
rs375912066 Chr.12: 107520488-107520488 on Build GRCh38
rs763598096 Chr.12: 107520483-107520483 on Build GRCh38
rs747538419 Chr.12: 107520611-107520611 on Build GRCh38
rs772146028 Chr.12: 107520601-107520601 on Build GRCh38
rs938558149 Chr.12: 107520427-107520427 on Build GRCh38
rs780079068 Chr.12: 107520573-107520573 on Build GRCh38
rs754934154 Chr.12: 107520575-107520575 on Build GRCh38
rs779042909 Chr.12: 107520644-107520644 on Build GRCh38
rs762880604 Chr.12: 107520440-107520440 on Build GRCh38
rs531702255 Chr.12: 107520635-107520635 on Build GRCh38
rs539681000 Chr.12: 107520712-107520712 on Build GRCh38
rs755923071 Chr.12: 107520643-107520643 on Build GRCh38
rs1026016441 Chr.12: 107520534-107520534 on Build GRCh38
rs545965414 Chr.12: 107520546-107520546 on Build GRCh38
rs774475983 Chr.12: 107520446-107520446 on Build GRCh38
rs781388063 Chr.12: 107520449-107520452 on Build GRCh38
rs778241000 Chr.12: 107520423-107520423 on Build GRCh38
rs947378908 Chr.12: 107520666-107520666 on Build GRCh38
rs561948556 Chr.12: 107520664-107520664 on Build GRCh38
rs756160204 Chr.12: 107520467-107520467 on Build GRCh38
rs979900783 Chr.12: 107520424-107520424 on Build GRCh38
rs748266575 Chr.12: 107520463-107520464 on Build GRCh38
rs376834648 Chr.12: 107520574-107520574 on Build GRCh38
rs765025008 Chr.12: 107520563-107520563 on Build GRCh38
rs200776966 Chr.12: 107520562-107520562 on Build GRCh38
rs191118285 Chr.12: 107520551-107520551 on Build GRCh38
rs748192269 Chr.12: 107520426-107520426 on Build GRCh38
rs1009991245 Chr.12: 107520747-107520747 on Build GRCh38
rs369929793 Chr.12: 107520729-107520729 on Build GRCh38
rs142870733 Chr.12: 107520614-107520614 on Build GRCh38
rs780174842 Chr.12: 107520517-107520517 on Build GRCh38
rs750006935 Chr.12: 107520498-107520498 on Build GRCh38
rs1006001902 Chr.12: 107520558-107520558 on Build GRCh38
rs771043482 Chr.12: 107520444-107520444 on Build GRCh38
rs1054306996 Chr.12: 107520713-107520713 on Build GRCh38
rs373812031 Chr.12: 107520626-107520626 on Build GRCh38
rs1041175301 Chr.12: 107520499-107520499 on Build GRCh38
rs369384423 Chr.12: 107520613-107520613 on Build GRCh38
rs747102172 Chr.12: 107520670-107520670 on Build GRCh38
rs756927512 Chr.12: 107520489-107520489 on Build GRCh38
rs527651988 Chr.12: 107520673-107520673 on Build GRCh38
rs753027981 Chr.12: 107520476-107520476 on Build GRCh38
rs369732393 Chr.12: 107520536-107520536 on Build GRCh38
rs778400600 Chr.12: 107520632-107520632 on Build GRCh38
rs534909008 Chr.12: 107520633-107520633 on Build GRCh38
rs978704781 Chr.12: 107520690-107520690 on Build GRCh38
rs764936824 Chr.12: 107520619-107520619 on Build GRCh38
rs200797164 Chr.12: 107520615-107520615 on Build GRCh38
rs535203696 Chr.12: 107520776-107520776 on Build GRCh38
rs768061028 Chr.12: 107520472-107520472 on Build GRCh38
rs775614897 Chr.12: 107520602-107520602 on Build GRCh38
rs570738828 Chr.12: 107520705-107520705 on Build GRCh38
rs867537282 Chr.12: 107520720-107520720 on Build GRCh38
rs933330309 Chr.12: 107520627-107520627 on Build GRCh38
rs767393830 Chr.12: 107520636-107520636 on Build GRCh38
rs749916863 Chr.12: 107520572-107520572 on Build GRCh38
rs771116406 Chr.12: 107520535-107520535 on Build GRCh38
rs58769417 Chr.12: 107520691-107520691 on Build GRCh38
rs201049274 Chr.12: 107520642-107520642 on Build GRCh38
rs754213567 Chr.12: 107520560-107520560 on Build GRCh38
rs186411315 Chr.12: 107520772-107520772 on Build GRCh38
rs777787569 Chr.12: 107520528-107520528 on Build GRCh38
rs779135626 Chr.12: 107520591-107520591 on Build GRCh38
rs745897907 Chr.12: 107520596-107520596 on Build GRCh38
rs761130494 Chr.12: 107520480-107520480 on Build GRCh38
rs185814432 Chr.12: 107520525-107520525 on Build GRCh38
rs746087194 Chr.12: 107520663-107520663 on Build GRCh38
rs747942150 Chr.12: 107520590-107520590 on Build GRCh38
rs372084873 Chr.12: 107520545-107520545 on Build GRCh38
rs764516288 Chr.12: 107520559-107520559 on Build GRCh38
rs781024108 Chr.12: 107520578-107520578 on Build GRCh38

View Full Product Details