Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 107581005-107581503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD11
Entrez Gene ID
121551
Gene Name
BTB domain containing 11
Gene Aliases
ABTB2B
UniGene
Hs.271272
Gene Chromosome Location
Chr.12: 107317168-107659642 on Build GRCh38
Transcripts
NM_001017523.1 NM_001018072.1 XM_005268645.3 XM_011537909.2 XM_011537910.2 XM_017018807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7291905 c.1556G>A p.R519H Chr.12: 107581217-107581217 on Build GRCh38
4889306 c.1504C>A p.L502M Chr.12: 107581165-107581165 on Build GRCh38
6205353 c.1529G>A p.R510Q Chr.12: 107581190-107581190 on Build GRCh38
6427337 c.101G>A p.R34K Chr.12: 107581007-107581007 on Build GRCh38
1605508 c.1607T>A p.L536Q Chr.12: 107581268-107581268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764863088 Chr.12: 107581105-107581105 on Build GRCh38
rs979072853 Chr.12: 107581343-107581343 on Build GRCh38
rs757948856 Chr.12: 107581101-107581101 on Build GRCh38
rs943288766 Chr.12: 107581027-107581027 on Build GRCh38
rs957981837 Chr.12: 107581106-107581107 on Build GRCh38
rs974692642 Chr.12: 107581366-107581366 on Build GRCh38
rs746467987 Chr.12: 107581129-107581129 on Build GRCh38
rs1016243253 Chr.12: 107581122-107581122 on Build GRCh38
rs927764959 Chr.12: 107581215-107581215 on Build GRCh38
rs561182828 Chr.12: 107581391-107581391 on Build GRCh38
rs1010168587 Chr.12: 107581359-107581359 on Build GRCh38
rs989425927 Chr.12: 107581316-107581316 on Build GRCh38
rs1006168394 Chr.12: 107581109-107581109 on Build GRCh38
rs1020842676 Chr.12: 107581360-107581360 on Build GRCh38
rs762041583 Chr.12: 107581136-107581136 on Build GRCh38
rs553932191 Chr.12: 107581038-107581038 on Build GRCh38
rs997975819 Chr.12: 107581041-107581041 on Build GRCh38
rs889852497 Chr.12: 107581054-107581055 on Build GRCh38
rs943236296 Chr.12: 107581356-107581356 on Build GRCh38
rs982324864 Chr.12: 107581155-107581155 on Build GRCh38
rs966587630 Chr.12: 107581381-107581381 on Build GRCh38
rs1048055482 Chr.12: 107581403-107581403 on Build GRCh38
rs889914170 Chr.12: 107581337-107581337 on Build GRCh38
rs1001755428 Chr.12: 107581293-107581293 on Build GRCh38
rs764491144 Chr.12: 107581136-107581138 on Build GRCh38
rs990986508 Chr.12: 107581433-107581433 on Build GRCh38
rs545104318 Chr.12: 107581029-107581029 on Build GRCh38
rs746285169 Chr.12: 107581056-107581056 on Build GRCh38
rs779707690 Chr.12: 107581123-107581123 on Build GRCh38
rs923058468 Chr.12: 107581384-107581384 on Build GRCh38
rs527397189 Chr.12: 107581055-107581073 on Build GRCh38
rs942139058 Chr.12: 107581412-107581412 on Build GRCh38
rs905881477 Chr.12: 107581455-107581455 on Build GRCh38
rs990542162 Chr.12: 107581292-107581292 on Build GRCh38
rs886561257 Chr.12: 107581081-107581081 on Build GRCh38
rs1037638386 Chr.12: 107581418-107581418 on Build GRCh38
rs933165100 Chr.12: 107581401-107581401 on Build GRCh38
rs1029758166 Chr.12: 107581043-107581043 on Build GRCh38
rs1038860551 Chr.12: 107581036-107581036 on Build GRCh38
rs960241068 Chr.12: 107581312-107581312 on Build GRCh38
rs768161404 Chr.12: 107581169-107581169 on Build GRCh38
rs986322882 Chr.12: 107581079-107581079 on Build GRCh38
rs776193996 Chr.12: 107581225-107581225 on Build GRCh38
rs757841406 Chr.12: 107581028-107581028 on Build GRCh38
rs572237402 Chr.12: 107581236-107581236 on Build GRCh38
rs1036234514 Chr.12: 107581311-107581311 on Build GRCh38
rs886511328 Chr.12: 107581404-107581404 on Build GRCh38
rs897381579 Chr.12: 107581318-107581318 on Build GRCh38
rs542853755 Chr.12: 107581323-107581323 on Build GRCh38
rs953545939 Chr.12: 107581073-107581073 on Build GRCh38
rs1047810307 Chr.12: 107581048-107581048 on Build GRCh38
rs1037290822 Chr.12: 107581313-107581313 on Build GRCh38
rs1001703015 Chr.12: 107581481-107581481 on Build GRCh38
rs776394793 Chr.12: 107581047-107581048 on Build GRCh38
rs924957673 Chr.12: 107581348-107581348 on Build GRCh38
rs937740400 Chr.12: 107581272-107581272 on Build GRCh38
rs966481187 Chr.12: 107581331-107581331 on Build GRCh38
rs1006914096 Chr.12: 107581066-107581066 on Build GRCh38
rs748019591 Chr.12: 107581269-107581269 on Build GRCh38

View Full Product Details