Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109450226-109450734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD10
Entrez Gene ID
83892
Gene Name
potassium channel tetramerization domain containing 10
Gene Aliases
BTBD28, MSTP028, ULRO61, hBACURD3
UniGene
Hs.524731
Gene Chromosome Location
Chr.12: 109448655-109477544 on Build GRCh38
Transcripts
NM_001317395.1 NM_001317399.1 NM_031954.4 NR_133897.1 NR_133898.1

SNP Detail

SNP ID Chromosome Location
rs367751021 Chr.12: 109450698-109450698 on Build GRCh38
rs980805121 Chr.12: 109450332-109450332 on Build GRCh38
rs919963747 Chr.12: 109450350-109450350 on Build GRCh38
rs141898856 Chr.12: 109450461-109450461 on Build GRCh38
rs1053641995 Chr.12: 109450284-109450284 on Build GRCh38
rs182871483 Chr.12: 109450355-109450355 on Build GRCh38
rs1037126801 Chr.12: 109450609-109450609 on Build GRCh38
rs771286268 Chr.12: 109450293-109450293 on Build GRCh38
rs368352301 Chr.12: 109450330-109450330 on Build GRCh38
rs760029667 Chr.12: 109450357-109450357 on Build GRCh38
rs1025129827 Chr.12: 109450380-109450380 on Build GRCh38
rs991023611 Chr.12: 109450372-109450372 on Build GRCh38
rs181754287 Chr.12: 109450551-109450551 on Build GRCh38
rs931892172 Chr.12: 109450481-109450481 on Build GRCh38
rs940491485 Chr.12: 109450592-109450592 on Build GRCh38
rs756840469 Chr.12: 109450314-109450314 on Build GRCh38
rs1025196506 Chr.12: 109450632-109450632 on Build GRCh38
rs138752845 Chr.12: 109450252-109450252 on Build GRCh38
rs73194269 Chr.12: 109450677-109450677 on Build GRCh38
rs936346017 Chr.12: 109450425-109450425 on Build GRCh38
rs562308838 Chr.12: 109450379-109450379 on Build GRCh38
rs572977957 Chr.12: 109450338-109450338 on Build GRCh38
rs892197243 Chr.12: 109450573-109450573 on Build GRCh38
rs908602284 Chr.12: 109450386-109450386 on Build GRCh38
rs959611271 Chr.12: 109450351-109450351 on Build GRCh38
rs527830023 Chr.12: 109450688-109450688 on Build GRCh38
rs772146344 Chr.12: 109450587-109450587 on Build GRCh38
rs1051602730 Chr.12: 109450311-109450311 on Build GRCh38
rs755544851 Chr.12: 109450540-109450540 on Build GRCh38
rs932930517 Chr.12: 109450460-109450460 on Build GRCh38
rs1027243531 Chr.12: 109450347-109450347 on Build GRCh38
rs540824674 Chr.12: 109450340-109450340 on Build GRCh38
rs747040862 Chr.12: 109450659-109450659 on Build GRCh38
rs977885467 Chr.12: 109450393-109450393 on Build GRCh38
rs192251818 Chr.12: 109450353-109450353 on Build GRCh38
rs765384728 Chr.12: 109450403-109450403 on Build GRCh38
rs34650913 Chr.12: 109450355-109450357 on Build GRCh38
rs779363694 Chr.12: 109450550-109450550 on Build GRCh38
rs1030769745 Chr.12: 109450549-109450549 on Build GRCh38
rs796811851 Chr.12: 109450357-109450359 on Build GRCh38
rs3217162 Chr.12: 109450354-109450356 on Build GRCh38
rs868777110 Chr.12: 109450519-109450519 on Build GRCh38
rs1026981613 Chr.12: 109450692-109450692 on Build GRCh38
rs1000656628 Chr.12: 109450270-109450270 on Build GRCh38
rs536262245 Chr.12: 109450283-109450283 on Build GRCh38
rs931818650 Chr.12: 109450308-109450308 on Build GRCh38
rs921891837 Chr.12: 109450479-109450479 on Build GRCh38
rs778097786 Chr.12: 109450621-109450621 on Build GRCh38
rs77531759 Chr.12: 109450667-109450667 on Build GRCh38
rs982856728 Chr.12: 109450381-109450381 on Build GRCh38
rs545796182 Chr.12: 109450313-109450313 on Build GRCh38
rs76929980 Chr.12: 109450251-109450251 on Build GRCh38
rs981138554 Chr.12: 109450672-109450672 on Build GRCh38
rs780693972 Chr.12: 109450331-109450331 on Build GRCh38
rs557527430 Chr.12: 109450322-109450322 on Build GRCh38
rs572892759 Chr.12: 109450580-109450580 on Build GRCh38
rs900089212 Chr.12: 109450418-109450418 on Build GRCh38
rs533141080 Chr.12: 109450626-109450626 on Build GRCh38
rs970909138 Chr.12: 109450389-109450389 on Build GRCh38
rs575418391 Chr.12: 109450710-109450710 on Build GRCh38
rs1052060238 Chr.12: 109450525-109450525 on Build GRCh38
rs995472242 Chr.12: 109450339-109450339 on Build GRCh38
rs1024911254 Chr.12: 109450319-109450319 on Build GRCh38
rs7967399 Chr.12: 109450402-109450402 on Build GRCh38

View Full Product Details