Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 109457850-109458359 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD10
Entrez Gene ID
83892
Gene Name
potassium channel tetramerization domain containing 10
Gene Aliases
BTBD28, MSTP028, ULRO61, hBACURD3
UniGene
Hs.524731
Gene Chromosome Location
Chr.12: 109448655-109477544 on Build GRCh38
Transcripts
NM_001317395.1 NM_001317399.1 NM_031954.4 NR_133897.1 NR_133898.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4038545 c.474+2T>C p.? Chr.12: 109457990-109457990 on Build GRCh38
691320 c.467C>A p.S156* Chr.12: 109457999-109457999 on Build GRCh38
5176067 c.474+10A>G p.? Chr.12: 109457982-109457982 on Build GRCh38
934813 c.406C>T p.P136S Chr.12: 109458060-109458060 on Build GRCh38
275655 c.462G>A p.A154A Chr.12: 109458004-109458004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747613147 Chr.12: 109458068-109458068 on Build GRCh38
rs185743759 Chr.12: 109457889-109457889 on Build GRCh38
rs763027938 Chr.12: 109458093-109458093 on Build GRCh38
rs12227283 Chr.12: 109458199-109458199 on Build GRCh38
rs1005650414 Chr.12: 109458326-109458326 on Build GRCh38
rs555660362 Chr.12: 109458004-109458004 on Build GRCh38
rs969334085 Chr.12: 109458112-109458112 on Build GRCh38
rs775215691 Chr.12: 109457969-109457969 on Build GRCh38
rs776032582 Chr.12: 109457977-109457977 on Build GRCh38
rs914998285 Chr.12: 109458184-109458184 on Build GRCh38
rs556768456 Chr.12: 109458144-109458144 on Build GRCh38
rs944313893 Chr.12: 109458201-109458201 on Build GRCh38
rs140610676 Chr.12: 109458043-109458043 on Build GRCh38
rs769406940 Chr.12: 109458070-109458070 on Build GRCh38
rs775817228 Chr.12: 109457973-109457973 on Build GRCh38
rs888761026 Chr.12: 109457899-109457899 on Build GRCh38
rs34495283 Chr.12: 109458128-109458129 on Build GRCh38
rs765097276 Chr.12: 109457982-109457982 on Build GRCh38
rs759348023 Chr.12: 109458107-109458107 on Build GRCh38
rs58168448 Chr.12: 109458049-109458049 on Build GRCh38
rs538076655 Chr.12: 109458143-109458143 on Build GRCh38
rs988319614 Chr.12: 109458162-109458162 on Build GRCh38
rs866323509 Chr.12: 109457922-109457922 on Build GRCh38
rs1027473726 Chr.12: 109458312-109458312 on Build GRCh38
rs764228628 Chr.12: 109458109-109458109 on Build GRCh38
rs1032003370 Chr.12: 109457887-109457887 on Build GRCh38
rs998309337 Chr.12: 109457914-109457914 on Build GRCh38
rs368114155 Chr.12: 109457981-109457981 on Build GRCh38
rs984332938 Chr.12: 109458188-109458188 on Build GRCh38
rs959254566 Chr.12: 109458161-109458161 on Build GRCh38
rs751372905 Chr.12: 109458008-109458008 on Build GRCh38
rs763115852 Chr.12: 109457932-109457932 on Build GRCh38
rs933943781 Chr.12: 109458284-109458284 on Build GRCh38
rs529038880 Chr.12: 109458296-109458296 on Build GRCh38
rs578058835 Chr.12: 109457895-109457895 on Build GRCh38
rs544087496 Chr.12: 109458310-109458310 on Build GRCh38
rs750305903 Chr.12: 109457998-109457998 on Build GRCh38
rs775908682 Chr.12: 109457878-109457878 on Build GRCh38
rs11609019 Chr.12: 109458153-109458153 on Build GRCh38
rs889844285 Chr.12: 109458319-109458319 on Build GRCh38
rs757297003 Chr.12: 109458034-109458034 on Build GRCh38
rs76932709 Chr.12: 109458200-109458200 on Build GRCh38
rs887585028 Chr.12: 109458318-109458318 on Build GRCh38
rs774311462 Chr.12: 109458096-109458096 on Build GRCh38
rs756436392 Chr.12: 109458010-109458010 on Build GRCh38
rs774498936 Chr.12: 109458137-109458137 on Build GRCh38
rs373694579 Chr.12: 109458064-109458064 on Build GRCh38
rs544990027 Chr.12: 109458179-109458179 on Build GRCh38
rs34225934 Chr.12: 109458235-109458236 on Build GRCh38
rs780436105 Chr.12: 109458060-109458060 on Build GRCh38
rs766383123 Chr.12: 109458094-109458094 on Build GRCh38
rs897736295 Chr.12: 109458273-109458273 on Build GRCh38
rs1026181168 Chr.12: 109457992-109457992 on Build GRCh38
rs985806734 Chr.12: 109458190-109458190 on Build GRCh38
rs766329850 Chr.12: 109458005-109458005 on Build GRCh38
rs12822891 Chr.12: 109458197-109458197 on Build GRCh38
rs746443007 Chr.12: 109458046-109458046 on Build GRCh38
rs772490436 Chr.12: 109457970-109457970 on Build GRCh38
rs753877578 Chr.12: 109458029-109458029 on Build GRCh38
rs778003685 Chr.12: 109458026-109458026 on Build GRCh38
rs1040351978 Chr.12: 109458236-109458236 on Build GRCh38
rs993765966 Chr.12: 109458295-109458295 on Build GRCh38
rs374862342 Chr.12: 109457976-109457976 on Build GRCh38
rs901156803 Chr.12: 109457905-109457905 on Build GRCh38
rs1008831975 Chr.12: 109457909-109457909 on Build GRCh38
rs772577045 Chr.12: 109458088-109458088 on Build GRCh38
rs561249170 Chr.12: 109458229-109458229 on Build GRCh38
rs763620974 Chr.12: 109458118-109458119 on Build GRCh38
rs754157928 Chr.12: 109458115-109458115 on Build GRCh38

View Full Product Details