Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 110651060-110651562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HVCN1
Entrez Gene ID
84329
Gene Name
hydrogen voltage gated channel 1
Gene Aliases
HV1, VSOP
UniGene
Hs.211511
Gene Chromosome Location
Chr.12: 110648686-110704945 on Build GRCh38
Transcripts
NM_001040107.1 NM_001256413.1 NM_032369.3 XM_005253948.2 XM_011538838.1 XM_011538839.2 XM_011538840.2 XM_011538841.2 XM_011538842.2 XM_011538844.2 XM_011538845.1 XM_011538846.2 XM_011538847.1 XM_017020026.1 XM_017020027.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7093086 c.449T>G p.F150C Chr.12: 110651411-110651411 on Build GRCh38
1358737 c.528C>T p.V176V Chr.12: 110651332-110651332 on Build GRCh38
1358736 c.529G>A p.V177M Chr.12: 110651331-110651331 on Build GRCh38
1358735 c.622C>T p.R208W Chr.12: 110651238-110651238 on Build GRCh38
4038675 c.601C>T p.L201L Chr.12: 110651259-110651259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375663985 Chr.12: 110651294-110651294 on Build GRCh38
rs372777561 Chr.12: 110651248-110651248 on Build GRCh38
rs543818690 Chr.12: 110651235-110651235 on Build GRCh38
rs748997855 Chr.12: 110651238-110651238 on Build GRCh38
rs781699201 Chr.12: 110651467-110651468 on Build GRCh38
rs199847095 Chr.12: 110651183-110651183 on Build GRCh38
rs766770167 Chr.12: 110651482-110651482 on Build GRCh38
rs994810445 Chr.12: 110651296-110651296 on Build GRCh38
rs765902906 Chr.12: 110651123-110651123 on Build GRCh38
rs767733937 Chr.12: 110651488-110651488 on Build GRCh38
rs748391921 Chr.12: 110651474-110651475 on Build GRCh38
rs907385822 Chr.12: 110651442-110651442 on Build GRCh38
rs751111228 Chr.12: 110651168-110651168 on Build GRCh38
rs374884864 Chr.12: 110651434-110651434 on Build GRCh38
rs778531975 Chr.12: 110651401-110651401 on Build GRCh38
rs934972701 Chr.12: 110651109-110651109 on Build GRCh38
rs770649056 Chr.12: 110651496-110651497 on Build GRCh38
rs768457352 Chr.12: 110651435-110651435 on Build GRCh38
rs773827729 Chr.12: 110651497-110651498 on Build GRCh38
rs369638051 Chr.12: 110651325-110651325 on Build GRCh38
rs371702882 Chr.12: 110651341-110651341 on Build GRCh38
rs112719731 Chr.12: 110651501-110651501 on Build GRCh38
rs376096473 Chr.12: 110651483-110651483 on Build GRCh38
rs900179793 Chr.12: 110651234-110651234 on Build GRCh38
rs759014369 Chr.12: 110651268-110651268 on Build GRCh38
rs146855907 Chr.12: 110651375-110651375 on Build GRCh38
rs572342262 Chr.12: 110651184-110651184 on Build GRCh38
rs933712812 Chr.12: 110651237-110651237 on Build GRCh38
rs1034325074 Chr.12: 110651105-110651105 on Build GRCh38
rs564954972 Chr.12: 110651503-110651503 on Build GRCh38
rs200277616 Chr.12: 110651180-110651180 on Build GRCh38
rs923609649 Chr.12: 110651095-110651095 on Build GRCh38
rs781764314 Chr.12: 110651210-110651210 on Build GRCh38
rs944615289 Chr.12: 110651202-110651202 on Build GRCh38
rs777929241 Chr.12: 110651219-110651219 on Build GRCh38
rs985863350 Chr.12: 110651530-110651530 on Build GRCh38
rs879171740 Chr.12: 110651139-110651139 on Build GRCh38
rs772064093 Chr.12: 110651285-110651285 on Build GRCh38
rs764475857 Chr.12: 110651499-110651499 on Build GRCh38
rs763287076 Chr.12: 110651305-110651305 on Build GRCh38
rs771575756 Chr.12: 110651416-110651416 on Build GRCh38
rs1001630313 Chr.12: 110651096-110651096 on Build GRCh38
rs971434071 Chr.12: 110651084-110651084 on Build GRCh38
rs770439659 Chr.12: 110651247-110651247 on Build GRCh38
rs781358524 Chr.12: 110651331-110651331 on Build GRCh38
rs374262901 Chr.12: 110651212-110651212 on Build GRCh38
rs537178438 Chr.12: 110651088-110651088 on Build GRCh38
rs544927951 Chr.12: 110651513-110651514 on Build GRCh38
rs1006259215 Chr.12: 110651523-110651523 on Build GRCh38
rs754543385 Chr.12: 110651307-110651307 on Build GRCh38
rs760539481 Chr.12: 110651298-110651298 on Build GRCh38
rs150325252 Chr.12: 110651146-110651146 on Build GRCh38
rs984429482 Chr.12: 110651147-110651147 on Build GRCh38
rs369188786 Chr.12: 110651457-110651457 on Build GRCh38
rs759761891 Chr.12: 110651487-110651487 on Build GRCh38
rs201614335 Chr.12: 110651365-110651365 on Build GRCh38
rs373217560 Chr.12: 110651332-110651332 on Build GRCh38
rs749592355 Chr.12: 110651344-110651344 on Build GRCh38
rs145106959 Chr.12: 110651335-110651335 on Build GRCh38
rs752772990 Chr.12: 110651492-110651492 on Build GRCh38
rs756690985 Chr.12: 110651494-110651494 on Build GRCh38
rs763981983 Chr.12: 110651299-110651299 on Build GRCh38
rs149175563 Chr.12: 110651334-110651334 on Build GRCh38
rs749906449 Chr.12: 110651376-110651376 on Build GRCh38
rs755585465 Chr.12: 110651188-110651188 on Build GRCh38
rs577208007 Chr.12: 110651162-110651162 on Build GRCh38
rs745440611 Chr.12: 110651410-110651410 on Build GRCh38
rs186695691 Chr.12: 110651301-110651301 on Build GRCh38
rs368753321 Chr.12: 110651213-110651213 on Build GRCh38
rs372611439 Chr.12: 110651471-110651471 on Build GRCh38
rs751759696 Chr.12: 110651323-110651323 on Build GRCh38
rs554039155 Chr.12: 110651422-110651422 on Build GRCh38
rs531985803 Chr.12: 110651532-110651532 on Build GRCh38
rs759135542 Chr.12: 110651499-110651500 on Build GRCh38

View Full Product Details