Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112125302-112125809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRAFD1
Entrez Gene ID
10906
Gene Name
TRAF-type zinc finger domain containing 1
Gene Aliases
FLN29
UniGene
Hs.5148
Gene Chromosome Location
Chr.12: 112125545-112153604 on Build GRCh38
Transcripts
NM_001143906.1 NM_006700.2

SNP Detail

SNP ID Chromosome Location
rs1010263292 Chr.12: 112125471-112125471 on Build GRCh38
rs906231088 Chr.12: 112125575-112125575 on Build GRCh38
rs868624081 Chr.12: 112125754-112125754 on Build GRCh38
rs985412728 Chr.12: 112125466-112125466 on Build GRCh38
rs1018198406 Chr.12: 112125785-112125785 on Build GRCh38
rs956752014 Chr.12: 112125382-112125382 on Build GRCh38
rs930154519 Chr.12: 112125525-112125525 on Build GRCh38
rs752555789 Chr.12: 112125392-112125392 on Build GRCh38
rs911245303 Chr.12: 112125482-112125482 on Build GRCh38
rs748430394 Chr.12: 112125720-112125720 on Build GRCh38
rs946283827 Chr.12: 112125713-112125713 on Build GRCh38
rs546136211 Chr.12: 112125570-112125570 on Build GRCh38
rs989371469 Chr.12: 112125396-112125396 on Build GRCh38
rs748160256 Chr.12: 112125590-112125590 on Build GRCh38
rs936193563 Chr.12: 112125651-112125651 on Build GRCh38
rs565620168 Chr.12: 112125567-112125567 on Build GRCh38
rs984598394 Chr.12: 112125533-112125533 on Build GRCh38
rs1011169105 Chr.12: 112125715-112125715 on Build GRCh38
rs570890948 Chr.12: 112125759-112125759 on Build GRCh38
rs550494771 Chr.12: 112125552-112125552 on Build GRCh38
rs542622048 Chr.12: 112125465-112125465 on Build GRCh38
rs1023336195 Chr.12: 112125339-112125339 on Build GRCh38
rs547284486 Chr.12: 112125676-112125676 on Build GRCh38
rs561320571 Chr.12: 112125496-112125496 on Build GRCh38
rs1000288358 Chr.12: 112125353-112125353 on Build GRCh38
rs1054728670 Chr.12: 112125440-112125440 on Build GRCh38
rs889952916 Chr.12: 112125348-112125348 on Build GRCh38
rs1034395543 Chr.12: 112125502-112125502 on Build GRCh38
rs766525708 Chr.12: 112125358-112125358 on Build GRCh38
rs1022113061 Chr.12: 112125753-112125753 on Build GRCh38
rs1011934829 Chr.12: 112125336-112125336 on Build GRCh38
rs915160926 Chr.12: 112125413-112125413 on Build GRCh38
rs531640025 Chr.12: 112125544-112125544 on Build GRCh38
rs560337802 Chr.12: 112125444-112125444 on Build GRCh38
rs944118868 Chr.12: 112125495-112125495 on Build GRCh38
rs951871922 Chr.12: 112125515-112125515 on Build GRCh38
rs925007016 Chr.12: 112125634-112125634 on Build GRCh38
rs778480719 Chr.12: 112125762-112125762 on Build GRCh38
rs1031168450 Chr.12: 112125629-112125629 on Build GRCh38
rs1001506238 Chr.12: 112125786-112125786 on Build GRCh38
rs888092254 Chr.12: 112125335-112125335 on Build GRCh38
rs965762963 Chr.12: 112125787-112125787 on Build GRCh38
rs918724110 Chr.12: 112125498-112125498 on Build GRCh38
rs758492502 Chr.12: 112125461-112125461 on Build GRCh38
rs986045457 Chr.12: 112125775-112125775 on Build GRCh38
rs931609082 Chr.12: 112125620-112125620 on Build GRCh38
rs1030824508 Chr.12: 112125359-112125359 on Build GRCh38
rs892273561 Chr.12: 112125650-112125650 on Build GRCh38
rs781747255 Chr.12: 112125611-112125611 on Build GRCh38
rs1003259670 Chr.12: 112125624-112125624 on Build GRCh38
rs548030476 Chr.12: 112125698-112125698 on Build GRCh38
rs948309450 Chr.12: 112125565-112125565 on Build GRCh38
rs970448413 Chr.12: 112125356-112125356 on Build GRCh38
rs888690685 Chr.12: 112125560-112125560 on Build GRCh38
rs1001618797 Chr.12: 112125497-112125497 on Build GRCh38
rs907205992 Chr.12: 112125764-112125764 on Build GRCh38
rs1048613818 Chr.12: 112125528-112125528 on Build GRCh38
rs751755433 Chr.12: 112125529-112125529 on Build GRCh38
rs1014867108 Chr.12: 112125557-112125557 on Build GRCh38
rs372761342 Chr.12: 112125447-112125447 on Build GRCh38
rs1055016693 Chr.12: 112125660-112125660 on Build GRCh38

View Full Product Details