Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112906847-112907352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OAS1
Entrez Gene ID
4938
Gene Name
2'-5'-oligoadenylate synthetase 1
Gene Aliases
E18/E16, IFI-4, OIAS, OIASI
UniGene
Hs.524760
Gene Chromosome Location
Chr.12: 112906777-112932190 on Build GRCh38
Transcripts
NM_001032409.2 NM_001320151.1 NM_002534.3 NM_016816.3 XM_006719434.1 XM_011538413.1 XM_017019361.1 XM_017019362.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3739440 c.43T>A p.F15I Chr.12: 112907082-112907082 on Build GRCh38
5958591 c.20delC p.P8fs*34 Chr.12: 112907059-112907059 on Build GRCh38
6732059 c.67G>C p.D23H Chr.12: 112907106-112907106 on Build GRCh38
389081 c.149C>G p.S50C Chr.12: 112907188-112907188 on Build GRCh38
3456297 c.152A>G p.Y51C Chr.12: 112907191-112907191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769441652 Chr.12: 112907034-112907034 on Build GRCh38
rs534305575 Chr.12: 112907088-112907088 on Build GRCh38
rs772789659 Chr.12: 112907037-112907037 on Build GRCh38
rs568931178 Chr.12: 112907142-112907142 on Build GRCh38
rs761844551 Chr.12: 112907064-112907064 on Build GRCh38
rs747004173 Chr.12: 112907026-112907026 on Build GRCh38
rs746226406 Chr.12: 112907040-112907040 on Build GRCh38
rs868732648 Chr.12: 112906950-112906950 on Build GRCh38
rs371488150 Chr.12: 112907135-112907135 on Build GRCh38
rs971215607 Chr.12: 112907120-112907120 on Build GRCh38
rs200259745 Chr.12: 112907178-112907178 on Build GRCh38
rs117835961 Chr.12: 112907118-112907118 on Build GRCh38
rs753984423 Chr.12: 112907283-112907284 on Build GRCh38
rs563149920 Chr.12: 112907268-112907268 on Build GRCh38
rs770540128 Chr.12: 112907155-112907155 on Build GRCh38
rs767460536 Chr.12: 112907192-112907192 on Build GRCh38
rs1032920285 Chr.12: 112907074-112907074 on Build GRCh38
rs940998521 Chr.12: 112906959-112906959 on Build GRCh38
rs912047764 Chr.12: 112907210-112907210 on Build GRCh38
rs773489189 Chr.12: 112907239-112907240 on Build GRCh38
rs944868103 Chr.12: 112907245-112907245 on Build GRCh38
rs985482632 Chr.12: 112907276-112907276 on Build GRCh38
rs772480478 Chr.12: 112907044-112907044 on Build GRCh38
rs777080728 Chr.12: 112907209-112907209 on Build GRCh38
rs1009068826 Chr.12: 112906881-112906881 on Build GRCh38
rs777467037 Chr.12: 112907119-112907119 on Build GRCh38
rs919444856 Chr.12: 112907185-112907185 on Build GRCh38
rs765469926 Chr.12: 112907072-112907072 on Build GRCh38
rs377053595 Chr.12: 112907115-112907115 on Build GRCh38
rs1050994 Chr.12: 112907130-112907130 on Build GRCh38
rs61096170 Chr.12: 112907285-112907285 on Build GRCh38
rs138921278 Chr.12: 112907202-112907202 on Build GRCh38
rs370284276 Chr.12: 112907104-112907104 on Build GRCh38
rs369215137 Chr.12: 112907048-112907048 on Build GRCh38
rs769182127 Chr.12: 112907141-112907141 on Build GRCh38
rs770574261 Chr.12: 112907122-112907122 on Build GRCh38
rs756850392 Chr.12: 112907241-112907241 on Build GRCh38
rs1036030154 Chr.12: 112906900-112906900 on Build GRCh38
rs761951929 Chr.12: 112907160-112907160 on Build GRCh38
rs763028185 Chr.12: 112907269-112907270 on Build GRCh38
rs999206387 Chr.12: 112907054-112907054 on Build GRCh38
rs758312258 Chr.12: 112907011-112907011 on Build GRCh38
rs781015631 Chr.12: 112907110-112907110 on Build GRCh38
rs747643556 Chr.12: 112907033-112907033 on Build GRCh38
rs775788574 Chr.12: 112907133-112907133 on Build GRCh38
rs1036081422 Chr.12: 112907246-112907246 on Build GRCh38
rs576801406 Chr.12: 112907045-112907045 on Build GRCh38
rs754926467 Chr.12: 112907109-112907109 on Build GRCh38
rs1023012042 Chr.12: 112907140-112907140 on Build GRCh38
rs1049302028 Chr.12: 112906892-112906892 on Build GRCh38
rs761571646 Chr.12: 112907025-112907028 on Build GRCh38
rs764613269 Chr.12: 112907057-112907057 on Build GRCh38
rs894425439 Chr.12: 112907315-112907315 on Build GRCh38
rs780688765 Chr.12: 112907029-112907029 on Build GRCh38
rs201403228 Chr.12: 112907198-112907198 on Build GRCh38
rs758259109 Chr.12: 112907093-112907093 on Build GRCh38
rs541677821 Chr.12: 112907214-112907214 on Build GRCh38
rs780140160 Chr.12: 112907013-112907013 on Build GRCh38
rs373546690 Chr.12: 112907092-112907092 on Build GRCh38
rs770069482 Chr.12: 112907195-112907198 on Build GRCh38
rs941008612 Chr.12: 112907289-112907289 on Build GRCh38
rs754872011 Chr.12: 112907182-112907182 on Build GRCh38
rs1039513103 Chr.12: 112907002-112907002 on Build GRCh38
rs777041841 Chr.12: 112907151-112907151 on Build GRCh38
rs377027609 Chr.12: 112907097-112907097 on Build GRCh38
rs751405629 Chr.12: 112907105-112907105 on Build GRCh38
rs754395067 Chr.12: 112907285-112907286 on Build GRCh38
rs778581219 Chr.12: 112907250-112907250 on Build GRCh38
rs373022452 Chr.12: 112907107-112907107 on Build GRCh38
rs147205471 Chr.12: 112907053-112907053 on Build GRCh38
rs1048709759 Chr.12: 112907306-112907306 on Build GRCh38
rs771615858 Chr.12: 112907324-112907324 on Build GRCh38
rs773358405 Chr.12: 112907170-112907170 on Build GRCh38
rs1013253247 Chr.12: 112907318-112907318 on Build GRCh38
rs763205404 Chr.12: 112907173-112907173 on Build GRCh38
rs150398756 Chr.12: 112907284-112907285 on Build GRCh38
rs994484366 Chr.12: 112907292-112907292 on Build GRCh38
rs776721646 Chr.12: 112907061-112907061 on Build GRCh38
rs909580725 Chr.12: 112906898-112906898 on Build GRCh38
rs757458413 Chr.12: 112906991-112906991 on Build GRCh38
rs748897961 Chr.12: 112906973-112906973 on Build GRCh38
rs751350524 Chr.12: 112907179-112907179 on Build GRCh38
rs779021235 Chr.12: 112907000-112907000 on Build GRCh38
rs750207645 Chr.12: 112907008-112907008 on Build GRCh38
rs897512272 Chr.12: 112906949-112906949 on Build GRCh38
rs781716825 Chr.12: 112907270-112907270 on Build GRCh38
rs374296254 Chr.12: 112907163-112907163 on Build GRCh38
rs558343010 Chr.12: 112907027-112907027 on Build GRCh38

View Full Product Details