Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112919730-112920240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OAS1
Entrez Gene ID
4938
Gene Name
2'-5'-oligoadenylate synthetase 1
Gene Aliases
E18/E16, IFI-4, OIAS, OIASI
UniGene
Hs.524760
Gene Chromosome Location
Chr.12: 112906777-112932190 on Build GRCh38
Transcripts
NM_001032409.2 NM_001320151.1 NM_016816.3 XM_006719434.1 XM_011538413.1 XM_017019361.1 XM_017019362.1

SNP Detail

SNP ID Chromosome Location
rs370200655 Chr.12: 112920058-112920058 on Build GRCh38
rs934091121 Chr.12: 112919842-112919842 on Build GRCh38
rs931929023 Chr.12: 112919802-112919802 on Build GRCh38
rs112149027 Chr.12: 112919903-112919903 on Build GRCh38
rs74616969 Chr.12: 112920090-112920090 on Build GRCh38
rs150256018 Chr.12: 112920136-112920136 on Build GRCh38
rs914937375 Chr.12: 112920106-112920106 on Build GRCh38
rs893407433 Chr.12: 112919895-112919895 on Build GRCh38
rs754098397 Chr.12: 112919870-112919870 on Build GRCh38
rs35264948 Chr.12: 112919880-112919880 on Build GRCh38
rs1012531370 Chr.12: 112919978-112919978 on Build GRCh38
rs34893035 Chr.12: 112919847-112919848 on Build GRCh38
rs112543402 Chr.12: 112919858-112919858 on Build GRCh38
rs921301284 Chr.12: 112920091-112920091 on Build GRCh38
rs1050758943 Chr.12: 112919824-112919824 on Build GRCh38
rs141050313 Chr.12: 112920076-112920076 on Build GRCh38
rs548240820 Chr.12: 112920155-112920155 on Build GRCh38
rs765377753 Chr.12: 112920203-112920203 on Build GRCh38
rs559857402 Chr.12: 112920099-112920099 on Build GRCh38
rs113325329 Chr.12: 112919883-112919883 on Build GRCh38
rs765559840 Chr.12: 112919912-112919912 on Build GRCh38
rs983038971 Chr.12: 112920053-112920053 on Build GRCh38
rs752863626 Chr.12: 112919968-112919968 on Build GRCh38
rs138943145 Chr.12: 112920138-112920138 on Build GRCh38
rs903383458 Chr.12: 112919862-112919862 on Build GRCh38
rs1023308408 Chr.12: 112919910-112919910 on Build GRCh38
rs944534397 Chr.12: 112919860-112919860 on Build GRCh38
rs970962215 Chr.12: 112920047-112920047 on Build GRCh38
rs138376387 Chr.12: 112919760-112919760 on Build GRCh38
rs934735878 Chr.12: 112920187-112920187 on Build GRCh38
rs570073768 Chr.12: 112919809-112919809 on Build GRCh38
rs986104722 Chr.12: 112920140-112920140 on Build GRCh38
rs79677566 Chr.12: 112919929-112919929 on Build GRCh38
rs989107904 Chr.12: 112919756-112919756 on Build GRCh38
rs899807602 Chr.12: 112919792-112919792 on Build GRCh38
rs956472047 Chr.12: 112920089-112920089 on Build GRCh38
rs34733355 Chr.12: 112920136-112920137 on Build GRCh38
rs888418111 Chr.12: 112919966-112919966 on Build GRCh38
rs758586420 Chr.12: 112920059-112920059 on Build GRCh38
rs1024291781 Chr.12: 112920014-112920014 on Build GRCh38
rs924710085 Chr.12: 112920184-112920184 on Build GRCh38
rs911790297 Chr.12: 112920171-112920171 on Build GRCh38

View Full Product Details