Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113208029-113208465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IQCD
Entrez Gene ID
115811
Gene Name
IQ motif containing D
Gene Aliases
4933433C09Rik, CFAP84, DRC10
UniGene
Hs.568276
Gene Chromosome Location
Chr.12: 113195441-113221344 on Build GRCh38
Transcripts
NM_138451.2 XM_005253832.4 XM_005253833.4 XM_005253834.4 XM_011537862.2 XM_011537863.2 XM_011537864.1 XM_011537865.2 XM_011537866.2 XM_017018771.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7188364 c.125C>T p.T42I Chr.12: 113208042-113208042 on Build GRCh38
7188363 c.125C>T p.T42I Chr.12: 113208042-113208042 on Build GRCh38
6686814 c.19G>A p.A7T Chr.12: 113208148-113208148 on Build GRCh38
5920431 c.86G>A p.R29K Chr.12: 113208081-113208081 on Build GRCh38
3998676 c.56G>A p.R19K Chr.12: 113208111-113208111 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970119714 Chr.12: 113208263-113208263 on Build GRCh38
rs548244523 Chr.12: 113208285-113208285 on Build GRCh38
rs185835807 Chr.12: 113208236-113208236 on Build GRCh38
rs749562171 Chr.12: 113208202-113208202 on Build GRCh38
rs976388545 Chr.12: 113208293-113208293 on Build GRCh38
rs778244633 Chr.12: 113208200-113208200 on Build GRCh38
rs757314322 Chr.12: 113208141-113208141 on Build GRCh38
rs1051732773 Chr.12: 113208433-113208433 on Build GRCh38
rs984273886 Chr.12: 113208336-113208336 on Build GRCh38
rs879753527 Chr.12: 113208384-113208384 on Build GRCh38
rs763586464 Chr.12: 113208076-113208076 on Build GRCh38
rs1025299284 Chr.12: 113208309-113208309 on Build GRCh38
rs779590248 Chr.12: 113208174-113208174 on Build GRCh38
rs372134716 Chr.12: 113208172-113208172 on Build GRCh38
rs534839458 Chr.12: 113208145-113208145 on Build GRCh38
rs201536583 Chr.12: 113208217-113208217 on Build GRCh38
rs983342812 Chr.12: 113208370-113208370 on Build GRCh38
rs774390657 Chr.12: 113208235-113208235 on Build GRCh38
rs141052395 Chr.12: 113208132-113208132 on Build GRCh38
rs764014584 Chr.12: 113208118-113208118 on Build GRCh38
rs150266692 Chr.12: 113208149-113208149 on Build GRCh38
rs951184145 Chr.12: 113208313-113208313 on Build GRCh38
rs776244170 Chr.12: 113208178-113208178 on Build GRCh38
rs550062589 Chr.12: 113208418-113208418 on Build GRCh38
rs909415334 Chr.12: 113208371-113208371 on Build GRCh38
rs372423641 Chr.12: 113208259-113208259 on Build GRCh38
rs768306958 Chr.12: 113208138-113208138 on Build GRCh38
rs376437746 Chr.12: 113208196-113208196 on Build GRCh38
rs569683487 Chr.12: 113208399-113208399 on Build GRCh38
rs749984572 Chr.12: 113208148-113208148 on Build GRCh38
rs922076923 Chr.12: 113208424-113208424 on Build GRCh38
rs1035766032 Chr.12: 113208311-113208311 on Build GRCh38
rs1052571737 Chr.12: 113208214-113208214 on Build GRCh38
rs927526433 Chr.12: 113208413-113208413 on Build GRCh38
rs60284450 Chr.12: 113208111-113208111 on Build GRCh38
rs34541861 Chr.12: 113208151-113208152 on Build GRCh38
rs3741990 Chr.12: 113208239-113208239 on Build GRCh38
rs140811110 Chr.12: 113208410-113208410 on Build GRCh38
rs193036533 Chr.12: 113208084-113208084 on Build GRCh38
rs748469466 Chr.12: 113208052-113208052 on Build GRCh38
rs34820322 Chr.12: 113208053-113208053 on Build GRCh38
rs904347714 Chr.12: 113208216-113208216 on Build GRCh38
rs760795023 Chr.12: 113208112-113208112 on Build GRCh38
rs928603328 Chr.12: 113208056-113208056 on Build GRCh38
rs748785231 Chr.12: 113208198-113208198 on Build GRCh38
rs907719787 Chr.12: 113208092-113208092 on Build GRCh38
rs771402177 Chr.12: 113208083-113208083 on Build GRCh38
rs145458075 Chr.12: 113208097-113208097 on Build GRCh38
rs942158135 Chr.12: 113208380-113208380 on Build GRCh38
rs767727440 Chr.12: 113208108-113208108 on Build GRCh38
rs931975290 Chr.12: 113208429-113208429 on Build GRCh38
rs746735317 Chr.12: 113208188-113208188 on Build GRCh38
rs3741991 Chr.12: 113208231-113208231 on Build GRCh38
rs759568288 Chr.12: 113208085-113208085 on Build GRCh38
rs565843767 Chr.12: 113208060-113208060 on Build GRCh38
rs963204145 Chr.12: 113208409-113208409 on Build GRCh38

View Full Product Details