Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 117212812-117213317 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOS1
Entrez Gene ID
4842
Gene Name
nitric oxide synthase 1
Gene Aliases
IHPS1, N-NOS, NC-NOS, NOS, bNOS, nNOS
UniGene
Hs.654410
Gene Chromosome Location
Chr.12: 117208142-117441843 on Build GRCh38
Transcripts
NM_000620.4 NM_001204213.1 NM_001204214.1 NM_001204218.1 XM_011538398.2 XM_017019345.1 XM_017019346.1 XM_017019347.1

SNP Detail

SNP ID Chromosome Location
rs925734980 Chr.12: 117212920-117212920 on Build GRCh38
rs9658573 Chr.12: 117212934-117212934 on Build GRCh38
rs904022247 Chr.12: 117213231-117213231 on Build GRCh38
rs1018456378 Chr.12: 117212844-117212844 on Build GRCh38
rs917123035 Chr.12: 117212941-117212941 on Build GRCh38
rs927137432 Chr.12: 117213010-117213010 on Build GRCh38
rs575915753 Chr.12: 117212967-117212969 on Build GRCh38
rs147417836 Chr.12: 117213247-117213247 on Build GRCh38
rs9658570 Chr.12: 117213068-117213068 on Build GRCh38
rs897261365 Chr.12: 117213067-117213067 on Build GRCh38
rs757277479 Chr.12: 117213039-117213039 on Build GRCh38
rs9658571 Chr.12: 117212977-117212977 on Build GRCh38
rs201438460 Chr.12: 117212974-117212976 on Build GRCh38
rs1000515400 Chr.12: 117212976-117212979 on Build GRCh38
rs1007157193 Chr.12: 117213157-117213157 on Build GRCh38
rs1042103693 Chr.12: 117212967-117212967 on Build GRCh38
rs182678396 Chr.12: 117212999-117212999 on Build GRCh38
rs901639108 Chr.12: 117212931-117212931 on Build GRCh38
rs973237605 Chr.12: 117213055-117213055 on Build GRCh38
rs9658572 Chr.12: 117212973-117212975 on Build GRCh38
rs577833773 Chr.12: 117213034-117213034 on Build GRCh38
rs9658574 Chr.12: 117212919-117212919 on Build GRCh38
rs964424344 Chr.12: 117212898-117212898 on Build GRCh38
rs762068243 Chr.12: 117213115-117213115 on Build GRCh38
rs1051122275 Chr.12: 117213074-117213074 on Build GRCh38
rs952845649 Chr.12: 117213114-117213114 on Build GRCh38
rs923102978 Chr.12: 117213246-117213246 on Build GRCh38
rs531445428 Chr.12: 117213251-117213251 on Build GRCh38
rs988522299 Chr.12: 117212940-117212940 on Build GRCh38
rs950013179 Chr.12: 117212966-117212966 on Build GRCh38
rs878860833 Chr.12: 117212921-117212921 on Build GRCh38
rs765406580 Chr.12: 117213043-117213043 on Build GRCh38
rs910450133 Chr.12: 117212845-117212846 on Build GRCh38
rs1013797997 Chr.12: 117212985-117212987 on Build GRCh38
rs529129655 Chr.12: 117213227-117213227 on Build GRCh38
rs941232371 Chr.12: 117213014-117213014 on Build GRCh38
rs891333389 Chr.12: 117213094-117213094 on Build GRCh38
rs9658569 Chr.12: 117213201-117213201 on Build GRCh38
rs143103197 Chr.12: 117213237-117213237 on Build GRCh38
rs536319412 Chr.12: 117213286-117213286 on Build GRCh38
rs895389468 Chr.12: 117212982-117212982 on Build GRCh38

View Full Product Details