Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20567781-20568288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFT88
Entrez Gene ID
8100
Gene Name
intraflagellar transport 88
Gene Aliases
D13S1056E, DAF19, TG737, TTC10, hTg737
UniGene
Hs.187376
Gene Chromosome Location
Chr.13: 20566446-20691437 on Build GRCh38
Transcripts
NM_001318491.1 NM_001318493.1 NM_006531.4 NM_175605.4 NR_134653.1 XM_005266553.2 XM_006719870.3 XM_011535241.2 XM_011535242.1 XM_011535243.1 XM_017020757.1 XM_017020758.1 XM_017020759.1 XM_017020760.1 XM_017020761.1 XM_017020762.1 XM_017020763.1 XM_017020764.1 XM_017020765.1 XM_017020766.1 XM_017020767.1 XM_017020768.1 XM_017020769.1 XM_017020770.1 XM_017020771.1 XM_017020772.1 XM_017020773.1 XM_017020774.1 XM_017020775.1 XM_017020776.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6237013 c.3G>A p.M1I Chr.13: 20567979-20567979 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768503252 Chr.13: 20567833-20567833 on Build GRCh38
rs191691526 Chr.13: 20568091-20568091 on Build GRCh38
rs767872180 Chr.13: 20567958-20567958 on Build GRCh38
rs1051624272 Chr.13: 20568074-20568074 on Build GRCh38
rs980334428 Chr.13: 20567824-20567824 on Build GRCh38
rs866742105 Chr.13: 20567872-20567872 on Build GRCh38
rs926475845 Chr.13: 20568145-20568145 on Build GRCh38
rs371948561 Chr.13: 20568244-20568244 on Build GRCh38
rs1033024540 Chr.13: 20568076-20568076 on Build GRCh38
rs577871937 Chr.13: 20568061-20568061 on Build GRCh38
rs190265640 Chr.13: 20567811-20567811 on Build GRCh38
rs368836743 Chr.13: 20568007-20568007 on Build GRCh38
rs1047407724 Chr.13: 20568191-20568191 on Build GRCh38
rs1026528127 Chr.13: 20567855-20567856 on Build GRCh38
rs925946546 Chr.13: 20567873-20567873 on Build GRCh38
rs776378449 Chr.13: 20567971-20567971 on Build GRCh38
rs913844427 Chr.13: 20568105-20568105 on Build GRCh38
rs1001953284 Chr.13: 20567983-20567983 on Build GRCh38
rs964804836 Chr.13: 20567869-20567869 on Build GRCh38
rs1005212284 Chr.13: 20568088-20568088 on Build GRCh38
rs949263634 Chr.13: 20567919-20567919 on Build GRCh38
rs770292076 Chr.13: 20568092-20568092 on Build GRCh38
rs1043547456 Chr.13: 20568029-20568029 on Build GRCh38
rs549097182 Chr.13: 20568230-20568230 on Build GRCh38
rs118009094 Chr.13: 20568229-20568229 on Build GRCh38
rs796785396 Chr.13: 20568180-20568180 on Build GRCh38
rs993128700 Chr.13: 20568003-20568003 on Build GRCh38
rs954866198 Chr.13: 20567856-20567859 on Build GRCh38
rs937802611 Chr.13: 20568149-20568149 on Build GRCh38
rs775749823 Chr.13: 20568157-20568157 on Build GRCh38
rs1037463161 Chr.13: 20568175-20568175 on Build GRCh38
rs1020421958 Chr.13: 20568213-20568213 on Build GRCh38
rs773485113 Chr.13: 20567952-20567954 on Build GRCh38
rs904971315 Chr.13: 20567945-20567945 on Build GRCh38
rs752208512 Chr.13: 20567998-20567998 on Build GRCh38
rs34335945 Chr.13: 20568179-20568180 on Build GRCh38
rs994574507 Chr.13: 20568100-20568100 on Build GRCh38
rs969064001 Chr.13: 20567817-20567817 on Build GRCh38
rs560746430 Chr.13: 20568238-20568238 on Build GRCh38
rs1003242308 Chr.13: 20568140-20568140 on Build GRCh38
rs1052313579 Chr.13: 20567892-20567892 on Build GRCh38
rs890412268 Chr.13: 20568082-20568082 on Build GRCh38
rs890685179 Chr.13: 20568009-20568009 on Build GRCh38
rs559121832 Chr.13: 20567868-20567869 on Build GRCh38
rs542590925 Chr.13: 20568237-20568237 on Build GRCh38
rs566416105 Chr.13: 20567861-20567864 on Build GRCh38
rs377387784 Chr.13: 20568002-20568002 on Build GRCh38
rs531225441 Chr.13: 20568246-20568246 on Build GRCh38
rs374667274 Chr.13: 20567992-20567992 on Build GRCh38
rs187137033 Chr.13: 20568049-20568049 on Build GRCh38
rs904844982 Chr.13: 20568112-20568112 on Build GRCh38
rs367929083 Chr.13: 20568180-20568180 on Build GRCh38
rs898885716 Chr.13: 20568180-20568184 on Build GRCh38
rs890729544 Chr.13: 20568214-20568214 on Build GRCh38
rs1046210371 Chr.13: 20567932-20567932 on Build GRCh38
rs982389626 Chr.13: 20568132-20568132 on Build GRCh38
rs373501203 Chr.13: 20568023-20568023 on Build GRCh38
rs35134506 Chr.13: 20568254-20568255 on Build GRCh38
rs1035939111 Chr.13: 20568160-20568160 on Build GRCh38
rs545341444 Chr.13: 20568104-20568104 on Build GRCh38
rs989474953 Chr.13: 20568187-20568187 on Build GRCh38
rs145788041 Chr.13: 20567864-20567864 on Build GRCh38
rs139295343 Chr.13: 20567910-20567910 on Build GRCh38
rs934625161 Chr.13: 20567880-20567880 on Build GRCh38
rs762084057 Chr.13: 20567866-20567868 on Build GRCh38
rs759519179 Chr.13: 20567979-20567979 on Build GRCh38
rs141865659 Chr.13: 20568107-20568107 on Build GRCh38
rs36104881 Chr.13: 20567936-20567937 on Build GRCh38
rs928898272 Chr.13: 20567863-20567863 on Build GRCh38
rs987627519 Chr.13: 20567860-20567860 on Build GRCh38
rs1000293389 Chr.13: 20568242-20568243 on Build GRCh38
rs987395011 Chr.13: 20568258-20568258 on Build GRCh38
rs963658609 Chr.13: 20567832-20567832 on Build GRCh38
rs756831560 Chr.13: 20568036-20568036 on Build GRCh38
rs182811302 Chr.13: 20568043-20568043 on Build GRCh38
rs751294205 Chr.13: 20568017-20568017 on Build GRCh38
rs141923780 Chr.13: 20567823-20567823 on Build GRCh38
rs931982200 Chr.13: 20568257-20568257 on Build GRCh38
rs949319191 Chr.13: 20568117-20568117 on Build GRCh38

View Full Product Details