Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 23759966-23760366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MIPEP
Entrez Gene ID
4285
Gene Name
mitochondrial intermediate peptidase
Gene Aliases
HMIP, MIP
UniGene
Hs.507498
Gene Chromosome Location
Chr.13: 23730189-23889448 on Build GRCh38
Transcripts
NM_005932.3 XM_011535097.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5882394 c.1889C>G p.A630G Chr.13: 23760177-23760177 on Build GRCh38
4887514 c.1924G>A p.A642T Chr.13: 23760142-23760142 on Build GRCh38
5479303 c.1919C>T p.A640V Chr.13: 23760147-23760147 on Build GRCh38
40329 c.1853G>A p.W618* Chr.13: 23760213-23760213 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778437046 Chr.13: 23760160-23760160 on Build GRCh38
rs555387574 Chr.13: 23760050-23760050 on Build GRCh38
rs771444720 Chr.13: 23760254-23760254 on Build GRCh38
rs746620137 Chr.13: 23760253-23760253 on Build GRCh38
rs906151681 Chr.13: 23760054-23760054 on Build GRCh38
rs539269421 Chr.13: 23759993-23759993 on Build GRCh38
rs752794654 Chr.13: 23760204-23760204 on Build GRCh38
rs969010822 Chr.13: 23760277-23760277 on Build GRCh38
rs936984105 Chr.13: 23760009-23760009 on Build GRCh38
rs1015130959 Chr.13: 23760196-23760196 on Build GRCh38
rs529288577 Chr.13: 23760265-23760265 on Build GRCh38
rs376771690 Chr.13: 23760228-23760228 on Build GRCh38
rs777034997 Chr.13: 23760255-23760255 on Build GRCh38
rs973177911 Chr.13: 23760017-23760017 on Build GRCh38
rs557408244 Chr.13: 23760001-23760001 on Build GRCh38
rs763154846 Chr.13: 23760205-23760205 on Build GRCh38
rs916882917 Chr.13: 23760252-23760252 on Build GRCh38
rs765285559 Chr.13: 23760190-23760190 on Build GRCh38
rs761135725 Chr.13: 23760180-23760180 on Build GRCh38
rs754340900 Chr.13: 23760109-23760109 on Build GRCh38
rs781008265 Chr.13: 23760133-23760133 on Build GRCh38
rs764894647 Chr.13: 23760068-23760068 on Build GRCh38
rs141693205 Chr.13: 23760142-23760142 on Build GRCh38
rs763148609 Chr.13: 23760023-23760023 on Build GRCh38
rs77219429 Chr.13: 23760170-23760170 on Build GRCh38
rs927646178 Chr.13: 23760295-23760295 on Build GRCh38
rs188050867 Chr.13: 23760288-23760288 on Build GRCh38
rs755280433 Chr.13: 23760122-23760122 on Build GRCh38
rs546454361 Chr.13: 23760146-23760146 on Build GRCh38
rs539230791 Chr.13: 23760326-23760326 on Build GRCh38
rs757925741 Chr.13: 23760247-23760247 on Build GRCh38
rs758629053 Chr.13: 23760125-23760125 on Build GRCh38
rs764290465 Chr.13: 23760206-23760206 on Build GRCh38
rs765320829 Chr.13: 23760120-23760120 on Build GRCh38
rs550431622 Chr.13: 23760281-23760281 on Build GRCh38
rs748779733 Chr.13: 23760061-23760061 on Build GRCh38
rs751661538 Chr.13: 23760112-23760112 on Build GRCh38
rs546719717 Chr.13: 23760147-23760147 on Build GRCh38
rs777574052 Chr.13: 23760248-23760248 on Build GRCh38
rs538389927 Chr.13: 23760098-23760098 on Build GRCh38
rs572925117 Chr.13: 23760018-23760018 on Build GRCh38
rs369304651 Chr.13: 23760091-23760091 on Build GRCh38
rs767624628 Chr.13: 23760329-23760329 on Build GRCh38
rs779781347 Chr.13: 23760238-23760238 on Build GRCh38
rs879030374 Chr.13: 23760032-23760032 on Build GRCh38
rs531105854 Chr.13: 23760145-23760145 on Build GRCh38
rs191532110 Chr.13: 23760055-23760055 on Build GRCh38
rs909665310 Chr.13: 23760290-23760290 on Build GRCh38
rs539834367 Chr.13: 23760041-23760041 on Build GRCh38
rs750217160 Chr.13: 23760138-23760138 on Build GRCh38
rs772345005 Chr.13: 23760090-23760090 on Build GRCh38
rs114529191 Chr.13: 23760240-23760240 on Build GRCh38
rs183315697 Chr.13: 23760186-23760186 on Build GRCh38
rs568404185 Chr.13: 23760241-23760241 on Build GRCh38
rs373511397 Chr.13: 23760157-23760157 on Build GRCh38
rs754920730 Chr.13: 23760243-23760243 on Build GRCh38
rs755904677 Chr.13: 23760229-23760229 on Build GRCh38
rs778324238 Chr.13: 23760244-23760244 on Build GRCh38
rs779977642 Chr.13: 23760143-23760143 on Build GRCh38
rs556005069 Chr.13: 23760245-23760245 on Build GRCh38
rs761303186 Chr.13: 23760099-23760099 on Build GRCh38
rs773876682 Chr.13: 23760071-23760071 on Build GRCh38
rs561723529 Chr.13: 23760308-23760308 on Build GRCh38
rs564084853 Chr.13: 23760037-23760037 on Build GRCh38
rs746406909 Chr.13: 23760264-23760264 on Build GRCh38
rs768150177 Chr.13: 23760062-23760062 on Build GRCh38
rs1046801523 Chr.13: 23760043-23760043 on Build GRCh38
rs765234272 Chr.13: 23760185-23760185 on Build GRCh38
rs773688171 Chr.13: 23760094-23760094 on Build GRCh38
rs11620036 Chr.13: 23760076-23760076 on Build GRCh38

View Full Product Details