Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24426158-24426665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP4
Entrez Gene ID
143
Gene Name
poly(ADP-ribose) polymerase family member 4
Gene Aliases
ADPRTL1, ARTD4, PARP-4, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193
UniGene
Hs.744855
Gene Chromosome Location
Chr.13: 24420930-24512993 on Build GRCh38
Transcripts
NM_006437.3 XM_011534931.1 XM_011534932.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
147645 c.4966G>C p.A1656P Chr.13: 24426479-24426479 on Build GRCh38
7135673 c.4930G>A p.G1644R Chr.13: 24426515-24426515 on Build GRCh38
4395387 c.4979G>A p.R1660K Chr.13: 24426466-24426466 on Build GRCh38
3667268 c.4923A>G p.E1641E Chr.13: 24426522-24426522 on Build GRCh38
4711223 c.4867C>T p.L1623F Chr.13: 24426578-24426578 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975688730 Chr.13: 24426189-24426189 on Build GRCh38
rs7571 Chr.13: 24426479-24426479 on Build GRCh38
rs755406445 Chr.13: 24426439-24426439 on Build GRCh38
rs4986821 Chr.13: 24426365-24426370 on Build GRCh38
rs748162886 Chr.13: 24426452-24426452 on Build GRCh38
rs369643554 Chr.13: 24426492-24426492 on Build GRCh38
rs148836083 Chr.13: 24426364-24426365 on Build GRCh38
rs773569597 Chr.13: 24426469-24426469 on Build GRCh38
rs4769352 Chr.13: 24426237-24426237 on Build GRCh38
rs771248465 Chr.13: 24426617-24426617 on Build GRCh38
rs1045640646 Chr.13: 24426248-24426248 on Build GRCh38
rs111651712 Chr.13: 24426363-24426364 on Build GRCh38
rs955697100 Chr.13: 24426562-24426562 on Build GRCh38
rs779283324 Chr.13: 24426582-24426582 on Build GRCh38
rs765468333 Chr.13: 24426607-24426607 on Build GRCh38
rs79836043 Chr.13: 24426544-24426544 on Build GRCh38
rs766986577 Chr.13: 24426535-24426535 on Build GRCh38
rs761848426 Chr.13: 24426444-24426445 on Build GRCh38
rs113208590 Chr.13: 24426570-24426570 on Build GRCh38
rs779097364 Chr.13: 24426451-24426451 on Build GRCh38
rs749023476 Chr.13: 24426629-24426629 on Build GRCh38
rs747263929 Chr.13: 24426612-24426612 on Build GRCh38
rs112883311 Chr.13: 24426222-24426222 on Build GRCh38
rs763919540 Chr.13: 24426493-24426493 on Build GRCh38
rs879439879 Chr.13: 24426188-24426188 on Build GRCh38
rs377673804 Chr.13: 24426603-24426603 on Build GRCh38
rs376375021 Chr.13: 24426527-24426527 on Build GRCh38
rs186652264 Chr.13: 24426607-24426607 on Build GRCh38
rs775387437 Chr.13: 24426480-24426480 on Build GRCh38
rs773973507 Chr.13: 24426504-24426504 on Build GRCh38
rs151093035 Chr.13: 24426594-24426594 on Build GRCh38
rs558791071 Chr.13: 24426260-24426260 on Build GRCh38
rs146698321 Chr.13: 24426365-24426366 on Build GRCh38
rs544037664 Chr.13: 24426331-24426331 on Build GRCh38
rs200620045 Chr.13: 24426362-24426363 on Build GRCh38
rs561462882 Chr.13: 24426316-24426316 on Build GRCh38
rs924295308 Chr.13: 24426203-24426203 on Build GRCh38
rs768930425 Chr.13: 24426220-24426220 on Build GRCh38
rs887898917 Chr.13: 24426412-24426415 on Build GRCh38
rs965939217 Chr.13: 24426184-24426184 on Build GRCh38
rs148540630 Chr.13: 24426536-24426536 on Build GRCh38
rs955768032 Chr.13: 24426221-24426221 on Build GRCh38
rs897812949 Chr.13: 24426357-24426357 on Build GRCh38
rs572128192 Chr.13: 24426234-24426234 on Build GRCh38
rs201200397 Chr.13: 24426618-24426618 on Build GRCh38
rs772400621 Chr.13: 24426463-24426463 on Build GRCh38
rs766768710 Chr.13: 24426557-24426557 on Build GRCh38
rs4769353 Chr.13: 24426318-24426318 on Build GRCh38
rs780340102 Chr.13: 24426420-24426420 on Build GRCh38
rs761225027 Chr.13: 24426534-24426534 on Build GRCh38
rs1056454117 Chr.13: 24426338-24426338 on Build GRCh38
rs754177942 Chr.13: 24426565-24426565 on Build GRCh38
rs993527187 Chr.13: 24426372-24426372 on Build GRCh38
rs1049138262 Chr.13: 24426404-24426404 on Build GRCh38
rs768710170 Chr.13: 24426639-24426639 on Build GRCh38
rs773728309 Chr.13: 24426488-24426488 on Build GRCh38
rs781543498 Chr.13: 24426435-24426435 on Build GRCh38
rs540236101 Chr.13: 24426511-24426511 on Build GRCh38

View Full Product Details