Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24431392-24431900 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP4
Entrez Gene ID
143
Gene Name
poly(ADP-ribose) polymerase family member 4
Gene Aliases
ADPRTL1, ARTD4, PARP-4, PARPL, PH5P, VAULT3, VPARP, VWA5C, p193
UniGene
Hs.744855
Gene Chromosome Location
Chr.13: 24420930-24512993 on Build GRCh38
Transcripts
NM_006437.3 XM_011534931.1 XM_011534932.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3467810 c.4793T>G p.L1598R Chr.13: 24431430-24431430 on Build GRCh38
946140 c.4748A>G p.D1583G Chr.13: 24431475-24431475 on Build GRCh38
1219314 c.4755C>G p.F1585L Chr.13: 24431468-24431468 on Build GRCh38
946139 c.4781T>G p.L1594R Chr.13: 24431442-24431442 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs770357568 Chr.13: 24431486-24431486 on Build GRCh38
rs767603597 Chr.13: 24431793-24431793 on Build GRCh38
rs7996670 Chr.13: 24431582-24431582 on Build GRCh38
rs772764244 Chr.13: 24431476-24431476 on Build GRCh38
rs957854935 Chr.13: 24431738-24431738 on Build GRCh38
rs764685686 Chr.13: 24431516-24431516 on Build GRCh38
rs751757493 Chr.13: 24431518-24431518 on Build GRCh38
rs758671857 Chr.13: 24431503-24431507 on Build GRCh38
rs768352917 Chr.13: 24431442-24431442 on Build GRCh38
rs969308484 Chr.13: 24431828-24431828 on Build GRCh38
rs1970483 Chr.13: 24431766-24431766 on Build GRCh38
rs906247978 Chr.13: 24431443-24431443 on Build GRCh38
rs759178436 Chr.13: 24431506-24431506 on Build GRCh38
rs753072530 Chr.13: 24431510-24431510 on Build GRCh38
rs1027151734 Chr.13: 24431566-24431566 on Build GRCh38
rs981751154 Chr.13: 24431831-24431831 on Build GRCh38
rs61947015 Chr.13: 24431636-24431636 on Build GRCh38
rs533661418 Chr.13: 24431797-24431797 on Build GRCh38
rs141166620 Chr.13: 24431853-24431853 on Build GRCh38
rs568855663 Chr.13: 24431656-24431656 on Build GRCh38
rs978187811 Chr.13: 24431688-24431688 on Build GRCh38
rs965782690 Chr.13: 24431666-24431666 on Build GRCh38
rs750635244 Chr.13: 24431525-24431525 on Build GRCh38
rs781560716 Chr.13: 24431422-24431422 on Build GRCh38
rs940472484 Chr.13: 24431483-24431483 on Build GRCh38
rs199950278 Chr.13: 24431524-24431524 on Build GRCh38
rs547524825 Chr.13: 24431804-24431804 on Build GRCh38
rs763421232 Chr.13: 24431514-24431514 on Build GRCh38
rs765550178 Chr.13: 24431508-24431508 on Build GRCh38
rs567368520 Chr.13: 24431841-24431841 on Build GRCh38
rs776161455 Chr.13: 24431503-24431503 on Build GRCh38
rs550512806 Chr.13: 24431638-24431638 on Build GRCh38
rs940381552 Chr.13: 24431849-24431849 on Build GRCh38
rs117315221 Chr.13: 24431740-24431740 on Build GRCh38
rs995668889 Chr.13: 24431546-24431546 on Build GRCh38
rs746039639 Chr.13: 24431439-24431439 on Build GRCh38
rs1006815768 Chr.13: 24431617-24431617 on Build GRCh38
rs1031538181 Chr.13: 24431693-24431693 on Build GRCh38
rs760169506 Chr.13: 24431485-24431485 on Build GRCh38
rs536510336 Chr.13: 24431844-24431844 on Build GRCh38
rs535500218 Chr.13: 24431440-24431440 on Build GRCh38
rs748070578 Chr.13: 24431468-24431468 on Build GRCh38
rs370844309 Chr.13: 24431522-24431522 on Build GRCh38
rs778723248 Chr.13: 24431462-24431462 on Build GRCh38
rs4770677 Chr.13: 24431772-24431772 on Build GRCh38
rs927635793 Chr.13: 24431832-24431832 on Build GRCh38

View Full Product Details