Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 12192352-12192857 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF1B
Entrez Gene ID
7133
Gene Name
TNF receptor superfamily member 1B
Gene Aliases
CD120b, TBPII, TNF-R-II, TNF-R75, TNFBR, TNFR1B, TNFR2, TNFR80, p75, p75TNFR
UniGene
Hs.256278
Gene Chromosome Location
Chr.1: 12166948-12209222 on Build GRCh38
Transcripts
NM_001066.2 XM_011542060.2 XM_011542063.2 XM_017002211.1 XM_017002212.1 XM_017002213.1 XM_017002214.1 XM_017002215.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
894798 c.475G>A p.V159M Chr.1: 12192448-12192448 on Build GRCh38
4734894 c.528G>A p.T176T Chr.1: 12192501-12192501 on Build GRCh38
6805057 c.503C>T p.T168M Chr.1: 12192476-12192476 on Build GRCh38
4550357 c.499G>A p.G167R Chr.1: 12192472-12192472 on Build GRCh38
6805054 c.482G>A p.C161Y Chr.1: 12192455-12192455 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs768823607 Chr.1: 12192379-12192379 on Build GRCh38
rs886628953 Chr.1: 12192809-12192809 on Build GRCh38
rs767782313 Chr.1: 12192461-12192461 on Build GRCh38
rs1025555995 Chr.1: 12192411-12192411 on Build GRCh38
rs1006857088 Chr.1: 12192762-12192762 on Build GRCh38
rs746890304 Chr.1: 12192514-12192514 on Build GRCh38
rs17886002 Chr.1: 12192415-12192415 on Build GRCh38
rs528310825 Chr.1: 12192767-12192768 on Build GRCh38
rs969055764 Chr.1: 12192500-12192500 on Build GRCh38
rs565635562 Chr.1: 12192627-12192627 on Build GRCh38
rs769329010 Chr.1: 12192524-12192524 on Build GRCh38
rs571129550 Chr.1: 12192488-12192488 on Build GRCh38
rs956227501 Chr.1: 12192656-12192656 on Build GRCh38
rs756427248 Chr.1: 12192470-12192470 on Build GRCh38
rs755130591 Chr.1: 12192477-12192477 on Build GRCh38
rs1010277883 Chr.1: 12192705-12192705 on Build GRCh38
rs754426669 Chr.1: 12192821-12192821 on Build GRCh38
rs554329036 Chr.1: 12192822-12192822 on Build GRCh38
rs923246282 Chr.1: 12192807-12192807 on Build GRCh38
rs776601667 Chr.1: 12192518-12192518 on Build GRCh38
rs559822776 Chr.1: 12192789-12192789 on Build GRCh38
rs375517338 Chr.1: 12192565-12192565 on Build GRCh38
rs760544693 Chr.1: 12192401-12192401 on Build GRCh38
rs1043290864 Chr.1: 12192732-12192732 on Build GRCh38
rs765420868 Chr.1: 12192823-12192823 on Build GRCh38
rs201574750 Chr.1: 12192476-12192476 on Build GRCh38
rs947601462 Chr.1: 12192720-12192720 on Build GRCh38
rs763871288 Chr.1: 12192560-12192560 on Build GRCh38
rs759635294 Chr.1: 12192793-12192793 on Build GRCh38
rs377401769 Chr.1: 12192501-12192501 on Build GRCh38
rs910402749 Chr.1: 12192764-12192764 on Build GRCh38
rs750854322 Chr.1: 12192462-12192462 on Build GRCh38
rs770961458 Chr.1: 12192413-12192413 on Build GRCh38
rs1035662943 Chr.1: 12192598-12192598 on Build GRCh38
rs751834930 Chr.1: 12192612-12192612 on Build GRCh38
rs1041528790 Chr.1: 12192769-12192769 on Build GRCh38
rs759423347 Chr.1: 12192522-12192522 on Build GRCh38
rs764708786 Chr.1: 12192421-12192421 on Build GRCh38
rs547920119 Chr.1: 12192750-12192750 on Build GRCh38
rs75924049 Chr.1: 12192393-12192393 on Build GRCh38
rs534441104 Chr.1: 12192728-12192728 on Build GRCh38
rs976411216 Chr.1: 12192794-12192794 on Build GRCh38
rs779326428 Chr.1: 12192386-12192386 on Build GRCh38
rs1053274623 Chr.1: 12192604-12192604 on Build GRCh38
rs866539950 Chr.1: 12192517-12192517 on Build GRCh38
rs1768642 Chr.1: 12192835-12192835 on Build GRCh38
rs752373710 Chr.1: 12192453-12192453 on Build GRCh38
rs749240503 Chr.1: 12192389-12192390 on Build GRCh38
rs780648249 Chr.1: 12192474-12192474 on Build GRCh38
rs762353199 Chr.1: 12192544-12192544 on Build GRCh38
rs552190870 Chr.1: 12192595-12192595 on Build GRCh38
rs916212512 Chr.1: 12192418-12192418 on Build GRCh38
rs113772467 Chr.1: 12192428-12192428 on Build GRCh38
rs889799410 Chr.1: 12192760-12192760 on Build GRCh38
rs779095658 Chr.1: 12192484-12192484 on Build GRCh38
rs754846502 Chr.1: 12192552-12192553 on Build GRCh38
rs372217736 Chr.1: 12192414-12192414 on Build GRCh38
rs766601906 Chr.1: 12192817-12192817 on Build GRCh38
rs1047451026 Chr.1: 12192831-12192831 on Build GRCh38
rs774919239 Chr.1: 12192539-12192539 on Build GRCh38
rs964618029 Chr.1: 12192792-12192792 on Build GRCh38
rs761148768 Chr.1: 12192568-12192568 on Build GRCh38
rs2275415 Chr.1: 12192516-12192516 on Build GRCh38
rs373216188 Chr.1: 12192460-12192460 on Build GRCh38
rs199963289 Chr.1: 12192408-12192408 on Build GRCh38
rs141628211 Chr.1: 12192489-12192489 on Build GRCh38
rs369774125 Chr.1: 12192556-12192556 on Build GRCh38
rs200865554 Chr.1: 12192387-12192387 on Build GRCh38
rs772993824 Chr.1: 12192399-12192399 on Build GRCh38

View Full Product Details