Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 13513142-13513642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LRRC38
Entrez Gene ID
126755
Gene Name
leucine rich repeat containing 38
Gene Aliases
-
UniGene
Hs.657356
Gene Chromosome Location
Chr.1: 13474977-13513747 on Build GRCh38
Transcripts
NM_001010847.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2229991 c.281C>T p.S94L Chr.1: 13513313-13513313 on Build GRCh38
6527507 c.143C>G p.P48R Chr.1: 13513451-13513451 on Build GRCh38
2229992 c.280T>A p.S94T Chr.1: 13513314-13513314 on Build GRCh38
6540101 c.81G>A p.A27A Chr.1: 13513513-13513513 on Build GRCh38
3376678 c.308C>T p.S103L Chr.1: 13513286-13513286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759480827 Chr.1: 13513279-13513279 on Build GRCh38
rs1003451543 Chr.1: 13513365-13513365 on Build GRCh38
rs758192768 Chr.1: 13513357-13513357 on Build GRCh38
rs953439910 Chr.1: 13513564-13513564 on Build GRCh38
rs867656672 Chr.1: 13513236-13513236 on Build GRCh38
rs930283316 Chr.1: 13513373-13513373 on Build GRCh38
rs771988041 Chr.1: 13513381-13513381 on Build GRCh38
rs1019517470 Chr.1: 13513590-13513590 on Build GRCh38
rs1053016315 Chr.1: 13513438-13513438 on Build GRCh38
rs765264170 Chr.1: 13513305-13513305 on Build GRCh38
rs554683538 Chr.1: 13513228-13513228 on Build GRCh38
rs772369396 Chr.1: 13513254-13513254 on Build GRCh38
rs763142199 Chr.1: 13513391-13513391 on Build GRCh38
rs766436549 Chr.1: 13513525-13513525 on Build GRCh38
rs553608233 Chr.1: 13513168-13513168 on Build GRCh38
rs893070170 Chr.1: 13513514-13513514 on Build GRCh38
rs897523846 Chr.1: 13513167-13513167 on Build GRCh38
rs528326776 Chr.1: 13513423-13513423 on Build GRCh38
rs749734395 Chr.1: 13513225-13513225 on Build GRCh38
rs998819341 Chr.1: 13513237-13513237 on Build GRCh38
rs576738467 Chr.1: 13513335-13513335 on Build GRCh38
rs1048724553 Chr.1: 13513377-13513377 on Build GRCh38
rs887879732 Chr.1: 13513282-13513282 on Build GRCh38
rs966214062 Chr.1: 13513604-13513604 on Build GRCh38
rs559382090 Chr.1: 13513380-13513380 on Build GRCh38
rs972372028 Chr.1: 13513354-13513354 on Build GRCh38
rs535663196 Chr.1: 13513226-13513226 on Build GRCh38
rs1035980391 Chr.1: 13513395-13513395 on Build GRCh38
rs1007675552 Chr.1: 13513588-13513588 on Build GRCh38
rs573432291 Chr.1: 13513213-13513213 on Build GRCh38
rs543038330 Chr.1: 13513273-13513273 on Build GRCh38
rs751171664 Chr.1: 13513432-13513432 on Build GRCh38
rs972654913 Chr.1: 13513615-13513615 on Build GRCh38
rs948513626 Chr.1: 13513397-13513397 on Build GRCh38
rs775910726 Chr.1: 13513419-13513419 on Build GRCh38
rs533073268 Chr.1: 13513342-13513342 on Build GRCh38
rs545643839 Chr.1: 13513347-13513347 on Build GRCh38
rs919521539 Chr.1: 13513356-13513356 on Build GRCh38
rs773451960 Chr.1: 13513269-13513269 on Build GRCh38
rs953447959 Chr.1: 13513309-13513309 on Build GRCh38
rs765538446 Chr.1: 13513227-13513227 on Build GRCh38
rs756870422 Chr.1: 13513447-13513447 on Build GRCh38
rs548065630 Chr.1: 13513600-13513600 on Build GRCh38
rs894243359 Chr.1: 13513215-13513215 on Build GRCh38
rs956712870 Chr.1: 13513259-13513259 on Build GRCh38
rs746693001 Chr.1: 13513163-13513163 on Build GRCh38
rs776864586 Chr.1: 13513276-13513276 on Build GRCh38
rs901752055 Chr.1: 13513264-13513264 on Build GRCh38
rs1022305863 Chr.1: 13513544-13513544 on Build GRCh38
rs149475966 Chr.1: 13513275-13513275 on Build GRCh38
rs574429504 Chr.1: 13513240-13513240 on Build GRCh38
rs780609755 Chr.1: 13513461-13513461 on Build GRCh38
rs986663597 Chr.1: 13513341-13513341 on Build GRCh38
rs943186744 Chr.1: 13513231-13513231 on Build GRCh38
rs1045968642 Chr.1: 13513416-13513416 on Build GRCh38
rs780588099 Chr.1: 13513211-13513211 on Build GRCh38
rs374550402 Chr.1: 13513333-13513333 on Build GRCh38
rs1006281427 Chr.1: 13513332-13513332 on Build GRCh38
rs999162220 Chr.1: 13513435-13513435 on Build GRCh38

View Full Product Details