Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15153467-15153967 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM51
Entrez Gene ID
55092
Gene Name
transmembrane protein 51
Gene Aliases
C1orf72
UniGene
Hs.465305
Gene Chromosome Location
Chr.1: 15152418-15220480 on Build GRCh38
Transcripts
NM_001136216.1 NM_001136217.1 NM_001136218.1 NM_001319665.1 NM_018022.2 NR_135082.1 XM_011541676.1 XM_017001590.1

SNP Detail

SNP ID Chromosome Location
rs929493712 Chr.1: 15153850-15153850 on Build GRCh38
rs1001211716 Chr.1: 15153727-15153727 on Build GRCh38
rs993677387 Chr.1: 15153540-15153540 on Build GRCh38
rs910514733 Chr.1: 15153654-15153654 on Build GRCh38
rs139948566 Chr.1: 15153841-15153841 on Build GRCh38
rs575147683 Chr.1: 15153583-15153583 on Build GRCh38
rs376862448 Chr.1: 15153903-15153903 on Build GRCh38
rs1037756466 Chr.1: 15153527-15153527 on Build GRCh38
rs191469718 Chr.1: 15153537-15153537 on Build GRCh38
rs149853364 Chr.1: 15153886-15153886 on Build GRCh38
rs554221874 Chr.1: 15153925-15153925 on Build GRCh38
rs955963164 Chr.1: 15153567-15153567 on Build GRCh38
rs546192889 Chr.1: 15153542-15153542 on Build GRCh38
rs1032011586 Chr.1: 15153588-15153588 on Build GRCh38
rs1021398378 Chr.1: 15153719-15153719 on Build GRCh38
rs199778642 Chr.1: 15153536-15153536 on Build GRCh38
rs397687545 Chr.1: 15153535-15153536 on Build GRCh38
rs979559301 Chr.1: 15153640-15153640 on Build GRCh38
rs538480911 Chr.1: 15153819-15153819 on Build GRCh38
rs968503477 Chr.1: 15153599-15153599 on Build GRCh38
rs1005979342 Chr.1: 15153495-15153495 on Build GRCh38
rs926619611 Chr.1: 15153771-15153771 on Build GRCh38
rs1051919491 Chr.1: 15153902-15153902 on Build GRCh38
rs750631641 Chr.1: 15153696-15153696 on Build GRCh38
rs988639386 Chr.1: 15153576-15153576 on Build GRCh38
rs942106977 Chr.1: 15153510-15153510 on Build GRCh38
rs756393337 Chr.1: 15153803-15153804 on Build GRCh38
rs1023016238 Chr.1: 15153551-15153551 on Build GRCh38
rs572756990 Chr.1: 15153653-15153653 on Build GRCh38
rs1002760854 Chr.1: 15153564-15153564 on Build GRCh38
rs943362456 Chr.1: 15153684-15153684 on Build GRCh38
rs905171013 Chr.1: 15153508-15153508 on Build GRCh38
rs956783175 Chr.1: 15153612-15153612 on Build GRCh38
rs533446648 Chr.1: 15153847-15153847 on Build GRCh38
rs968767547 Chr.1: 15153723-15153723 on Build GRCh38
rs742675 Chr.1: 15153552-15153552 on Build GRCh38
rs540147854 Chr.1: 15153698-15153699 on Build GRCh38
rs923524134 Chr.1: 15153708-15153708 on Build GRCh38
rs949352021 Chr.1: 15153710-15153710 on Build GRCh38
rs756538347 Chr.1: 15153778-15153778 on Build GRCh38
rs869227385 Chr.1: 15153531-15153532 on Build GRCh38
rs742674 Chr.1: 15153566-15153566 on Build GRCh38
rs905901874 Chr.1: 15153564-15153586 on Build GRCh38
rs1021493324 Chr.1: 15153580-15153580 on Build GRCh38
rs5772620 Chr.1: 15153530-15153531 on Build GRCh38
rs7530108 Chr.1: 15153790-15153790 on Build GRCh38
rs866902026 Chr.1: 15153795-15153795 on Build GRCh38
rs1017776228 Chr.1: 15153942-15153942 on Build GRCh38
rs980655300 Chr.1: 15153728-15153728 on Build GRCh38
rs3737685 Chr.1: 15153909-15153909 on Build GRCh38

View Full Product Details