Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27925204-27925745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDX1
Entrez Gene ID
3651
Gene Name
pancreatic and duodenal homeobox 1
Gene Aliases
GSF, IDX-1, IPF1, IUF1, MODY4, PAGEN1, PDX-1, STF-1
UniGene
Hs.32938
Gene Chromosome Location
Chr.13: 27919982-27926314 on Build GRCh38
Transcripts
NM_000209.3

SNP Detail

SNP ID Chromosome Location
rs1050351636 Chr.13: 27925241-27925241 on Build GRCh38
rs941562010 Chr.13: 27925467-27925467 on Build GRCh38
rs71433245 Chr.13: 27925444-27925444 on Build GRCh38
rs749161312 Chr.13: 27925446-27925447 on Build GRCh38
rs543696560 Chr.13: 27925466-27925466 on Build GRCh38
rs558803711 Chr.13: 27925277-27925277 on Build GRCh38
rs1052228687 Chr.13: 27925683-27925683 on Build GRCh38
rs112637982 Chr.13: 27925716-27925716 on Build GRCh38
rs900006009 Chr.13: 27925339-27925341 on Build GRCh38
rs932682831 Chr.13: 27925369-27925369 on Build GRCh38
rs1001153429 Chr.13: 27925615-27925616 on Build GRCh38
rs932839795 Chr.13: 27925592-27925594 on Build GRCh38
rs200414219 Chr.13: 27925264-27925265 on Build GRCh38
rs556636257 Chr.13: 27925246-27925246 on Build GRCh38
rs201227980 Chr.13: 27925465-27925467 on Build GRCh38
rs11620272 Chr.13: 27925558-27925558 on Build GRCh38
rs903792294 Chr.13: 27925242-27925242 on Build GRCh38
rs12560601 Chr.13: 27925582-27925582 on Build GRCh38
rs541144902 Chr.13: 27925706-27925706 on Build GRCh38
rs903746479 Chr.13: 27925493-27925493 on Build GRCh38
rs959203747 Chr.13: 27925519-27925519 on Build GRCh38
rs892520918 Chr.13: 27925343-27925343 on Build GRCh38
rs1024957406 Chr.13: 27925475-27925475 on Build GRCh38
rs1013679855 Chr.13: 27925367-27925367 on Build GRCh38
rs71993976 Chr.13: 27925553-27925555 on Build GRCh38
rs113506161 Chr.13: 27925343-27925344 on Build GRCh38
rs11617239 Chr.13: 27925552-27925552 on Build GRCh38
rs1009600899 Chr.13: 27925425-27925425 on Build GRCh38
rs576516071 Chr.13: 27925247-27925247 on Build GRCh38
rs1027221289 Chr.13: 27925540-27925540 on Build GRCh38
rs139276646 Chr.13: 27925447-27925455 on Build GRCh38
rs1033644298 Chr.13: 27925670-27925670 on Build GRCh38
rs143062765 Chr.13: 27925495-27925495 on Build GRCh38
rs374204593 Chr.13: 27925546-27925546 on Build GRCh38
rs201597488 Chr.13: 27925349-27925351 on Build GRCh38
rs373152010 Chr.13: 27925444-27925473 on Build GRCh38
rs982907578 Chr.13: 27925544-27925544 on Build GRCh38
rs560079500 Chr.13: 27925607-27925610 on Build GRCh38
rs11617238 Chr.13: 27925537-27925537 on Build GRCh38
rs561511065 Chr.13: 27925416-27925416 on Build GRCh38
rs941637120 Chr.13: 27925292-27925294 on Build GRCh38
rs986239239 Chr.13: 27925608-27925608 on Build GRCh38
rs60008665 Chr.13: 27925564-27925566 on Build GRCh38
rs891123842 Chr.13: 27925391-27925391 on Build GRCh38
rs750218974 Chr.13: 27925295-27925343 on Build GRCh38
rs567194464 Chr.13: 27925375-27925377 on Build GRCh38
rs1024705968 Chr.13: 27925542-27925542 on Build GRCh38
rs541297096 Chr.13: 27925368-27925368 on Build GRCh38
rs548132595 Chr.13: 27925593-27925593 on Build GRCh38
rs372179991 Chr.13: 27925273-27925273 on Build GRCh38
rs538988629 Chr.13: 27925264-27925264 on Build GRCh38
rs371209265 Chr.13: 27925563-27925564 on Build GRCh38
rs894976456 Chr.13: 27925691-27925691 on Build GRCh38
rs925216352 Chr.13: 27925626-27925626 on Build GRCh38
rs563597142 Chr.13: 27925487-27925487 on Build GRCh38
rs78382412 Chr.13: 27925572-27925572 on Build GRCh38
rs1033898357 Chr.13: 27925518-27925518 on Build GRCh38
rs386769404 Chr.13: 27925488-27925489 on Build GRCh38
rs11617235 Chr.13: 27925482-27925482 on Build GRCh38
rs74799049 Chr.13: 27925564-27925564 on Build GRCh38
rs190800391 Chr.13: 27925545-27925545 on Build GRCh38
rs1020852139 Chr.13: 27925484-27925484 on Build GRCh38
rs892322969 Chr.13: 27925701-27925701 on Build GRCh38
rs551783385 Chr.13: 27925278-27925278 on Build GRCh38
rs761083009 Chr.13: 27925662-27925662 on Build GRCh38
rs112525679 Chr.13: 27925723-27925723 on Build GRCh38
rs11620280 Chr.13: 27925574-27925574 on Build GRCh38
rs1051220684 Chr.13: 27925390-27925390 on Build GRCh38
rs147791430 Chr.13: 27925580-27925580 on Build GRCh38
rs908888622 Chr.13: 27925567-27925567 on Build GRCh38
rs537340336 Chr.13: 27925232-27925232 on Build GRCh38
rs1052300577 Chr.13: 27925326-27925326 on Build GRCh38
rs771050094 Chr.13: 27925398-27925400 on Build GRCh38
rs1013394384 Chr.13: 27925700-27925700 on Build GRCh38
rs921428973 Chr.13: 27925583-27925591 on Build GRCh38
rs574837616 Chr.13: 27925445-27925445 on Build GRCh38
rs764496076 Chr.13: 27925571-27925572 on Build GRCh38
rs936580078 Chr.13: 27925645-27925645 on Build GRCh38
rs572467700 Chr.13: 27925328-27925328 on Build GRCh38
rs537515100 Chr.13: 27925543-27925543 on Build GRCh38
rs9554205 Chr.13: 27925604-27925604 on Build GRCh38
rs537204017 Chr.13: 27925605-27925605 on Build GRCh38

View Full Product Details