Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 27988411-27988907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
URAD
Entrez Gene ID
646625
Gene Name
ureidoimidazoline (2-oxo-4-hydroxy-4-carboxy-5-) decarboxylase
Gene Aliases
PRHOXNB
UniGene
Hs.705356
Gene Chromosome Location
Chr.13: 27978106-27988637 on Build GRCh38
Transcripts
NM_001105577.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
696847 c.21C>T p.N7N Chr.13: 27988617-27988617 on Build GRCh38
6817723 c.147delT p.F49fs*45 Chr.13: 27988491-27988491 on Build GRCh38
5052105 c.16G>A p.V6I Chr.13: 27988622-27988622 on Build GRCh38
4823537 c.67G>C p.E23Q Chr.13: 27988571-27988571 on Build GRCh38
396414 c.107G>A p.R36Q Chr.13: 27988531-27988531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779009570 Chr.13: 27988663-27988663 on Build GRCh38
rs771396757 Chr.13: 27988442-27988442 on Build GRCh38
rs775279767 Chr.13: 27988682-27988682 on Build GRCh38
rs58908725 Chr.13: 27988785-27988785 on Build GRCh38
rs367643288 Chr.13: 27988496-27988496 on Build GRCh38
rs866313495 Chr.13: 27988710-27988710 on Build GRCh38
rs781575495 Chr.13: 27988511-27988511 on Build GRCh38
rs541769135 Chr.13: 27988715-27988715 on Build GRCh38
rs369657865 Chr.13: 27988661-27988661 on Build GRCh38
rs868346858 Chr.13: 27988567-27988567 on Build GRCh38
rs370378739 Chr.13: 27988724-27988724 on Build GRCh38
rs186848050 Chr.13: 27988726-27988726 on Build GRCh38
rs755271949 Chr.13: 27988662-27988662 on Build GRCh38
rs781417316 Chr.13: 27988443-27988443 on Build GRCh38
rs3897926 Chr.13: 27988468-27988468 on Build GRCh38
rs749372365 Chr.13: 27988557-27988557 on Build GRCh38
rs181737747 Chr.13: 27988671-27988671 on Build GRCh38
rs944977681 Chr.13: 27988721-27988721 on Build GRCh38
rs776456405 Chr.13: 27988601-27988601 on Build GRCh38
rs759376316 Chr.13: 27988604-27988604 on Build GRCh38
rs997775772 Chr.13: 27988639-27988639 on Build GRCh38
rs762601122 Chr.13: 27988618-27988618 on Build GRCh38
rs7339283 Chr.13: 27988664-27988664 on Build GRCh38
rs937261788 Chr.13: 27988718-27988718 on Build GRCh38
rs267603794 Chr.13: 27988596-27988596 on Build GRCh38
rs760532638 Chr.13: 27988462-27988462 on Build GRCh38
rs371322824 Chr.13: 27988499-27988499 on Build GRCh38
rs563513361 Chr.13: 27988750-27988750 on Build GRCh38
rs112398440 Chr.13: 27988737-27988737 on Build GRCh38
rs572691016 Chr.13: 27988713-27988713 on Build GRCh38
rs1006721708 Chr.13: 27988585-27988585 on Build GRCh38
rs370796527 Chr.13: 27988457-27988457 on Build GRCh38
rs553150890 Chr.13: 27988709-27988709 on Build GRCh38
rs753597768 Chr.13: 27988472-27988472 on Build GRCh38
rs746253853 Chr.13: 27988531-27988531 on Build GRCh38
rs367941920 Chr.13: 27988622-27988622 on Build GRCh38
rs138307982 Chr.13: 27988766-27988766 on Build GRCh38
rs111850310 Chr.13: 27988568-27988568 on Build GRCh38
rs770879827 Chr.13: 27988667-27988667 on Build GRCh38
rs769514810 Chr.13: 27988680-27988680 on Build GRCh38
rs150488539 Chr.13: 27988730-27988730 on Build GRCh38
rs1030306274 Chr.13: 27988801-27988801 on Build GRCh38
rs535381414 Chr.13: 27988654-27988654 on Build GRCh38
rs746915022 Chr.13: 27988569-27988569 on Build GRCh38
rs375161217 Chr.13: 27988651-27988651 on Build GRCh38
rs887846269 Chr.13: 27988581-27988581 on Build GRCh38
rs1032462222 Chr.13: 27988834-27988834 on Build GRCh38
rs1045666639 Chr.13: 27988705-27988705 on Build GRCh38
rs780477671 Chr.13: 27988555-27988555 on Build GRCh38
rs568788742 Chr.13: 27988808-27988808 on Build GRCh38
rs559154441 Chr.13: 27988704-27988704 on Build GRCh38
rs757962505 Chr.13: 27988480-27988480 on Build GRCh38
rs535238504 Chr.13: 27988796-27988796 on Build GRCh38
rs775675781 Chr.13: 27988451-27988451 on Build GRCh38
rs376394906 Chr.13: 27988696-27988696 on Build GRCh38
rs771206096 Chr.13: 27988871-27988871 on Build GRCh38
rs763448634 Chr.13: 27988439-27988439 on Build GRCh38
rs1014893480 Chr.13: 27988595-27988595 on Build GRCh38
rs775192997 Chr.13: 27988607-27988607 on Build GRCh38
rs545555367 Chr.13: 27988697-27988697 on Build GRCh38
rs770115628 Chr.13: 27988532-27988532 on Build GRCh38
rs936168851 Chr.13: 27988749-27988749 on Build GRCh38
rs953643605 Chr.13: 27988666-27988666 on Build GRCh38
rs768786975 Chr.13: 27988565-27988565 on Build GRCh38
rs907019290 Chr.13: 27988521-27988521 on Build GRCh38
rs746047937 Chr.13: 27988445-27988445 on Build GRCh38
rs752181906 Chr.13: 27988479-27988479 on Build GRCh38
rs76294919 Chr.13: 27988452-27988452 on Build GRCh38
rs1028476190 Chr.13: 27988865-27988865 on Build GRCh38
rs989896049 Chr.13: 27988872-27988872 on Build GRCh38
rs958484652 Chr.13: 27988843-27988843 on Build GRCh38
rs371238042 Chr.13: 27988475-27988475 on Build GRCh38
rs375509965 Chr.13: 27988678-27988678 on Build GRCh38
rs537316275 Chr.13: 27988847-27988847 on Build GRCh38
rs368115210 Chr.13: 27988765-27988765 on Build GRCh38
rs770805634 Chr.13: 27988570-27988570 on Build GRCh38
rs750782891 Chr.13: 27988645-27988645 on Build GRCh38
rs113311108 Chr.13: 27988852-27988852 on Build GRCh38
rs9554219 Chr.13: 27988805-27988805 on Build GRCh38
rs770167025 Chr.13: 27988448-27988448 on Build GRCh38
rs375493324 Chr.13: 27988458-27988458 on Build GRCh38
rs3897927 Chr.13: 27988764-27988764 on Build GRCh38
rs774307922 Chr.13: 27988566-27988566 on Build GRCh38
rs374399842 Chr.13: 27988606-27988606 on Build GRCh38
rs764911617 Chr.13: 27988477-27988477 on Build GRCh38
rs757747107 Chr.13: 27988632-27988632 on Build GRCh38
rs755587451 Chr.13: 27988450-27988451 on Build GRCh38
rs371642764 Chr.13: 27988635-27988635 on Build GRCh38
rs7323025 Chr.13: 27988685-27988685 on Build GRCh38
rs376297191 Chr.13: 27988483-27988483 on Build GRCh38

View Full Product Details