Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28311581-28312114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT1
Entrez Gene ID
2321
Gene Name
fms related tyrosine kinase 1
Gene Aliases
FLT, FLT-1, VEGFR-1, VEGFR1
UniGene
Hs.594454
Gene Chromosome Location
Chr.13: 28300346-28495128 on Build GRCh38
Transcripts
NM_002019.4 XM_017020485.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4786217 c.509T>G p.F170C Chr.13: 28312045-28312045 on Build GRCh38
5608865 c.694G>A p.D232N Chr.13: 28311600-28311600 on Build GRCh38
6964891 c.3490C>T p.Q1164* Chr.13: 28311995-28311995 on Build GRCh38
1582173 c.3421C>A p.P1141T Chr.13: 28312064-28312064 on Build GRCh38
6963748 c.490C>A p.P164T Chr.13: 28312064-28312064 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567015873 Chr.13: 28311958-28311958 on Build GRCh38
rs77857617 Chr.13: 28311843-28311843 on Build GRCh38
rs778937990 Chr.13: 28311757-28311757 on Build GRCh38
rs61763564 Chr.13: 28311820-28311820 on Build GRCh38
rs148958662 Chr.13: 28312006-28312006 on Build GRCh38
rs773436133 Chr.13: 28312091-28312091 on Build GRCh38
rs868190326 Chr.13: 28311658-28311658 on Build GRCh38
rs750549806 Chr.13: 28311747-28311747 on Build GRCh38
rs777484833 Chr.13: 28311779-28311779 on Build GRCh38
rs114152863 Chr.13: 28311792-28311792 on Build GRCh38
rs757545042 Chr.13: 28311722-28311722 on Build GRCh38
rs767803039 Chr.13: 28312072-28312072 on Build GRCh38
rs903838562 Chr.13: 28311929-28311929 on Build GRCh38
rs765799505 Chr.13: 28311657-28311657 on Build GRCh38
rs756222379 Chr.13: 28311962-28311962 on Build GRCh38
rs372303417 Chr.13: 28312032-28312032 on Build GRCh38
rs781361526 Chr.13: 28311739-28311739 on Build GRCh38
rs759083224 Chr.13: 28311641-28311641 on Build GRCh38
rs531666463 Chr.13: 28311832-28311832 on Build GRCh38
rs55673217 Chr.13: 28311673-28311673 on Build GRCh38
rs374608009 Chr.13: 28311781-28311781 on Build GRCh38
rs61763563 Chr.13: 28311951-28311951 on Build GRCh38
rs762195216 Chr.13: 28312071-28312071 on Build GRCh38
rs573870646 Chr.13: 28312014-28312014 on Build GRCh38
rs561880240 Chr.13: 28312002-28312002 on Build GRCh38
rs7328692 Chr.13: 28311976-28311976 on Build GRCh38
rs113867782 Chr.13: 28311802-28311802 on Build GRCh38
rs150821981 Chr.13: 28311999-28311999 on Build GRCh38
rs778692239 Chr.13: 28311616-28311616 on Build GRCh38
rs560320808 Chr.13: 28312062-28312062 on Build GRCh38
rs776081415 Chr.13: 28311945-28311945 on Build GRCh38
rs756256507 Chr.13: 28312088-28312088 on Build GRCh38
rs539518963 Chr.13: 28311647-28311647 on Build GRCh38
rs932217572 Chr.13: 28311877-28311877 on Build GRCh38
rs147277608 Chr.13: 28311873-28311873 on Build GRCh38
rs370382868 Chr.13: 28311780-28311780 on Build GRCh38
rs145693340 Chr.13: 28311998-28311998 on Build GRCh38
rs138094419 Chr.13: 28311829-28311829 on Build GRCh38
rs774792502 Chr.13: 28312068-28312068 on Build GRCh38
rs555066797 Chr.13: 28311919-28311920 on Build GRCh38
rs747215759 Chr.13: 28311862-28311863 on Build GRCh38
rs771911806 Chr.13: 28311776-28311776 on Build GRCh38
rs747998054 Chr.13: 28311617-28311617 on Build GRCh38
rs764087668 Chr.13: 28311626-28311626 on Build GRCh38
rs776343811 Chr.13: 28311636-28311636 on Build GRCh38
rs61950205 Chr.13: 28311842-28311842 on Build GRCh38
rs772551966 Chr.13: 28311861-28311862 on Build GRCh38
rs143590198 Chr.13: 28311855-28311855 on Build GRCh38
rs377442842 Chr.13: 28311667-28311667 on Build GRCh38
rs199706466 Chr.13: 28311694-28311694 on Build GRCh38
rs192757238 Chr.13: 28311899-28311899 on Build GRCh38
rs941841930 Chr.13: 28311764-28311764 on Build GRCh38
rs762144429 Chr.13: 28311954-28311954 on Build GRCh38
rs971646396 Chr.13: 28312042-28312042 on Build GRCh38
rs763429331 Chr.13: 28311948-28311948 on Build GRCh38
rs140861115 Chr.13: 28311623-28311623 on Build GRCh38
rs769123519 Chr.13: 28311943-28311943 on Build GRCh38
rs1000832120 Chr.13: 28311930-28311930 on Build GRCh38
rs773210044 Chr.13: 28311947-28311947 on Build GRCh38
rs777732747 Chr.13: 28311994-28311994 on Build GRCh38
rs368097084 Chr.13: 28311950-28311950 on Build GRCh38
rs954489468 Chr.13: 28312031-28312031 on Build GRCh38
rs764310611 Chr.13: 28311687-28311687 on Build GRCh38
rs770692157 Chr.13: 28311627-28311627 on Build GRCh38
rs139434208 Chr.13: 28312081-28312081 on Build GRCh38
rs2296186 Chr.13: 28311956-28311956 on Build GRCh38
rs751909538 Chr.13: 28311715-28311715 on Build GRCh38
rs1033722050 Chr.13: 28311920-28311920 on Build GRCh38
rs933012783 Chr.13: 28311662-28311662 on Build GRCh38
rs758330992 Chr.13: 28311986-28311986 on Build GRCh38
rs768621630 Chr.13: 28311606-28311606 on Build GRCh38
rs572908814 Chr.13: 28311874-28311874 on Build GRCh38
rs143577219 Chr.13: 28311786-28311786 on Build GRCh38
rs374796929 Chr.13: 28311622-28311622 on Build GRCh38
rs141658286 Chr.13: 28311921-28311921 on Build GRCh38
rs538506425 Chr.13: 28312067-28312067 on Build GRCh38
rs756137527 Chr.13: 28311752-28311752 on Build GRCh38
rs139091615 Chr.13: 28311917-28311917 on Build GRCh38
rs759150886 Chr.13: 28311942-28311942 on Build GRCh38
rs776439365 Chr.13: 28311856-28311856 on Build GRCh38
rs776377967 Chr.13: 28311784-28311784 on Build GRCh38
rs562619634 Chr.13: 28311822-28311822 on Build GRCh38
rs138652408 Chr.13: 28311685-28311685 on Build GRCh38
rs1040544036 Chr.13: 28311823-28311823 on Build GRCh38
rs375433146 Chr.13: 28311871-28311874 on Build GRCh38
rs764941678 Chr.13: 28311920-28311920 on Build GRCh38
rs528850307 Chr.13: 28311814-28311814 on Build GRCh38
rs66484805 Chr.13: 28311877-28311878 on Build GRCh38
rs148901042 Chr.13: 28311727-28311727 on Build GRCh38
rs148817356 Chr.13: 28311758-28311758 on Build GRCh38
rs1034287323 Chr.13: 28312077-28312077 on Build GRCh38
rs375487770 Chr.13: 28312047-28312047 on Build GRCh38

View Full Product Details