Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28658924-28659427 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
POMP
Entrez Gene ID
51371
Gene Name
proteasome maturation protein
Gene Aliases
C13orf12, HSPC014, PNAS-110, UMP1
UniGene
Hs.268742
Gene Chromosome Location
Chr.13: 28659004-28678957 on Build GRCh38
Transcripts
NM_015932.5

SNP Detail

SNP ID Chromosome Location
rs761573969 Chr.13: 28659157-28659158 on Build GRCh38
rs771652313 Chr.13: 28659164-28659164 on Build GRCh38
rs960463677 Chr.13: 28659049-28659049 on Build GRCh38
rs888179376 Chr.13: 28659263-28659263 on Build GRCh38
rs918761117 Chr.13: 28658973-28658973 on Build GRCh38
rs552853976 Chr.13: 28659315-28659315 on Build GRCh38
rs766897923 Chr.13: 28659135-28659135 on Build GRCh38
rs962731264 Chr.13: 28659076-28659076 on Build GRCh38
rs983980708 Chr.13: 28659031-28659031 on Build GRCh38
rs942652654 Chr.13: 28659022-28659022 on Build GRCh38
rs944969100 Chr.13: 28659205-28659205 on Build GRCh38
rs550484205 Chr.13: 28659152-28659152 on Build GRCh38
rs754830362 Chr.13: 28659212-28659212 on Build GRCh38
rs772572905 Chr.13: 28659165-28659165 on Build GRCh38
rs113700550 Chr.13: 28659189-28659189 on Build GRCh38
rs1040602001 Chr.13: 28659388-28659388 on Build GRCh38
rs767136186 Chr.13: 28658968-28658968 on Build GRCh38
rs763058898 Chr.13: 28659199-28659199 on Build GRCh38
rs939961140 Chr.13: 28659035-28659035 on Build GRCh38
rs1044417110 Chr.13: 28658957-28658957 on Build GRCh38
rs891238474 Chr.13: 28659016-28659016 on Build GRCh38
rs995604076 Chr.13: 28659284-28659284 on Build GRCh38
rs1019557030 Chr.13: 28659006-28659006 on Build GRCh38
rs989335697 Chr.13: 28659356-28659356 on Build GRCh38
rs539211409 Chr.13: 28659196-28659196 on Build GRCh38
rs986931026 Chr.13: 28659294-28659294 on Build GRCh38
rs974343189 Chr.13: 28659079-28659079 on Build GRCh38
rs112927425 Chr.13: 28659188-28659188 on Build GRCh38
rs7330455 Chr.13: 28659227-28659227 on Build GRCh38
rs758567259 Chr.13: 28659141-28659141 on Build GRCh38
rs112576010 Chr.13: 28659167-28659167 on Build GRCh38
rs765698118 Chr.13: 28659116-28659116 on Build GRCh38
rs547781197 Chr.13: 28659090-28659090 on Build GRCh38
rs914673185 Chr.13: 28659333-28659333 on Build GRCh38
rs918862935 Chr.13: 28659083-28659083 on Build GRCh38
rs1005299683 Chr.13: 28659278-28659279 on Build GRCh38
rs952354415 Chr.13: 28659111-28659111 on Build GRCh38
rs373450514 Chr.13: 28659134-28659134 on Build GRCh38
rs554552544 Chr.13: 28659241-28659241 on Build GRCh38
rs941635300 Chr.13: 28658949-28658949 on Build GRCh38
rs1051166990 Chr.13: 28659015-28659015 on Build GRCh38
rs904483846 Chr.13: 28659056-28659056 on Build GRCh38
rs531435814 Chr.13: 28659132-28659132 on Build GRCh38
rs572507865 Chr.13: 28659365-28659365 on Build GRCh38
rs965138868 Chr.13: 28659013-28659013 on Build GRCh38
rs951506003 Chr.13: 28659129-28659129 on Build GRCh38
rs947392993 Chr.13: 28659349-28659349 on Build GRCh38
rs766452964 Chr.13: 28659249-28659249 on Build GRCh38
rs999346357 Chr.13: 28659039-28659039 on Build GRCh38
rs912620533 Chr.13: 28659155-28659155 on Build GRCh38
rs563184710 Chr.13: 28659030-28659030 on Build GRCh38
rs983738871 Chr.13: 28659123-28659123 on Build GRCh38
rs1023307895 Chr.13: 28659054-28659054 on Build GRCh38
rs570293774 Chr.13: 28659154-28659154 on Build GRCh38
rs1009077647 Chr.13: 28659005-28659005 on Build GRCh38
rs112368783 Chr.13: 28659090-28659090 on Build GRCh38
rs1618098 Chr.13: 28659285-28659285 on Build GRCh38
rs574402095 Chr.13: 28658959-28658959 on Build GRCh38
rs543265240 Chr.13: 28658980-28658980 on Build GRCh38
rs962391835 Chr.13: 28659276-28659276 on Build GRCh38
rs574091904 Chr.13: 28659257-28659257 on Build GRCh38
rs777375693 Chr.13: 28659226-28659226 on Build GRCh38
rs766243253 Chr.13: 28659203-28659203 on Build GRCh38
rs576612933 Chr.13: 28659038-28659038 on Build GRCh38
rs913197147 Chr.13: 28659361-28659361 on Build GRCh38
rs779853022 Chr.13: 28659146-28659146 on Build GRCh38
rs28372944 Chr.13: 28659206-28659206 on Build GRCh38
rs1010824014 Chr.13: 28659028-28659028 on Build GRCh38
rs776926791 Chr.13: 28659051-28659051 on Build GRCh38
rs59496599 Chr.13: 28659369-28659369 on Build GRCh38
rs556815417 Chr.13: 28659264-28659264 on Build GRCh38
rs923853246 Chr.13: 28659378-28659378 on Build GRCh38
rs974613239 Chr.13: 28658964-28658964 on Build GRCh38
rs377702347 Chr.13: 28659105-28659105 on Build GRCh38
rs749012649 Chr.13: 28659149-28659149 on Build GRCh38
rs964242884 Chr.13: 28659302-28659302 on Build GRCh38
rs374606472 Chr.13: 28659060-28659060 on Build GRCh38
rs951621477 Chr.13: 28659286-28659286 on Build GRCh38
rs756047393 Chr.13: 28659201-28659201 on Build GRCh38
rs199642264 Chr.13: 28659200-28659200 on Build GRCh38
rs908469239 Chr.13: 28659032-28659032 on Build GRCh38
rs565603764 Chr.13: 28659026-28659026 on Build GRCh38
rs776659765 Chr.13: 28659385-28659385 on Build GRCh38
rs778680440 Chr.13: 28659156-28659156 on Build GRCh38
rs756025303 Chr.13: 28659145-28659145 on Build GRCh38
rs932260238 Chr.13: 28659402-28659402 on Build GRCh38
rs750418216 Chr.13: 28659142-28659142 on Build GRCh38
rs747883302 Chr.13: 28659221-28659221 on Build GRCh38
rs778540639 Chr.13: 28659219-28659219 on Build GRCh38
rs765000515 Chr.13: 28659164-28659165 on Build GRCh38
rs747751391 Chr.13: 28659157-28659157 on Build GRCh38
rs925798374 Chr.13: 28659311-28659311 on Build GRCh38
rs975171789 Chr.13: 28659021-28659021 on Build GRCh38
rs1039738805 Chr.13: 28659024-28659024 on Build GRCh38
rs777140811 Chr.13: 28659179-28659179 on Build GRCh38
rs996516877 Chr.13: 28659108-28659108 on Build GRCh38
rs1000417646 Chr.13: 28658975-28658975 on Build GRCh38
rs1031929213 Chr.13: 28658981-28658981 on Build GRCh38
rs1022675801 Chr.13: 28659029-28659029 on Build GRCh38
rs1023839248 Chr.13: 28659274-28659274 on Build GRCh38
rs759869912 Chr.13: 28659138-28659138 on Build GRCh38
rs762173139 Chr.13: 28659068-28659068 on Build GRCh38
rs545995833 Chr.13: 28659301-28659301 on Build GRCh38
rs1042131216 Chr.13: 28659354-28659354 on Build GRCh38
rs1024658529 Chr.13: 28659299-28659300 on Build GRCh38
rs527752032 Chr.13: 28659067-28659067 on Build GRCh38
rs1019109312 Chr.13: 28659089-28659089 on Build GRCh38
rs759923011 Chr.13: 28659181-28659181 on Build GRCh38
rs186909938 Chr.13: 28659047-28659047 on Build GRCh38
rs956377868 Chr.13: 28658987-28658988 on Build GRCh38
rs900751057 Chr.13: 28659390-28659390 on Build GRCh38
rs771579292 Chr.13: 28659239-28659239 on Build GRCh38
rs757936789 Chr.13: 28659225-28659225 on Build GRCh38
rs754208042 Chr.13: 28659137-28659137 on Build GRCh38
rs930170628 Chr.13: 28658990-28658990 on Build GRCh38
rs545127969 Chr.13: 28659044-28659044 on Build GRCh38
rs765684384 Chr.13: 28659191-28659191 on Build GRCh38
rs536984217 Chr.13: 28659262-28659262 on Build GRCh38
rs892032346 Chr.13: 28659070-28659070 on Build GRCh38
rs993190902 Chr.13: 28659151-28659151 on Build GRCh38
rs376702047 Chr.13: 28659171-28659171 on Build GRCh38
rs936096892 Chr.13: 28659172-28659172 on Build GRCh38

View Full Product Details