Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 31761395-31761941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RXFP2
Entrez Gene ID
122042
Gene Name
relaxin/insulin like family peptide receptor 2
Gene Aliases
GPR106, GREAT, INSL3R, LGR8, LGR8.1, RXFPR2
UniGene
Hs.680763
Gene Chromosome Location
Chr.13: 31739542-31802872 on Build GRCh38
Transcripts
NM_001166058.1 NM_130806.3 XM_017020389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6341262 c.253G>T p.G85* Chr.13: 31761735-31761735 on Build GRCh38
3955527 c.313G>T p.E105* Chr.13: 31761795-31761795 on Build GRCh38
147661 c.307A>C p.T103P Chr.13: 31761789-31761789 on Build GRCh38
5970957 c.281A>G p.H94R Chr.13: 31761763-31761763 on Build GRCh38
2070977 c.283G>A p.G95R Chr.13: 31761765-31761765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373570227 Chr.13: 31761742-31761742 on Build GRCh38
rs771127707 Chr.13: 31761829-31761829 on Build GRCh38
rs771198129 Chr.13: 31761795-31761795 on Build GRCh38
rs1536635 Chr.13: 31761903-31761903 on Build GRCh38
rs9532478 Chr.13: 31761703-31761703 on Build GRCh38
rs780595679 Chr.13: 31761763-31761763 on Build GRCh38
rs778938790 Chr.13: 31761426-31761426 on Build GRCh38
rs184512681 Chr.13: 31761546-31761546 on Build GRCh38
rs748172742 Chr.13: 31761557-31761557 on Build GRCh38
rs778112518 Chr.13: 31761736-31761738 on Build GRCh38
rs767730385 Chr.13: 31761701-31761701 on Build GRCh38
rs531281283 Chr.13: 31761776-31761776 on Build GRCh38
rs1010200678 Chr.13: 31761905-31761905 on Build GRCh38
rs551201424 Chr.13: 31761904-31761904 on Build GRCh38
rs1014212474 Chr.13: 31761748-31761748 on Build GRCh38
rs774703024 Chr.13: 31761690-31761690 on Build GRCh38
rs1035549362 Chr.13: 31761624-31761624 on Build GRCh38
rs9532479 Chr.13: 31761743-31761743 on Build GRCh38
rs760740889 Chr.13: 31761709-31761709 on Build GRCh38
rs148840796 Chr.13: 31761779-31761779 on Build GRCh38
rs541741749 Chr.13: 31761478-31761478 on Build GRCh38
rs758481894 Chr.13: 31761647-31761647 on Build GRCh38
rs779166506 Chr.13: 31761780-31761780 on Build GRCh38
rs758330882 Chr.13: 31761733-31761733 on Build GRCh38
rs762964942 Chr.13: 31761851-31761851 on Build GRCh38
rs751277993 Chr.13: 31761737-31761737 on Build GRCh38
rs890118384 Chr.13: 31761542-31761542 on Build GRCh38
rs140976146 Chr.13: 31761889-31761889 on Build GRCh38
rs748505158 Chr.13: 31761798-31761798 on Build GRCh38
rs956000792 Chr.13: 31761541-31761541 on Build GRCh38
rs868153087 Chr.13: 31761436-31761436 on Build GRCh38
rs35384029 Chr.13: 31761483-31761483 on Build GRCh38
rs1051829026 Chr.13: 31761524-31761524 on Build GRCh38
rs777096033 Chr.13: 31761869-31761869 on Build GRCh38
rs951371919 Chr.13: 31761683-31761683 on Build GRCh38
rs773421766 Chr.13: 31761809-31761809 on Build GRCh38
rs188981298 Chr.13: 31761592-31761592 on Build GRCh38
rs760891727 Chr.13: 31761814-31761814 on Build GRCh38
rs1032998390 Chr.13: 31761532-31761532 on Build GRCh38
rs1036665708 Chr.13: 31761718-31761718 on Build GRCh38
rs765276273 Chr.13: 31761728-31761728 on Build GRCh38
rs909655448 Chr.13: 31761699-31761699 on Build GRCh38
rs931759068 Chr.13: 31761497-31761497 on Build GRCh38
rs565347402 Chr.13: 31761692-31761692 on Build GRCh38
rs867167451 Chr.13: 31761596-31761596 on Build GRCh38
rs1023813218 Chr.13: 31761573-31761573 on Build GRCh38
rs932377502 Chr.13: 31761622-31761622 on Build GRCh38
rs1003627479 Chr.13: 31761623-31761623 on Build GRCh38
rs199862083 Chr.13: 31761850-31761850 on Build GRCh38
rs973612451 Chr.13: 31761724-31761724 on Build GRCh38
rs922156247 Chr.13: 31761556-31761556 on Build GRCh38
rs1011050154 Chr.13: 31761566-31761566 on Build GRCh38
rs752664547 Chr.13: 31761731-31761731 on Build GRCh38
rs912113133 Chr.13: 31761545-31761545 on Build GRCh38
rs921649297 Chr.13: 31761493-31761493 on Build GRCh38
rs969837392 Chr.13: 31761577-31761577 on Build GRCh38
rs764045429 Chr.13: 31761853-31761853 on Build GRCh38
rs745375797 Chr.13: 31761760-31761760 on Build GRCh38
rs772187543 Chr.13: 31761801-31761801 on Build GRCh38
rs1014770538 Chr.13: 31761702-31761702 on Build GRCh38
rs746339710 Chr.13: 31761874-31761874 on Build GRCh38
rs551970530 Chr.13: 31761668-31761668 on Build GRCh38
rs1052668211 Chr.13: 31761554-31761554 on Build GRCh38
rs768995785 Chr.13: 31761681-31761681 on Build GRCh38
rs747250203 Chr.13: 31761544-31761544 on Build GRCh38

View Full Product Details