Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 32404091-32404589 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
N4BP2L1
Entrez Gene ID
90634
Gene Name
NEDD4 binding protein 2 like 1
Gene Aliases
CG018
UniGene
Hs.732118
Gene Chromosome Location
Chr.13: 32400723-32429649 on Build GRCh38
Transcripts
NM_001079691.1 NM_001286459.1 NM_001286460.1 NM_001286461.1 NM_052818.2 XM_006719890.3 XM_011535301.2 XM_011535302.2 XM_011535303.2 XM_011535304.2 XM_017020836.1 XM_017020837.1 XM_017020838.1 XM_017020840.1 XM_017020841.1 XM_017020842.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6202070 c.455G>T p.R152M Chr.13: 32404213-32404213 on Build GRCh38
7154270 c.403G>C p.E135Q Chr.13: 32404391-32404391 on Build GRCh38
1158168 c.453C>T p.F151F Chr.13: 32404341-32404341 on Build GRCh38
946892 c.460G>T p.E154* Chr.13: 32404208-32404208 on Build GRCh38
257484 c.417G>T p.K139N Chr.13: 32404251-32404251 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs983747626 Chr.13: 32404124-32404124 on Build GRCh38
rs375051779 Chr.13: 32404440-32404440 on Build GRCh38
rs373017254 Chr.13: 32404333-32404333 on Build GRCh38
rs3210648 Chr.13: 32404371-32404371 on Build GRCh38
rs773200881 Chr.13: 32404228-32404228 on Build GRCh38
rs750976191 Chr.13: 32404295-32404295 on Build GRCh38
rs777550361 Chr.13: 32404387-32404387 on Build GRCh38
rs746991013 Chr.13: 32404400-32404400 on Build GRCh38
rs921597527 Chr.13: 32404201-32404201 on Build GRCh38
rs759717796 Chr.13: 32404545-32404545 on Build GRCh38
rs764485230 Chr.13: 32404260-32404260 on Build GRCh38
rs774777506 Chr.13: 32404428-32404428 on Build GRCh38
rs767428531 Chr.13: 32404238-32404238 on Build GRCh38
rs377189702 Chr.13: 32404259-32404259 on Build GRCh38
rs80025343 Chr.13: 32404352-32404352 on Build GRCh38
rs1062947 Chr.13: 32404373-32404373 on Build GRCh38
rs751984302 Chr.13: 32404263-32404263 on Build GRCh38
rs775057878 Chr.13: 32404166-32404168 on Build GRCh38
rs750184384 Chr.13: 32404312-32404314 on Build GRCh38
rs569741562 Chr.13: 32404172-32404172 on Build GRCh38
rs372082286 Chr.13: 32404434-32404434 on Build GRCh38
rs1001641952 Chr.13: 32404468-32404468 on Build GRCh38
rs569882860 Chr.13: 32404417-32404417 on Build GRCh38
rs762233369 Chr.13: 32404435-32404435 on Build GRCh38
rs1045789 Chr.13: 32404356-32404356 on Build GRCh38
rs1128611 Chr.13: 32404344-32404344 on Build GRCh38
rs567237618 Chr.13: 32404520-32404520 on Build GRCh38
rs180916545 Chr.13: 32404561-32404561 on Build GRCh38
rs778687431 Chr.13: 32404171-32404171 on Build GRCh38
rs753368209 Chr.13: 32404339-32404339 on Build GRCh38
rs1062946 Chr.13: 32404375-32404375 on Build GRCh38
rs754642145 Chr.13: 32404351-32404351 on Build GRCh38
rs771962589 Chr.13: 32404142-32404142 on Build GRCh38
rs909185552 Chr.13: 32404461-32404461 on Build GRCh38
rs756241336 Chr.13: 32404311-32404313 on Build GRCh38
rs941721029 Chr.13: 32404165-32404165 on Build GRCh38
rs752851759 Chr.13: 32404304-32404305 on Build GRCh38
rs747937300 Chr.13: 32404366-32404366 on Build GRCh38
rs760751564 Chr.13: 32404403-32404403 on Build GRCh38
rs200269903 Chr.13: 32404241-32404241 on Build GRCh38
rs771988656 Chr.13: 32404188-32404188 on Build GRCh38
rs973437303 Chr.13: 32404196-32404196 on Build GRCh38
rs957174373 Chr.13: 32404121-32404121 on Build GRCh38
rs989842632 Chr.13: 32404138-32404138 on Build GRCh38
rs11841349 Chr.13: 32404291-32404291 on Build GRCh38
rs372468111 Chr.13: 32404251-32404251 on Build GRCh38
rs377155457 Chr.13: 32404434-32404437 on Build GRCh38
rs908089036 Chr.13: 32404132-32404132 on Build GRCh38
rs372358284 Chr.13: 32404153-32404153 on Build GRCh38
rs762325298 Chr.13: 32404287-32404287 on Build GRCh38
rs768986907 Chr.13: 32404425-32404425 on Build GRCh38
rs748025144 Chr.13: 32404187-32404187 on Build GRCh38
rs1045789 Chr.13: 32404356-32404356 on Build GRCh38
rs764424567 Chr.13: 32404311-32404311 on Build GRCh38
rs890318556 Chr.13: 32404264-32404264 on Build GRCh38
rs534361206 Chr.13: 32404553-32404553 on Build GRCh38
rs941987384 Chr.13: 32404502-32404502 on Build GRCh38
rs754083841 Chr.13: 32404453-32404453 on Build GRCh38
rs931774391 Chr.13: 32404237-32404237 on Build GRCh38
rs755553911 Chr.13: 32404326-32404326 on Build GRCh38
rs1128610 Chr.13: 32404346-32404346 on Build GRCh38
rs80249163 Chr.13: 32404433-32404433 on Build GRCh38

View Full Product Details